Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38685394-38685956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE8
Entrez Gene ID
144402
Gene Name
copine 8
Gene Aliases
-
UniGene
Hs.40910
Gene Chromosome Location
Chr.12: 38652200-38908593 on Build GRCh38
Transcripts
NM_153634.2 XM_017018851.1 XM_017018852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202175 c.1215A>C p.K405N Chr.12: 38685546-38685546 on Build GRCh38
1202174 c.1234C>T p.P412S Chr.12: 38685527-38685527 on Build GRCh38
7163389 c.1243T>A p.F415I Chr.12: 38685518-38685518 on Build GRCh38
2206535 c.1151A>G p.N384S Chr.12: 38685610-38685610 on Build GRCh38
6634711 c.1153C>A p.P385T Chr.12: 38685608-38685608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573296580 Chr.12: 38685634-38685635 on Build GRCh38
rs978258344 Chr.12: 38685625-38685625 on Build GRCh38
rs183270935 Chr.12: 38685544-38685544 on Build GRCh38
rs1022340780 Chr.12: 38685565-38685565 on Build GRCh38
rs769038121 Chr.12: 38685742-38685742 on Build GRCh38
rs775338705 Chr.12: 38685813-38685815 on Build GRCh38
rs768031708 Chr.12: 38685451-38685451 on Build GRCh38
rs755854274 Chr.12: 38685468-38685468 on Build GRCh38
rs541704501 Chr.12: 38685627-38685627 on Build GRCh38
rs772403462 Chr.12: 38685677-38685677 on Build GRCh38
rs766578041 Chr.12: 38685597-38685597 on Build GRCh38
rs776970840 Chr.12: 38685775-38685775 on Build GRCh38
rs760818241 Chr.12: 38685458-38685458 on Build GRCh38
rs573272903 Chr.12: 38685903-38685903 on Build GRCh38
rs755168060 Chr.12: 38685479-38685479 on Build GRCh38
rs199991967 Chr.12: 38685557-38685557 on Build GRCh38
rs1012569459 Chr.12: 38685754-38685754 on Build GRCh38
rs767508731 Chr.12: 38685635-38685635 on Build GRCh38
rs755990175 Chr.12: 38685657-38685657 on Build GRCh38
rs889240471 Chr.12: 38685554-38685554 on Build GRCh38
rs774613687 Chr.12: 38685558-38685558 on Build GRCh38
rs201967260 Chr.12: 38685453-38685453 on Build GRCh38
rs1022818069 Chr.12: 38685899-38685899 on Build GRCh38
rs568834083 Chr.12: 38685662-38685662 on Build GRCh38
rs903027833 Chr.12: 38685673-38685673 on Build GRCh38
rs78596070 Chr.12: 38685765-38685765 on Build GRCh38
rs1026666397 Chr.12: 38685445-38685445 on Build GRCh38
rs374379534 Chr.12: 38685542-38685542 on Build GRCh38
rs1035235569 Chr.12: 38685914-38685914 on Build GRCh38
rs759704790 Chr.12: 38685603-38685603 on Build GRCh38
rs754097312 Chr.12: 38685464-38685464 on Build GRCh38
rs3759140 Chr.12: 38685706-38685706 on Build GRCh38
rs773827247 Chr.12: 38685590-38685590 on Build GRCh38
rs570415553 Chr.12: 38685429-38685429 on Build GRCh38
rs561869190 Chr.12: 38685930-38685930 on Build GRCh38
rs546377605 Chr.12: 38685792-38685792 on Build GRCh38
rs373045704 Chr.12: 38685442-38685442 on Build GRCh38
rs371442242 Chr.12: 38685469-38685469 on Build GRCh38
rs146995905 Chr.12: 38685908-38685908 on Build GRCh38
rs3834473 Chr.12: 38685758-38685758 on Build GRCh38
rs890265974 Chr.12: 38685800-38685800 on Build GRCh38
rs757044788 Chr.12: 38685631-38685631 on Build GRCh38
rs751325087 Chr.12: 38685486-38685486 on Build GRCh38
rs900507498 Chr.12: 38685884-38685884 on Build GRCh38
rs187679996 Chr.12: 38685708-38685708 on Build GRCh38
rs761852709 Chr.12: 38685793-38685793 on Build GRCh38
rs748836600 Chr.12: 38685635-38685639 on Build GRCh38
rs991031127 Chr.12: 38685697-38685697 on Build GRCh38
rs1050411106 Chr.12: 38685472-38685472 on Build GRCh38
rs550433849 Chr.12: 38685419-38685419 on Build GRCh38
rs147852781 Chr.12: 38685446-38685446 on Build GRCh38
rs777430861 Chr.12: 38685484-38685484 on Build GRCh38
rs769290248 Chr.12: 38685534-38685534 on Build GRCh38
rs757164412 Chr.12: 38685497-38685497 on Build GRCh38
rs192772318 Chr.12: 38685808-38685808 on Build GRCh38
rs144807766 Chr.12: 38685773-38685776 on Build GRCh38
rs781083875 Chr.12: 38685528-38685528 on Build GRCh38
rs944814168 Chr.12: 38685653-38685653 on Build GRCh38
rs1022583747 Chr.12: 38685758-38685758 on Build GRCh38
rs773097368 Chr.12: 38685825-38685825 on Build GRCh38
rs1008805799 Chr.12: 38685455-38685455 on Build GRCh38
rs745420513 Chr.12: 38685532-38685532 on Build GRCh38
rs374257875 Chr.12: 38685845-38685845 on Build GRCh38
rs1010888262 Chr.12: 38685894-38685894 on Build GRCh38
rs765449118 Chr.12: 38685620-38685620 on Build GRCh38
rs760891922 Chr.12: 38685596-38685596 on Build GRCh38
rs201874856 Chr.12: 38685579-38685579 on Build GRCh38
rs776780548 Chr.12: 38685600-38685600 on Build GRCh38
rs981222507 Chr.12: 38685821-38685821 on Build GRCh38
rs748500905 Chr.12: 38685575-38685575 on Build GRCh38
rs967299608 Chr.12: 38685614-38685614 on Build GRCh38
rs750348072 Chr.12: 38685645-38685645 on Build GRCh38
rs766751770 Chr.12: 38685463-38685463 on Build GRCh38
rs989573392 Chr.12: 38685511-38685511 on Build GRCh38
rs1040828641 Chr.12: 38685671-38685671 on Build GRCh38
rs939631559 Chr.12: 38685695-38685695 on Build GRCh38
rs79114072 Chr.12: 38685718-38685718 on Build GRCh38

View Full Product Details