Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8065997-8066523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3AR1
Entrez Gene ID
719
Gene Name
complement component 3a receptor 1
Gene Aliases
AZ3B, C3AR, HNFAG09
UniGene
Hs.504656
Gene Chromosome Location
Chr.12: 8058323-8066359 on Build GRCh38
Transcripts
NM_001326475.1 NM_001326477.1 NM_004054.3

SNP Detail

SNP ID Chromosome Location
rs948387313 Chr.12: 8066050-8066050 on Build GRCh38
rs765118736 Chr.12: 8066221-8066221 on Build GRCh38
rs561378145 Chr.12: 8066139-8066144 on Build GRCh38
rs1034392373 Chr.12: 8066398-8066398 on Build GRCh38
rs1003938570 Chr.12: 8066058-8066058 on Build GRCh38
rs1021597281 Chr.12: 8066254-8066254 on Build GRCh38
rs900967701 Chr.12: 8066085-8066085 on Build GRCh38
rs758538720 Chr.12: 8066326-8066326 on Build GRCh38
rs778747429 Chr.12: 8066425-8066425 on Build GRCh38
rs969140063 Chr.12: 8066261-8066261 on Build GRCh38
rs200437789 Chr.12: 8066061-8066062 on Build GRCh38
rs998028885 Chr.12: 8066089-8066089 on Build GRCh38
rs762942106 Chr.12: 8066120-8066120 on Build GRCh38
rs992996812 Chr.12: 8066312-8066312 on Build GRCh38
rs1048640463 Chr.12: 8066349-8066349 on Build GRCh38
rs1015779120 Chr.12: 8066081-8066081 on Build GRCh38
rs934027787 Chr.12: 8066145-8066145 on Build GRCh38
rs374175548 Chr.12: 8066198-8066198 on Build GRCh38
rs531617244 Chr.12: 8066053-8066053 on Build GRCh38
rs11567784 Chr.12: 8066142-8066142 on Build GRCh38
rs986115512 Chr.12: 8066203-8066203 on Build GRCh38
rs1024030252 Chr.12: 8066351-8066351 on Build GRCh38
rs764043109 Chr.12: 8066357-8066357 on Build GRCh38
rs370403393 Chr.12: 8066269-8066269 on Build GRCh38
rs1050164108 Chr.12: 8066227-8066227 on Build GRCh38
rs953964811 Chr.12: 8066166-8066166 on Build GRCh38
rs891996043 Chr.12: 8066343-8066343 on Build GRCh38
rs949002465 Chr.12: 8066334-8066334 on Build GRCh38
rs1036751654 Chr.12: 8066414-8066414 on Build GRCh38
rs773193657 Chr.12: 8066410-8066413 on Build GRCh38
rs939686498 Chr.12: 8066408-8066408 on Build GRCh38
rs11567785 Chr.12: 8066057-8066057 on Build GRCh38
rs748699641 Chr.12: 8066399-8066401 on Build GRCh38
rs374074556 Chr.12: 8066303-8066303 on Build GRCh38
rs984740446 Chr.12: 8066410-8066411 on Build GRCh38
rs890049599 Chr.12: 8066281-8066281 on Build GRCh38
rs527304207 Chr.12: 8066189-8066189 on Build GRCh38
rs938844430 Chr.12: 8066311-8066311 on Build GRCh38
rs979804678 Chr.12: 8066370-8066370 on Build GRCh38
rs547259614 Chr.12: 8066475-8066475 on Build GRCh38
rs894765258 Chr.12: 8066020-8066020 on Build GRCh38
rs146982786 Chr.12: 8066055-8066055 on Build GRCh38
rs541996969 Chr.12: 8066395-8066395 on Build GRCh38
rs765568857 Chr.12: 8066143-8066143 on Build GRCh38
rs900955012 Chr.12: 8066424-8066424 on Build GRCh38
rs766765318 Chr.12: 8066492-8066492 on Build GRCh38
rs888098151 Chr.12: 8066364-8066364 on Build GRCh38

View Full Product Details