Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100204858-100205325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR6
Entrez Gene ID
64431
Gene Name
ARP6 actin-related protein 6 homolog
Gene Aliases
ARP6, CDA12, HSPC281, MSTP136, hARP6, hARPX
UniGene
Hs.115088
Gene Chromosome Location
Chr.12: 100200087-100224425 on Build GRCh38
Transcripts
NM_022496.4 NR_048568.1 NR_048569.1 XM_017019821.1 NR_135073.1

Gene Symbol
DEPDC4
Entrez Gene ID
120863
Gene Name
DEP domain containing 4
Gene Aliases
DEP.4
UniGene
Hs.653118
Gene Chromosome Location
Chr.12: 100203669-100268094 on Build GRCh38
Transcripts
NM_022496.4 NR_048568.1 NR_048569.1 XM_017019821.1 NR_135073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5638862 c.70G>A p.V24I Chr.12: 100204941-100204941 on Build GRCh38
268576 c.162C>A p.L54L Chr.12: 100205033-100205033 on Build GRCh38
4829760 c.122C>A p.T41N Chr.12: 100204993-100204993 on Build GRCh38
1747321 c.172C>T p.L58F Chr.12: 100205043-100205043 on Build GRCh38
6036919 c.92G>A p.R31Q Chr.12: 100204963-100204963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758150828 Chr.12: 100205013-100205013 on Build GRCh38
rs766532903 Chr.12: 100205213-100205213 on Build GRCh38
rs768030046 Chr.12: 100205036-100205036 on Build GRCh38
rs202026087 Chr.12: 100204941-100204941 on Build GRCh38
rs571190790 Chr.12: 100205244-100205244 on Build GRCh38
rs546543126 Chr.12: 100205209-100205209 on Build GRCh38
rs776974514 Chr.12: 100204891-100204891 on Build GRCh38
rs762289056 Chr.12: 100204893-100204893 on Build GRCh38
rs567305636 Chr.12: 100205145-100205145 on Build GRCh38
rs533239449 Chr.12: 100204888-100204888 on Build GRCh38
rs534438742 Chr.12: 100205195-100205195 on Build GRCh38
rs765573548 Chr.12: 100204896-100204896 on Build GRCh38
rs369354926 Chr.12: 100205001-100205001 on Build GRCh38
rs745605106 Chr.12: 100205004-100205004 on Build GRCh38
rs943664445 Chr.12: 100205100-100205100 on Build GRCh38
rs1019079191 Chr.12: 100205276-100205276 on Build GRCh38
rs773491494 Chr.12: 100204901-100204901 on Build GRCh38
rs202129245 Chr.12: 100205079-100205079 on Build GRCh38
rs759812024 Chr.12: 100204889-100204890 on Build GRCh38
rs767632802 Chr.12: 100204890-100204891 on Build GRCh38
rs773579312 Chr.12: 100205086-100205086 on Build GRCh38
rs771171280 Chr.12: 100205098-100205098 on Build GRCh38
rs769239726 Chr.12: 100204890-100204890 on Build GRCh38
rs772372420 Chr.12: 100204978-100204978 on Build GRCh38
rs766529937 Chr.12: 100204924-100204924 on Build GRCh38
rs1040724267 Chr.12: 100205138-100205138 on Build GRCh38
rs540106593 Chr.12: 100205264-100205264 on Build GRCh38
rs746457585 Chr.12: 100204889-100204889 on Build GRCh38
rs764129838 Chr.12: 100204977-100204977 on Build GRCh38
rs763166222 Chr.12: 100204913-100204913 on Build GRCh38
rs904012187 Chr.12: 100205247-100205247 on Build GRCh38
rs747971263 Chr.12: 100204962-100204962 on Build GRCh38
rs757171838 Chr.12: 100204994-100204994 on Build GRCh38
rs200199803 Chr.12: 100205070-100205070 on Build GRCh38
rs912923625 Chr.12: 100205162-100205162 on Build GRCh38
rs746571745 Chr.12: 100205022-100205022 on Build GRCh38
rs752882205 Chr.12: 100204899-100204899 on Build GRCh38
rs563139606 Chr.12: 100205045-100205045 on Build GRCh38
rs779535184 Chr.12: 100205021-100205021 on Build GRCh38
rs149793338 Chr.12: 100205040-100205040 on Build GRCh38
rs945609085 Chr.12: 100205182-100205182 on Build GRCh38
rs188726229 Chr.12: 100205292-100205292 on Build GRCh38
rs1042687814 Chr.12: 100205217-100205217 on Build GRCh38
rs763253380 Chr.12: 100205090-100205090 on Build GRCh38

View Full Product Details