Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89352206-89352802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP6
Entrez Gene ID
1848
Gene Name
dual specificity phosphatase 6
Gene Aliases
HH19, MKP3, PYST1
UniGene
Hs.298654
Gene Chromosome Location
Chr.12: 89347825-89352859 on Build GRCh38
Transcripts
NM_001946.3 NM_022652.3

SNP Detail

SNP ID Chromosome Location
rs189094409 Chr.12: 89352379-89352379 on Build GRCh38
rs981713651 Chr.12: 89352578-89352578 on Build GRCh38
rs566014700 Chr.12: 89352688-89352688 on Build GRCh38
rs991680122 Chr.12: 89352527-89352527 on Build GRCh38
rs192148001 Chr.12: 89352429-89352429 on Build GRCh38
rs543873089 Chr.12: 89352331-89352331 on Build GRCh38
rs966892763 Chr.12: 89352271-89352271 on Build GRCh38
rs927031715 Chr.12: 89352551-89352551 on Build GRCh38
rs1027559997 Chr.12: 89352238-89352238 on Build GRCh38
rs1048890557 Chr.12: 89352570-89352570 on Build GRCh38
rs998302319 Chr.12: 89352450-89352450 on Build GRCh38
rs762984683 Chr.12: 89352516-89352516 on Build GRCh38
rs563195334 Chr.12: 89352704-89352704 on Build GRCh38
rs539217017 Chr.12: 89352662-89352662 on Build GRCh38
rs912920816 Chr.12: 89352526-89352526 on Build GRCh38
rs949806515 Chr.12: 89352344-89352344 on Build GRCh38
rs1015687828 Chr.12: 89352245-89352245 on Build GRCh38
rs574573165 Chr.12: 89352266-89352266 on Build GRCh38
rs991752060 Chr.12: 89352305-89352305 on Build GRCh38
rs17016713 Chr.12: 89352671-89352671 on Build GRCh38
rs183833024 Chr.12: 89352332-89352332 on Build GRCh38
rs926207447 Chr.12: 89352313-89352313 on Build GRCh38
rs754701115 Chr.12: 89352703-89352703 on Build GRCh38
rs1045872811 Chr.12: 89352351-89352351 on Build GRCh38
rs369920146 Chr.12: 89352712-89352712 on Build GRCh38
rs772457203 Chr.12: 89352273-89352273 on Build GRCh38
rs897190958 Chr.12: 89352624-89352624 on Build GRCh38
rs981474201 Chr.12: 89352723-89352723 on Build GRCh38
rs766337953 Chr.12: 89352592-89352592 on Build GRCh38
rs961448955 Chr.12: 89352270-89352270 on Build GRCh38
rs79053845 Chr.12: 89352355-89352355 on Build GRCh38
rs1023125434 Chr.12: 89352561-89352561 on Build GRCh38
rs998592931 Chr.12: 89352653-89352653 on Build GRCh38
rs971457988 Chr.12: 89352534-89352534 on Build GRCh38
rs932722585 Chr.12: 89352617-89352617 on Build GRCh38
rs902176219 Chr.12: 89352497-89352497 on Build GRCh38
rs765005164 Chr.12: 89352392-89352392 on Build GRCh38
rs17016710 Chr.12: 89352535-89352535 on Build GRCh38
rs567001636 Chr.12: 89352350-89352351 on Build GRCh38
rs760935219 Chr.12: 89352368-89352368 on Build GRCh38
rs750201612 Chr.12: 89352408-89352408 on Build GRCh38
rs988403057 Chr.12: 89352334-89352334 on Build GRCh38
rs549805853 Chr.12: 89352577-89352577 on Build GRCh38
rs552491007 Chr.12: 89352685-89352685 on Build GRCh38
rs1021075904 Chr.12: 89352482-89352482 on Build GRCh38
rs1020601976 Chr.12: 89352269-89352269 on Build GRCh38
rs919988162 Chr.12: 89352596-89352598 on Build GRCh38
rs554568236 Chr.12: 89352734-89352734 on Build GRCh38
rs919915976 Chr.12: 89352739-89352739 on Build GRCh38
rs897248513 Chr.12: 89352439-89352439 on Build GRCh38
rs141042035 Chr.12: 89352455-89352455 on Build GRCh38
rs765239006 Chr.12: 89352286-89352287 on Build GRCh38
rs888482227 Chr.12: 89352242-89352242 on Build GRCh38
rs754420906 Chr.12: 89352602-89352602 on Build GRCh38
rs565177000 Chr.12: 89352414-89352414 on Build GRCh38
rs546544311 Chr.12: 89352693-89352693 on Build GRCh38
rs898342542 Chr.12: 89352409-89352409 on Build GRCh38
rs1033233738 Chr.12: 89352476-89352476 on Build GRCh38
rs985712229 Chr.12: 89352283-89352283 on Build GRCh38
rs972089560 Chr.12: 89352575-89352575 on Build GRCh38
rs912391239 Chr.12: 89352659-89352659 on Build GRCh38
rs1029805409 Chr.12: 89352276-89352276 on Build GRCh38
rs954164360 Chr.12: 89352747-89352747 on Build GRCh38
rs151076733 Chr.12: 89352226-89352226 on Build GRCh38
rs961694110 Chr.12: 89352733-89352733 on Build GRCh38
rs980281237 Chr.12: 89352750-89352750 on Build GRCh38
rs1049045138 Chr.12: 89352370-89352370 on Build GRCh38
rs1030154871 Chr.12: 89352454-89352454 on Build GRCh38
rs1006975876 Chr.12: 89352475-89352475 on Build GRCh38
rs187052148 Chr.12: 89352637-89352637 on Build GRCh38
rs554674631 Chr.12: 89352243-89352243 on Build GRCh38
rs575824258 Chr.12: 89352250-89352250 on Build GRCh38
rs560840124 Chr.12: 89352484-89352484 on Build GRCh38
rs543062381 Chr.12: 89352479-89352479 on Build GRCh38
rs1051193811 Chr.12: 89352664-89352664 on Build GRCh38
rs775794238 Chr.12: 89352342-89352342 on Build GRCh38
rs1010516522 Chr.12: 89352700-89352700 on Build GRCh38
rs943994057 Chr.12: 89352665-89352665 on Build GRCh38
rs376343537 Chr.12: 89352680-89352680 on Build GRCh38
rs1036668124 Chr.12: 89352595-89352595 on Build GRCh38
rs957517018 Chr.12: 89352699-89352699 on Build GRCh38
rs981877883 Chr.12: 89352343-89352343 on Build GRCh38
rs957491339 Chr.12: 89352287-89352287 on Build GRCh38
rs796691930 Chr.12: 89352351-89352351 on Build GRCh38
rs751099361 Chr.12: 89352513-89352513 on Build GRCh38
rs937158988 Chr.12: 89352559-89352559 on Build GRCh38
rs780836918 Chr.12: 89352674-89352674 on Build GRCh38
rs1032064387 Chr.12: 89352517-89352517 on Build GRCh38
rs534721824 Chr.12: 89352732-89352732 on Build GRCh38
rs760626772 Chr.12: 89352281-89352281 on Build GRCh38
rs554969209 Chr.12: 89352749-89352749 on Build GRCh38
rs1007026869 Chr.12: 89352683-89352683 on Build GRCh38
rs894924927 Chr.12: 89352695-89352695 on Build GRCh38
rs909140786 Chr.12: 89352583-89352583 on Build GRCh38

View Full Product Details