Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16050687-16051170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKB
Entrez Gene ID
1188
Gene Name
chloride voltage-gated channel Kb
Gene Aliases
CLCKB, ClC-K2, ClC-Kb
UniGene
Hs.352243
Gene Chromosome Location
Chr.1: 16043736-16057326 on Build GRCh38
Transcripts
NM_000085.4 NM_001165945.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381255 c.1119C>A p.N373K Chr.1: 16050940-16050940 on Build GRCh38
5917095 c.673C>T p.P225S Chr.1: 16051001-16051001 on Build GRCh38
4523117 c.1199G>A p.G400E Chr.1: 16051020-16051020 on Build GRCh38
4765062 c.642_643insC p.Q216fs*66 Chr.1: 16050970-16050971 on Build GRCh38
6385010 c.1058C>T p.S353F Chr.1: 16050879-16050879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748987694 Chr.1: 16050912-16050912 on Build GRCh38
rs763173537 Chr.1: 16051023-16051023 on Build GRCh38
rs1051767497 Chr.1: 16051022-16051022 on Build GRCh38
rs777020165 Chr.1: 16051054-16051054 on Build GRCh38
rs372390371 Chr.1: 16050759-16050759 on Build GRCh38
rs775949416 Chr.1: 16050959-16050959 on Build GRCh38
rs769053704 Chr.1: 16051049-16051049 on Build GRCh38
rs764159319 Chr.1: 16050967-16050967 on Build GRCh38
rs753799104 Chr.1: 16051082-16051082 on Build GRCh38
rs372482510 Chr.1: 16050845-16050845 on Build GRCh38
rs569630570 Chr.1: 16050797-16050797 on Build GRCh38
rs369375366 Chr.1: 16050958-16050958 on Build GRCh38
rs34255952 Chr.1: 16051004-16051004 on Build GRCh38
rs891000534 Chr.1: 16051040-16051040 on Build GRCh38
rs201479763 Chr.1: 16051059-16051059 on Build GRCh38
rs368794099 Chr.1: 16050758-16050758 on Build GRCh38
rs757343183 Chr.1: 16050970-16050970 on Build GRCh38
rs777146902 Chr.1: 16050851-16050851 on Build GRCh38
rs149762502 Chr.1: 16051103-16051104 on Build GRCh38
rs2275165 Chr.1: 16051126-16051126 on Build GRCh38
rs752344124 Chr.1: 16051099-16051099 on Build GRCh38
rs771693877 Chr.1: 16050941-16050941 on Build GRCh38
rs988333756 Chr.1: 16050755-16050755 on Build GRCh38
rs13306237 Chr.1: 16050982-16050982 on Build GRCh38
rs771370918 Chr.1: 16051010-16051021 on Build GRCh38
rs1045254484 Chr.1: 16050766-16050766 on Build GRCh38
rs60943704 Chr.1: 16051109-16051110 on Build GRCh38
rs147999316 Chr.1: 16051005-16051005 on Build GRCh38
rs918198207 Chr.1: 16050760-16050761 on Build GRCh38
rs151266668 Chr.1: 16050913-16050913 on Build GRCh38
rs565896323 Chr.1: 16050846-16050846 on Build GRCh38
rs763270686 Chr.1: 16050962-16050962 on Build GRCh38
rs12093430 Chr.1: 16051071-16051071 on Build GRCh38
rs368997387 Chr.1: 16050807-16050807 on Build GRCh38
rs761275599 Chr.1: 16050847-16050847 on Build GRCh38
rs779851182 Chr.1: 16050981-16050981 on Build GRCh38
rs897827928 Chr.1: 16050806-16050806 on Build GRCh38
rs762559176 Chr.1: 16050831-16050831 on Build GRCh38
rs545117992 Chr.1: 16051119-16051119 on Build GRCh38
rs755992420 Chr.1: 16050980-16050980 on Build GRCh38
rs112933556 Chr.1: 16050789-16050789 on Build GRCh38
rs1008435493 Chr.1: 16050840-16050840 on Build GRCh38
rs374911075 Chr.1: 16050867-16050867 on Build GRCh38
rs949665265 Chr.1: 16050972-16050972 on Build GRCh38
rs373351140 Chr.1: 16051051-16051051 on Build GRCh38
rs751676745 Chr.1: 16050968-16050968 on Build GRCh38
rs775187299 Chr.1: 16050828-16050828 on Build GRCh38
rs200272823 Chr.1: 16050997-16050997 on Build GRCh38
rs777345240 Chr.1: 16050946-16050946 on Build GRCh38
rs754672425 Chr.1: 16050992-16050992 on Build GRCh38
rs767602749 Chr.1: 16051056-16051056 on Build GRCh38
rs765694706 Chr.1: 16050860-16050860 on Build GRCh38
rs369520464 Chr.1: 16051103-16051103 on Build GRCh38
rs538743005 Chr.1: 16050811-16050811 on Build GRCh38
rs140705060 Chr.1: 16050901-16050901 on Build GRCh38
rs942137627 Chr.1: 16050796-16050796 on Build GRCh38
rs369169829 Chr.1: 16051013-16051013 on Build GRCh38
rs764256848 Chr.1: 16050875-16050875 on Build GRCh38
rs773916073 Chr.1: 16050925-16050925 on Build GRCh38
rs185451420 Chr.1: 16050868-16050868 on Build GRCh38
rs549793997 Chr.1: 16050736-16050736 on Build GRCh38
rs949556066 Chr.1: 16050763-16050763 on Build GRCh38
rs766728474 Chr.1: 16050850-16050850 on Build GRCh38
rs1001135349 Chr.1: 16051102-16051102 on Build GRCh38
rs935036505 Chr.1: 16050737-16050737 on Build GRCh38
rs769451791 Chr.1: 16050825-16050825 on Build GRCh38
rs779798035 Chr.1: 16050908-16050908 on Build GRCh38
rs760610564 Chr.1: 16051069-16051069 on Build GRCh38
rs755939381 Chr.1: 16050907-16050907 on Build GRCh38
rs778485688 Chr.1: 16050993-16050993 on Build GRCh38
rs569936954 Chr.1: 16050812-16050812 on Build GRCh38
rs760090728 Chr.1: 16050947-16050947 on Build GRCh38
rs201245211 Chr.1: 16050892-16050892 on Build GRCh38
rs201333658 Chr.1: 16051100-16051101 on Build GRCh38
rs768258386 Chr.1: 16050837-16050837 on Build GRCh38
rs767551505 Chr.1: 16050974-16050974 on Build GRCh38
rs754795852 Chr.1: 16051083-16051083 on Build GRCh38
rs751558214 Chr.1: 16050883-16050883 on Build GRCh38
rs760033582 Chr.1: 16050852-16050852 on Build GRCh38
rs888026091 Chr.1: 16050826-16050826 on Build GRCh38
rs781053314 Chr.1: 16050899-16050899 on Build GRCh38
rs201840674 Chr.1: 16050933-16050933 on Build GRCh38
rs778739554 Chr.1: 16051096-16051096 on Build GRCh38
rs35258749 Chr.1: 16050975-16050975 on Build GRCh38
rs45573234 Chr.1: 16050975-16050975 on Build GRCh38

View Full Product Details