Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28200844-28201442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC8
Entrez Gene ID
22826
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Gene Aliases
HSPC331, SPF31
UniGene
Hs.433540
Gene Chromosome Location
Chr.1: 28200278-28233031 on Build GRCh38
Transcripts
NM_014280.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5557855 c.711A>G p.K237K Chr.1: 28201299-28201299 on Build GRCh38
7247575 c.640-7T>A p.? Chr.1: 28201377-28201377 on Build GRCh38
280544 c.643A>C p.S215R Chr.1: 28201367-28201367 on Build GRCh38
6332902 c.730C>A p.P244T Chr.1: 28201280-28201280 on Build GRCh38
3487765 c.699G>A p.K233K Chr.1: 28201311-28201311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761918013 Chr.1: 28201339-28201339 on Build GRCh38
rs769205621 Chr.1: 28201321-28201321 on Build GRCh38
rs772566038 Chr.1: 28201305-28201305 on Build GRCh38
rs1802449 Chr.1: 28200984-28200984 on Build GRCh38
rs531182164 Chr.1: 28201208-28201208 on Build GRCh38
rs956940837 Chr.1: 28201010-28201010 on Build GRCh38
rs765612789 Chr.1: 28201346-28201346 on Build GRCh38
rs35570191 Chr.1: 28201160-28201161 on Build GRCh38
rs754951090 Chr.1: 28201223-28201223 on Build GRCh38
rs1031619989 Chr.1: 28201005-28201005 on Build GRCh38
rs9447 Chr.1: 28200900-28200900 on Build GRCh38
rs781343972 Chr.1: 28201231-28201231 on Build GRCh38
rs965847466 Chr.1: 28200992-28200992 on Build GRCh38
rs1004757217 Chr.1: 28200969-28200969 on Build GRCh38
rs915511868 Chr.1: 28201052-28201052 on Build GRCh38
rs771688424 Chr.1: 28200972-28200972 on Build GRCh38
rs762318992 Chr.1: 28201377-28201378 on Build GRCh38
rs766578064 Chr.1: 28201391-28201391 on Build GRCh38
rs187868657 Chr.1: 28201294-28201294 on Build GRCh38
rs530029122 Chr.1: 28201331-28201331 on Build GRCh38
rs944472320 Chr.1: 28201173-28201173 on Build GRCh38
rs757522302 Chr.1: 28201210-28201210 on Build GRCh38
rs773537189 Chr.1: 28201237-28201237 on Build GRCh38
rs927697971 Chr.1: 28201083-28201083 on Build GRCh38
rs201063813 Chr.1: 28201222-28201222 on Build GRCh38
rs989715419 Chr.1: 28201051-28201051 on Build GRCh38
rs8678 Chr.1: 28201143-28201143 on Build GRCh38
rs367997548 Chr.1: 28201417-28201417 on Build GRCh38
rs576452091 Chr.1: 28201228-28201228 on Build GRCh38
rs756105567 Chr.1: 28201402-28201402 on Build GRCh38
rs919106138 Chr.1: 28201171-28201171 on Build GRCh38
rs778607449 Chr.1: 28201413-28201413 on Build GRCh38
rs763483655 Chr.1: 28201315-28201315 on Build GRCh38
rs111458452 Chr.1: 28201406-28201406 on Build GRCh38
rs200079350 Chr.1: 28201269-28201269 on Build GRCh38
rs564024593 Chr.1: 28201204-28201204 on Build GRCh38
rs368080312 Chr.1: 28201256-28201256 on Build GRCh38
rs768853922 Chr.1: 28201330-28201330 on Build GRCh38
rs17163970 Chr.1: 28200914-28200914 on Build GRCh38
rs753949516 Chr.1: 28201206-28201206 on Build GRCh38
rs775911040 Chr.1: 28201314-28201314 on Build GRCh38
rs552202690 Chr.1: 28200924-28200924 on Build GRCh38
rs939139442 Chr.1: 28201098-28201098 on Build GRCh38
rs561902825 Chr.1: 28201295-28201295 on Build GRCh38
rs779067310 Chr.1: 28200930-28200930 on Build GRCh38
rs754226248 Chr.1: 28201202-28201203 on Build GRCh38
rs35039225 Chr.1: 28201377-28201377 on Build GRCh38
rs114065436 Chr.1: 28201002-28201002 on Build GRCh38
rs568855403 Chr.1: 28200980-28200980 on Build GRCh38
rs764524776 Chr.1: 28201202-28201202 on Build GRCh38
rs777396368 Chr.1: 28201403-28201403 on Build GRCh38
rs200774904 Chr.1: 28201217-28201217 on Build GRCh38
rs764515963 Chr.1: 28201199-28201202 on Build GRCh38
rs138121284 Chr.1: 28201071-28201071 on Build GRCh38
rs770294979 Chr.1: 28201174-28201174 on Build GRCh38
rs1049274 Chr.1: 28201322-28201322 on Build GRCh38
rs143936244 Chr.1: 28201094-28201094 on Build GRCh38
rs770661212 Chr.1: 28201275-28201275 on Build GRCh38
rs925579536 Chr.1: 28201101-28201101 on Build GRCh38
rs1040147258 Chr.1: 28201320-28201320 on Build GRCh38
rs370942935 Chr.1: 28201246-28201246 on Build GRCh38
rs956864396 Chr.1: 28201104-28201104 on Build GRCh38
rs777642919 Chr.1: 28201247-28201247 on Build GRCh38
rs546835925 Chr.1: 28200888-28200888 on Build GRCh38
rs748098606 Chr.1: 28201233-28201233 on Build GRCh38
rs746557361 Chr.1: 28201044-28201044 on Build GRCh38
rs748847741 Chr.1: 28201254-28201254 on Build GRCh38
rs912945523 Chr.1: 28201159-28201159 on Build GRCh38
rs191748539 Chr.1: 28201058-28201058 on Build GRCh38
rs184086871 Chr.1: 28201193-28201193 on Build GRCh38
rs147272528 Chr.1: 28201116-28201116 on Build GRCh38
rs139274022 Chr.1: 28201126-28201126 on Build GRCh38
rs750524396 Chr.1: 28201354-28201354 on Build GRCh38
rs560390631 Chr.1: 28201398-28201398 on Build GRCh38
rs887590997 Chr.1: 28200936-28200936 on Build GRCh38

View Full Product Details