Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110029268-110029785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13A
Entrez Gene ID
88455
Gene Name
ankyrin repeat domain 13A
Gene Aliases
ANKRD13, NY-REN-25
UniGene
Hs.528703
Gene Chromosome Location
Chr.12: 109999169-110039432 on Build GRCh38
Transcripts
NM_033121.1 XM_005253980.2 XM_005253981.3 XM_005253982.3 XM_005253984.1 XM_011538937.1 XM_011538938.2 XM_017020157.1 XM_017020158.1 XM_017020159.1 XM_017020160.1 XM_017020161.1 XM_017020162.1 XM_017020163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5507866 c.1222C>T p.P408S Chr.12: 110029623-110029623 on Build GRCh38
934906 c.1160G>A p.R387Q Chr.12: 110029561-110029561 on Build GRCh38
3980753 c.1167T>G p.S389R Chr.12: 110029568-110029568 on Build GRCh38
6282420 c.1196T>C p.I399T Chr.12: 110029597-110029597 on Build GRCh38
278837 c.1164G>A p.T388T Chr.12: 110029565-110029565 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766349615 Chr.12: 110029644-110029644 on Build GRCh38
rs758997511 Chr.12: 110029530-110029530 on Build GRCh38
rs992313512 Chr.12: 110029567-110029567 on Build GRCh38
rs565247205 Chr.12: 110029305-110029305 on Build GRCh38
rs765416553 Chr.12: 110029513-110029513 on Build GRCh38
rs373804069 Chr.12: 110029560-110029560 on Build GRCh38
rs564377050 Chr.12: 110029408-110029408 on Build GRCh38
rs1047500750 Chr.12: 110029330-110029330 on Build GRCh38
rs377451135 Chr.12: 110029669-110029669 on Build GRCh38
rs748752939 Chr.12: 110029571-110029571 on Build GRCh38
rs774392967 Chr.12: 110029586-110029586 on Build GRCh38
rs975768242 Chr.12: 110029709-110029709 on Build GRCh38
rs773457423 Chr.12: 110029637-110029637 on Build GRCh38
rs775362594 Chr.12: 110029617-110029617 on Build GRCh38
rs931733893 Chr.12: 110029747-110029747 on Build GRCh38
rs1025004067 Chr.12: 110029461-110029461 on Build GRCh38
rs772641974 Chr.12: 110029428-110029428 on Build GRCh38
rs367703492 Chr.12: 110029564-110029564 on Build GRCh38
rs766643944 Chr.12: 110029704-110029704 on Build GRCh38
rs907815239 Chr.12: 110029675-110029675 on Build GRCh38
rs961723843 Chr.12: 110029697-110029697 on Build GRCh38
rs750473015 Chr.12: 110029520-110029520 on Build GRCh38
rs61940911 Chr.12: 110029339-110029339 on Build GRCh38
rs559102605 Chr.12: 110029307-110029307 on Build GRCh38
rs943237793 Chr.12: 110029762-110029762 on Build GRCh38
rs749004666 Chr.12: 110029430-110029430 on Build GRCh38
rs755427453 Chr.12: 110029561-110029561 on Build GRCh38
rs762887689 Chr.12: 110029448-110029448 on Build GRCh38
rs745782030 Chr.12: 110029599-110029599 on Build GRCh38
rs182050000 Chr.12: 110029648-110029648 on Build GRCh38
rs765507638 Chr.12: 110029626-110029626 on Build GRCh38
rs770837475 Chr.12: 110029720-110029720 on Build GRCh38
rs1006298742 Chr.12: 110029473-110029473 on Build GRCh38
rs774042457 Chr.12: 110029458-110029458 on Build GRCh38
rs766870223 Chr.12: 110029533-110029533 on Build GRCh38
rs1014039629 Chr.12: 110029441-110029441 on Build GRCh38
rs767937571 Chr.12: 110029670-110029670 on Build GRCh38
rs770555876 Chr.12: 110029441-110029445 on Build GRCh38
rs372319311 Chr.12: 110029568-110029568 on Build GRCh38
rs770313128 Chr.12: 110029572-110029572 on Build GRCh38
rs753007415 Chr.12: 110029679-110029679 on Build GRCh38
rs767197481 Chr.12: 110029475-110029475 on Build GRCh38
rs529913834 Chr.12: 110029734-110029734 on Build GRCh38
rs778371129 Chr.12: 110029455-110029456 on Build GRCh38
rs189748288 Chr.12: 110029312-110029312 on Build GRCh38
rs1052824595 Chr.12: 110029347-110029347 on Build GRCh38
rs762126665 Chr.12: 110029488-110029488 on Build GRCh38
rs576977206 Chr.12: 110029625-110029625 on Build GRCh38
rs755705630 Chr.12: 110029565-110029565 on Build GRCh38
rs771435129 Chr.12: 110029682-110029683 on Build GRCh38
rs755373408 Chr.12: 110029663-110029663 on Build GRCh38
rs771973183 Chr.12: 110029600-110029600 on Build GRCh38
rs747802155 Chr.12: 110029562-110029562 on Build GRCh38
rs755791451 Chr.12: 110029680-110029680 on Build GRCh38
rs745388390 Chr.12: 110029683-110029696 on Build GRCh38
rs892464432 Chr.12: 110029374-110029374 on Build GRCh38
rs759413867 Chr.12: 110029463-110029463 on Build GRCh38
rs763135045 Chr.12: 110029641-110029641 on Build GRCh38
rs559242747 Chr.12: 110029698-110029698 on Build GRCh38
rs969469946 Chr.12: 110029489-110029489 on Build GRCh38
rs776973378 Chr.12: 110029481-110029481 on Build GRCh38
rs36049730 Chr.12: 110029439-110029440 on Build GRCh38
rs922991866 Chr.12: 110029712-110029712 on Build GRCh38

View Full Product Details