Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44218321-44218847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMAP1
Entrez Gene ID
55929
Gene Name
DNA methyltransferase 1 associated protein 1
Gene Aliases
DNMAP1, DNMTAP1, EAF2, MEAF2, SWC4
UniGene
Hs.8008
Gene Chromosome Location
Chr.1: 44213428-44220679 on Build GRCh38
Transcripts
NM_001034023.1 NM_001034024.1 NM_019100.4 XM_005271037.4 XM_005271039.1 XM_005271040.1 XM_006710771.3 XM_011541784.2 XM_011541785.2 XM_017001807.1 XM_017001808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6326796 c.553C>T p.H185Y Chr.1: 44218392-44218392 on Build GRCh38
3930793 c.716T>G p.L239R Chr.1: 44218673-44218673 on Build GRCh38
5901024 c.739G>A p.G247R Chr.1: 44218696-44218696 on Build GRCh38
6645148 c.773G>A p.R258H Chr.1: 44218730-44218730 on Build GRCh38
4927698 c.441C>T p.L147L Chr.1: 44218358-44218358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147271334 Chr.1: 44218654-44218654 on Build GRCh38
rs748658570 Chr.1: 44218544-44218544 on Build GRCh38
rs187902214 Chr.1: 44218433-44218433 on Build GRCh38
rs144462164 Chr.1: 44218457-44218457 on Build GRCh38
rs968979370 Chr.1: 44218556-44218556 on Build GRCh38
rs775655920 Chr.1: 44218682-44218682 on Build GRCh38
rs964044718 Chr.1: 44218530-44218530 on Build GRCh38
rs747249971 Chr.1: 44218393-44218394 on Build GRCh38
rs35249619 Chr.1: 44218701-44218701 on Build GRCh38
rs769065111 Chr.1: 44218359-44218359 on Build GRCh38
rs368524464 Chr.1: 44218741-44218741 on Build GRCh38
rs764864892 Chr.1: 44218709-44218709 on Build GRCh38
rs779646178 Chr.1: 44218363-44218363 on Build GRCh38
rs778216027 Chr.1: 44218569-44218569 on Build GRCh38
rs748510377 Chr.1: 44218444-44218444 on Build GRCh38
rs748765935 Chr.1: 44218343-44218343 on Build GRCh38
rs898444637 Chr.1: 44218419-44218419 on Build GRCh38
rs778241048 Chr.1: 44218648-44218648 on Build GRCh38
rs767278967 Chr.1: 44218398-44218398 on Build GRCh38
rs774579658 Chr.1: 44218460-44218460 on Build GRCh38
rs775360259 Chr.1: 44218591-44218591 on Build GRCh38
rs372548904 Chr.1: 44218418-44218418 on Build GRCh38
rs764742862 Chr.1: 44218758-44218758 on Build GRCh38
rs756913898 Chr.1: 44218514-44218514 on Build GRCh38
rs1056895210 Chr.1: 44218659-44218659 on Build GRCh38
rs781590748 Chr.1: 44218547-44218547 on Build GRCh38
rs368742800 Chr.1: 44218410-44218410 on Build GRCh38
rs757527244 Chr.1: 44218655-44218655 on Build GRCh38
rs760508897 Chr.1: 44218606-44218606 on Build GRCh38
rs187803864 Chr.1: 44218553-44218553 on Build GRCh38
rs764893235 Chr.1: 44218621-44218621 on Build GRCh38
rs756647722 Chr.1: 44218647-44218647 on Build GRCh38
rs762679185 Chr.1: 44218712-44218712 on Build GRCh38
rs747268999 Chr.1: 44218624-44218624 on Build GRCh38
rs755291277 Chr.1: 44218408-44218408 on Build GRCh38
rs762027853 Chr.1: 44218362-44218362 on Build GRCh38
rs770115089 Chr.1: 44218555-44218555 on Build GRCh38
rs369079971 Chr.1: 44218571-44218571 on Build GRCh38
rs749397742 Chr.1: 44218535-44218535 on Build GRCh38
rs773374215 Chr.1: 44218454-44218454 on Build GRCh38
rs767381622 Chr.1: 44218485-44218485 on Build GRCh38
rs759735232 Chr.1: 44218583-44218583 on Build GRCh38
rs766415972 Chr.1: 44218381-44218381 on Build GRCh38
rs899384322 Chr.1: 44218765-44218765 on Build GRCh38
rs367767883 Chr.1: 44218536-44218536 on Build GRCh38
rs763741160 Chr.1: 44218612-44218612 on Build GRCh38
rs192276687 Chr.1: 44218446-44218446 on Build GRCh38
rs772018540 Chr.1: 44218590-44218590 on Build GRCh38
rs185871556 Chr.1: 44218486-44218486 on Build GRCh38
rs140090594 Chr.1: 44218637-44218637 on Build GRCh38
rs150342757 Chr.1: 44218388-44218388 on Build GRCh38
rs781441767 Chr.1: 44218429-44218431 on Build GRCh38
rs748797447 Chr.1: 44218413-44218413 on Build GRCh38
rs760424165 Chr.1: 44218490-44218490 on Build GRCh38
rs772541742 Chr.1: 44218346-44218346 on Build GRCh38
rs746022722 Chr.1: 44218802-44218802 on Build GRCh38
rs771712127 Chr.1: 44218770-44218770 on Build GRCh38
rs770254055 Chr.1: 44218626-44218631 on Build GRCh38
rs199594206 Chr.1: 44218421-44218421 on Build GRCh38
rs779267606 Chr.1: 44218791-44218791 on Build GRCh38
rs376072861 Chr.1: 44218679-44218679 on Build GRCh38
rs754220119 Chr.1: 44218766-44218766 on Build GRCh38
rs138626857 Chr.1: 44218412-44218412 on Build GRCh38
rs781579736 Chr.1: 44218435-44218435 on Build GRCh38
rs772683814 Chr.1: 44218812-44218812 on Build GRCh38
rs776362349 Chr.1: 44218705-44218705 on Build GRCh38
rs753543846 Chr.1: 44218501-44218501 on Build GRCh38
rs372465567 Chr.1: 44218574-44218574 on Build GRCh38
rs544496296 Chr.1: 44218646-44218646 on Build GRCh38
rs1012570208 Chr.1: 44218702-44218702 on Build GRCh38
rs749903843 Chr.1: 44218622-44218622 on Build GRCh38
rs769155056 Chr.1: 44218426-44218426 on Build GRCh38
rs745436055 Chr.1: 44218764-44218764 on Build GRCh38
rs367917027 Chr.1: 44218361-44218361 on Build GRCh38
rs749690011 Chr.1: 44218652-44218652 on Build GRCh38
rs772103921 Chr.1: 44218680-44218680 on Build GRCh38
rs201828316 Chr.1: 44218414-44218414 on Build GRCh38
rs141586055 Chr.1: 44218592-44218592 on Build GRCh38
rs763166810 Chr.1: 44218380-44218380 on Build GRCh38
rs779322512 Chr.1: 44218661-44218661 on Build GRCh38
rs111713256 Chr.1: 44218349-44218349 on Build GRCh38
rs151219508 Chr.1: 44218370-44218370 on Build GRCh38
rs751108364 Chr.1: 44218739-44218739 on Build GRCh38
rs142415325 Chr.1: 44218394-44218394 on Build GRCh38
rs771313523 Chr.1: 44218579-44218579 on Build GRCh38
rs919828221 Chr.1: 44218716-44218716 on Build GRCh38
rs773901665 Chr.1: 44218705-44218706 on Build GRCh38
rs947256201 Chr.1: 44218614-44218614 on Build GRCh38
rs997258908 Chr.1: 44218532-44218532 on Build GRCh38
rs774528208 Chr.1: 44218581-44218581 on Build GRCh38
rs375445340 Chr.1: 44218478-44218478 on Build GRCh38
rs755464308 Chr.1: 44218519-44218519 on Build GRCh38
rs746856748 Chr.1: 44218696-44218696 on Build GRCh38
rs192410775 Chr.1: 44218706-44218706 on Build GRCh38
rs867684794 Chr.1: 44218534-44218534 on Build GRCh38
rs921168667 Chr.1: 44218608-44218608 on Build GRCh38
rs376305922 Chr.1: 44218730-44218730 on Build GRCh38
rs755747117 Chr.1: 44218411-44218411 on Build GRCh38
rs1043259224 Chr.1: 44218615-44218615 on Build GRCh38
rs772907285 Chr.1: 44218711-44218711 on Build GRCh38
rs977548813 Chr.1: 44218572-44218573 on Build GRCh38
rs762447948 Chr.1: 44218746-44218746 on Build GRCh38
rs375398371 Chr.1: 44218493-44218493 on Build GRCh38
rs757766717 Chr.1: 44218772-44218772 on Build GRCh38
rs931241626 Chr.1: 44218742-44218742 on Build GRCh38
rs370762869 Chr.1: 44218523-44218523 on Build GRCh38
rs943372569 Chr.1: 44218546-44218546 on Build GRCh38
rs540607515 Chr.1: 44218813-44218813 on Build GRCh38
rs757912210 Chr.1: 44218628-44218628 on Build GRCh38
rs778325844 Chr.1: 44218516-44218516 on Build GRCh38
rs765409416 Chr.1: 44218364-44218364 on Build GRCh38
rs747366744 Chr.1: 44218356-44218356 on Build GRCh38
rs560510137 Chr.1: 44218819-44218819 on Build GRCh38
rs761415756 Chr.1: 44218616-44218616 on Build GRCh38

View Full Product Details