Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40984533-40985070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001905.3 NR_125440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1602390 c.306G>A p.R102R Chr.1: 40984960-40984960 on Build GRCh38
6063551 c.246C>G p.L82L Chr.1: 40984900-40984900 on Build GRCh38
5735417 c.248G>A p.R83Q Chr.1: 40984881-40984881 on Build GRCh38
4675442 c.325A>T p.K109* Chr.1: 40984979-40984979 on Build GRCh38
5773290 c.190G>A p.E64K Chr.1: 40984823-40984823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765840007 Chr.1: 40984959-40984959 on Build GRCh38
rs751260098 Chr.1: 40984875-40984875 on Build GRCh38
rs777081020 Chr.1: 40984906-40984906 on Build GRCh38
rs761124469 Chr.1: 40984808-40984808 on Build GRCh38
rs372087700 Chr.1: 40984802-40984802 on Build GRCh38
rs777790111 Chr.1: 40985019-40985019 on Build GRCh38
rs1008571617 Chr.1: 40985020-40985020 on Build GRCh38
rs201994529 Chr.1: 40984876-40984876 on Build GRCh38
rs986628487 Chr.1: 40984558-40984558 on Build GRCh38
rs769195447 Chr.1: 40984929-40984929 on Build GRCh38
rs938949732 Chr.1: 40984732-40984732 on Build GRCh38
rs772827860 Chr.1: 40984794-40984794 on Build GRCh38
rs376970699 Chr.1: 40984896-40984896 on Build GRCh38
rs756834548 Chr.1: 40985027-40985027 on Build GRCh38
rs374103645 Chr.1: 40985007-40985007 on Build GRCh38
rs369866440 Chr.1: 40984651-40984651 on Build GRCh38
rs748131892 Chr.1: 40984895-40984895 on Build GRCh38
rs774085033 Chr.1: 40984920-40984920 on Build GRCh38
rs925195448 Chr.1: 40984618-40984618 on Build GRCh38
rs754557223 Chr.1: 40984765-40984765 on Build GRCh38
rs765474888 Chr.1: 40984837-40984837 on Build GRCh38
rs773103801 Chr.1: 40984963-40984963 on Build GRCh38
rs925193846 Chr.1: 40985006-40985006 on Build GRCh38
rs188902675 Chr.1: 40984721-40984721 on Build GRCh38
rs559679792 Chr.1: 40984629-40984629 on Build GRCh38
rs548657642 Chr.1: 40984710-40984710 on Build GRCh38
rs552263230 Chr.1: 40984628-40984628 on Build GRCh38
rs1047189199 Chr.1: 40984998-40984998 on Build GRCh38
rs745961668 Chr.1: 40985036-40985038 on Build GRCh38
rs200144652 Chr.1: 40984815-40984815 on Build GRCh38
rs943761223 Chr.1: 40984666-40984666 on Build GRCh38
rs766634874 Chr.1: 40984863-40984863 on Build GRCh38
rs747349849 Chr.1: 40984921-40984921 on Build GRCh38
rs150868456 Chr.1: 40984966-40984966 on Build GRCh38
rs112398640 Chr.1: 40984648-40984648 on Build GRCh38
rs992854556 Chr.1: 40984893-40984893 on Build GRCh38
rs750649864 Chr.1: 40984842-40984842 on Build GRCh38
rs752494489 Chr.1: 40985005-40985005 on Build GRCh38
rs761967231 Chr.1: 40984817-40984817 on Build GRCh38
rs201019829 Chr.1: 40985038-40985038 on Build GRCh38
rs770622124 Chr.1: 40984800-40984800 on Build GRCh38
rs762480558 Chr.1: 40984939-40984939 on Build GRCh38
rs778411782 Chr.1: 40985031-40985031 on Build GRCh38
rs78630393 Chr.1: 40984717-40984717 on Build GRCh38
rs967373645 Chr.1: 40984674-40984674 on Build GRCh38
rs967797072 Chr.1: 40984958-40984958 on Build GRCh38
rs896795332 Chr.1: 40984718-40984718 on Build GRCh38
rs1027669458 Chr.1: 40984740-40984740 on Build GRCh38
rs945337203 Chr.1: 40984581-40984581 on Build GRCh38
rs758478895 Chr.1: 40984847-40984847 on Build GRCh38
rs748844510 Chr.1: 40984911-40984911 on Build GRCh38
rs754744599 Chr.1: 40984984-40984984 on Build GRCh38
rs964066441 Chr.1: 40984621-40984621 on Build GRCh38
rs764658979 Chr.1: 40984813-40984813 on Build GRCh38
rs767357701 Chr.1: 40984999-40984999 on Build GRCh38
rs754438957 Chr.1: 40984816-40984816 on Build GRCh38
rs748708761 Chr.1: 40985021-40985021 on Build GRCh38
rs143597296 Chr.1: 40984576-40984576 on Build GRCh38
rs780693225 Chr.1: 40984771-40984771 on Build GRCh38
rs978402727 Chr.1: 40984595-40984595 on Build GRCh38
rs936506892 Chr.1: 40984645-40984645 on Build GRCh38
rs770443898 Chr.1: 40984915-40984915 on Build GRCh38
rs143501659 Chr.1: 40984882-40984882 on Build GRCh38
rs769333191 Chr.1: 40984775-40984775 on Build GRCh38
rs191736269 Chr.1: 40984795-40984795 on Build GRCh38
rs183200970 Chr.1: 40984553-40984553 on Build GRCh38
rs751846370 Chr.1: 40984980-40984980 on Build GRCh38
rs367835354 Chr.1: 40984676-40984676 on Build GRCh38
rs554198167 Chr.1: 40985042-40985042 on Build GRCh38
rs777269285 Chr.1: 40984934-40984934 on Build GRCh38
rs766098132 Chr.1: 40984586-40984586 on Build GRCh38

View Full Product Details