Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32365879-32366442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSDC1
Entrez Gene ID
55108
Gene Name
BSD domain containing 1
Gene Aliases
-
UniGene
Hs.353454
Gene Chromosome Location
Chr.1: 32364633-32394461 on Build GRCh38
Transcripts
NM_001143888.2 NM_001143889.2 NM_001143890.2 NM_001300958.1 NM_018045.7 NR_125352.1

SNP Detail

SNP ID Chromosome Location
rs576928164 Chr.1: 32366249-32366249 on Build GRCh38
rs944552369 Chr.1: 32366132-32366132 on Build GRCh38
rs476153 Chr.1: 32365942-32365942 on Build GRCh38
rs957483539 Chr.1: 32366076-32366076 on Build GRCh38
rs764986437 Chr.1: 32366330-32366330 on Build GRCh38
rs938936254 Chr.1: 32366051-32366051 on Build GRCh38
rs1024435556 Chr.1: 32366385-32366385 on Build GRCh38
rs187812170 Chr.1: 32365899-32365899 on Build GRCh38
rs187991738 Chr.1: 32366357-32366357 on Build GRCh38
rs558741156 Chr.1: 32366239-32366239 on Build GRCh38
rs1019911702 Chr.1: 32366123-32366123 on Build GRCh38
rs767446752 Chr.1: 32366389-32366389 on Build GRCh38
rs7539731 Chr.1: 32366084-32366084 on Build GRCh38
rs1004580942 Chr.1: 32365974-32365974 on Build GRCh38
rs1043196515 Chr.1: 32366068-32366068 on Build GRCh38
rs201769400 Chr.1: 32366199-32366202 on Build GRCh38
rs542580846 Chr.1: 32366360-32366360 on Build GRCh38
rs34410839 Chr.1: 32366161-32366162 on Build GRCh38
rs553483969 Chr.1: 32366174-32366174 on Build GRCh38
rs921847364 Chr.1: 32366175-32366175 on Build GRCh38
rs192341448 Chr.1: 32365905-32365905 on Build GRCh38
rs916155856 Chr.1: 32366322-32366322 on Build GRCh38
rs559417190 Chr.1: 32366297-32366297 on Build GRCh38
rs1032975866 Chr.1: 32366060-32366060 on Build GRCh38
rs963823415 Chr.1: 32365902-32365902 on Build GRCh38
rs200387251 Chr.1: 32366198-32366200 on Build GRCh38
rs988878548 Chr.1: 32366083-32366083 on Build GRCh38
rs766432003 Chr.1: 32365954-32365954 on Build GRCh38
rs948902601 Chr.1: 32366350-32366351 on Build GRCh38
rs1014536221 Chr.1: 32365994-32365994 on Build GRCh38
rs1048671698 Chr.1: 32365951-32365951 on Build GRCh38
rs548451147 Chr.1: 32366299-32366300 on Build GRCh38
rs980099469 Chr.1: 32366032-32366032 on Build GRCh38
rs998961629 Chr.1: 32366364-32366366 on Build GRCh38
rs1057497930 Chr.1: 32366054-32366054 on Build GRCh38
rs774938212 Chr.1: 32365921-32365921 on Build GRCh38
rs1024990249 Chr.1: 32366002-32366002 on Build GRCh38
rs999284822 Chr.1: 32366077-32366077 on Build GRCh38
rs1053589249 Chr.1: 32366371-32366371 on Build GRCh38
rs535870621 Chr.1: 32366015-32366015 on Build GRCh38
rs1013054186 Chr.1: 32366061-32366061 on Build GRCh38
rs554136938 Chr.1: 32366045-32366045 on Build GRCh38
rs1036507899 Chr.1: 32366250-32366250 on Build GRCh38
rs183491996 Chr.1: 32366259-32366259 on Build GRCh38
rs760937557 Chr.1: 32366238-32366238 on Build GRCh38
rs41265847 Chr.1: 32365982-32365982 on Build GRCh38
rs573239350 Chr.1: 32366280-32366280 on Build GRCh38
rs954842640 Chr.1: 32366096-32366096 on Build GRCh38
rs908216167 Chr.1: 32366292-32366292 on Build GRCh38
rs1008165098 Chr.1: 32366121-32366121 on Build GRCh38
rs897296189 Chr.1: 32365983-32365983 on Build GRCh38
rs536704300 Chr.1: 32366173-32366173 on Build GRCh38
rs377550059 Chr.1: 32366286-32366286 on Build GRCh38
rs764140118 Chr.1: 32366130-32366130 on Build GRCh38
rs931866171 Chr.1: 32366235-32366235 on Build GRCh38
rs890464984 Chr.1: 32366395-32366395 on Build GRCh38
rs900390088 Chr.1: 32365910-32365910 on Build GRCh38
rs940467391 Chr.1: 32366247-32366247 on Build GRCh38
rs906119854 Chr.1: 32366409-32366409 on Build GRCh38

View Full Product Details