Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109250964-109251562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR2
Entrez Gene ID
1952
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Gene Aliases
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3
UniGene
Hs.57652
Gene Chromosome Location
Chr.1: 109249705-109275756 on Build GRCh38
Transcripts
NM_001408.2 XM_005270580.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5900442 c.1132C>T p.P378S Chr.1: 109251211-109251211 on Build GRCh38
7286374 c.1423C>T p.R475* Chr.1: 109251502-109251502 on Build GRCh38
3933991 c.941C>T p.T314M Chr.1: 109251020-109251020 on Build GRCh38
7165848 c.1031C>T p.P344L Chr.1: 109251110-109251110 on Build GRCh38
1294672 c.1265C>T p.T422I Chr.1: 109251344-109251344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369951311 Chr.1: 109251278-109251278 on Build GRCh38
rs755549032 Chr.1: 109251177-109251177 on Build GRCh38
rs746126393 Chr.1: 109251384-109251384 on Build GRCh38
rs769734269 Chr.1: 109251216-109251216 on Build GRCh38
rs375455011 Chr.1: 109251291-109251291 on Build GRCh38
rs927757225 Chr.1: 109251133-109251133 on Build GRCh38
rs952303398 Chr.1: 109251048-109251048 on Build GRCh38
rs573558774 Chr.1: 109251286-109251286 on Build GRCh38
rs748320682 Chr.1: 109251112-109251112 on Build GRCh38
rs369703876 Chr.1: 109251391-109251391 on Build GRCh38
rs778686097 Chr.1: 109251503-109251503 on Build GRCh38
rs377036999 Chr.1: 109251375-109251375 on Build GRCh38
rs771660911 Chr.1: 109251531-109251531 on Build GRCh38
rs41279704 Chr.1: 109251501-109251501 on Build GRCh38
rs749274117 Chr.1: 109251424-109251436 on Build GRCh38
rs149311467 Chr.1: 109251229-109251229 on Build GRCh38
rs887956763 Chr.1: 109251404-109251404 on Build GRCh38
rs775721854 Chr.1: 109251460-109251460 on Build GRCh38
rs759949577 Chr.1: 109251348-109251348 on Build GRCh38
rs780591973 Chr.1: 109251167-109251167 on Build GRCh38
rs764686331 Chr.1: 109251134-109251134 on Build GRCh38
rs768494078 Chr.1: 109251410-109251410 on Build GRCh38
rs571436385 Chr.1: 109251130-109251130 on Build GRCh38
rs768813176 Chr.1: 109251089-109251089 on Build GRCh38
rs774591844 Chr.1: 109251106-109251106 on Build GRCh38
rs200447397 Chr.1: 109251272-109251272 on Build GRCh38
rs773624411 Chr.1: 109251113-109251113 on Build GRCh38
rs760425869 Chr.1: 109251428-109251428 on Build GRCh38
rs760119454 Chr.1: 109251376-109251376 on Build GRCh38
rs1012205619 Chr.1: 109251273-109251273 on Build GRCh38
rs753199359 Chr.1: 109251355-109251355 on Build GRCh38
rs775461482 Chr.1: 109251220-109251220 on Build GRCh38
rs756465634 Chr.1: 109251153-109251153 on Build GRCh38
rs528285800 Chr.1: 109251025-109251025 on Build GRCh38
rs193224402 Chr.1: 109251223-109251223 on Build GRCh38
rs911446385 Chr.1: 109251489-109251489 on Build GRCh38
rs199829910 Chr.1: 109251210-109251218 on Build GRCh38
rs748247760 Chr.1: 109251187-109251187 on Build GRCh38
rs146721276 Chr.1: 109251108-109251108 on Build GRCh38
rs373566231 Chr.1: 109251338-109251338 on Build GRCh38
rs981761180 Chr.1: 109251120-109251120 on Build GRCh38
rs370246986 Chr.1: 109251455-109251455 on Build GRCh38
rs749186325 Chr.1: 109251409-109251409 on Build GRCh38
rs550918065 Chr.1: 109251350-109251350 on Build GRCh38
rs201416867 Chr.1: 109251199-109251199 on Build GRCh38
rs567653065 Chr.1: 109251206-109251206 on Build GRCh38
rs748146968 Chr.1: 109251510-109251510 on Build GRCh38
rs780906021 Chr.1: 109251287-109251287 on Build GRCh38
rs766137017 Chr.1: 109251123-109251123 on Build GRCh38
rs368408692 Chr.1: 109251080-109251080 on Build GRCh38
rs373714726 Chr.1: 109251416-109251416 on Build GRCh38
rs747213922 Chr.1: 109251282-109251282 on Build GRCh38
rs755453775 Chr.1: 109251266-109251266 on Build GRCh38
rs779843956 Chr.1: 109251497-109251497 on Build GRCh38
rs758092786 Chr.1: 109251037-109251037 on Build GRCh38
rs769640761 Chr.1: 109251296-109251296 on Build GRCh38
rs961341501 Chr.1: 109251110-109251110 on Build GRCh38
rs776440581 Chr.1: 109251212-109251212 on Build GRCh38
rs751233523 Chr.1: 109251150-109251150 on Build GRCh38
rs772880054 Chr.1: 109251537-109251537 on Build GRCh38
rs971668527 Chr.1: 109251486-109251486 on Build GRCh38
rs773068309 Chr.1: 109251346-109251346 on Build GRCh38
rs772978368 Chr.1: 109251420-109251420 on Build GRCh38
rs377605875 Chr.1: 109251453-109251453 on Build GRCh38
rs760504883 Chr.1: 109251115-109251115 on Build GRCh38
rs768176557 Chr.1: 109251331-109251331 on Build GRCh38
rs754271169 Chr.1: 109251264-109251264 on Build GRCh38
rs374336247 Chr.1: 109251021-109251021 on Build GRCh38
rs747244877 Chr.1: 109251202-109251202 on Build GRCh38
rs775463779 Chr.1: 109251325-109251325 on Build GRCh38
rs934845086 Chr.1: 109251066-109251066 on Build GRCh38
rs764494127 Chr.1: 109251471-109251471 on Build GRCh38
rs773741912 Chr.1: 109251337-109251337 on Build GRCh38
rs759400632 Chr.1: 109251214-109251214 on Build GRCh38
rs141506583 Chr.1: 109251415-109251415 on Build GRCh38
rs201185292 Chr.1: 109251491-109251491 on Build GRCh38
rs148805303 Chr.1: 109251223-109251224 on Build GRCh38
rs142614123 Chr.1: 109251351-109251351 on Build GRCh38
rs758860471 Chr.1: 109251356-109251356 on Build GRCh38
rs753831092 Chr.1: 109251017-109251017 on Build GRCh38
rs371403541 Chr.1: 109251228-109251228 on Build GRCh38
rs375029971 Chr.1: 109251145-109251145 on Build GRCh38
rs756506681 Chr.1: 109251070-109251070 on Build GRCh38
rs752306754 Chr.1: 109251035-109251035 on Build GRCh38
rs773071253 Chr.1: 109250988-109250988 on Build GRCh38
rs781355873 Chr.1: 109251045-109251045 on Build GRCh38
rs200405611 Chr.1: 109251477-109251477 on Build GRCh38
rs746250775 Chr.1: 109251051-109251051 on Build GRCh38
rs573063472 Chr.1: 109251013-109251013 on Build GRCh38
rs149911274 Chr.1: 109251176-109251176 on Build GRCh38
rs763786482 Chr.1: 109251148-109251148 on Build GRCh38
rs766334537 Chr.1: 109250999-109250999 on Build GRCh38
rs146832064 Chr.1: 109251394-109251394 on Build GRCh38
rs764166933 Chr.1: 109251371-109251371 on Build GRCh38
rs367687072 Chr.1: 109251200-109251200 on Build GRCh38
rs944709395 Chr.1: 109251301-109251301 on Build GRCh38
rs746593465 Chr.1: 109251539-109251539 on Build GRCh38
rs192199164 Chr.1: 109251128-109251128 on Build GRCh38
rs752173128 Chr.1: 109251476-109251476 on Build GRCh38
rs868134638 Chr.1: 109251057-109251057 on Build GRCh38
rs781385956 Chr.1: 109251381-109251381 on Build GRCh38
rs199581862 Chr.1: 109251465-109251465 on Build GRCh38
rs926059478 Chr.1: 109251368-109251368 on Build GRCh38
rs202054206 Chr.1: 109251186-109251186 on Build GRCh38
rs760461083 Chr.1: 109250998-109250998 on Build GRCh38
rs749847773 Chr.1: 109251088-109251088 on Build GRCh38
rs773248244 Chr.1: 109251024-109251026 on Build GRCh38
rs372637628 Chr.1: 109251496-109251496 on Build GRCh38
rs371574420 Chr.1: 109251138-109251138 on Build GRCh38
rs745646802 Chr.1: 109251290-109251290 on Build GRCh38
rs779560542 Chr.1: 109251267-109251267 on Build GRCh38

View Full Product Details