Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100872794-100873320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL2
Entrez Gene ID
2135
Gene Name
exostosin like glycosyltransferase 2
Gene Aliases
EXTR2
UniGene
Hs.357637
Gene Chromosome Location
Chr.1: 100872372-100895411 on Build GRCh38
Transcripts
NM_001033025.2 NM_001261440.1 NM_001261441.1 NM_001261442.1 NM_001439.3 NR_048570.1 XM_005270621.1 XM_011540994.2 XM_011540995.2 XM_011540996.2 XM_017000651.1

SNP Detail

SNP ID Chromosome Location
rs529247033 Chr.1: 100873035-100873035 on Build GRCh38
rs562865469 Chr.1: 100873194-100873194 on Build GRCh38
rs982529859 Chr.1: 100873184-100873184 on Build GRCh38
rs999579802 Chr.1: 100872889-100872889 on Build GRCh38
rs1034826791 Chr.1: 100873142-100873142 on Build GRCh38
rs975040137 Chr.1: 100873235-100873235 on Build GRCh38
rs893785805 Chr.1: 100872923-100872923 on Build GRCh38
rs531441928 Chr.1: 100872888-100872888 on Build GRCh38
rs932213936 Chr.1: 100873252-100873252 on Build GRCh38
rs76577854 Chr.1: 100873112-100873112 on Build GRCh38
rs560345180 Chr.1: 100873074-100873074 on Build GRCh38
rs561882067 Chr.1: 100872994-100872994 on Build GRCh38
rs556000633 Chr.1: 100872829-100872829 on Build GRCh38
rs778316313 Chr.1: 100873186-100873186 on Build GRCh38
rs765465718 Chr.1: 100872911-100872911 on Build GRCh38
rs988134858 Chr.1: 100873256-100873256 on Build GRCh38
rs758795330 Chr.1: 100873156-100873156 on Build GRCh38
rs558087289 Chr.1: 100872939-100872939 on Build GRCh38
rs905109117 Chr.1: 100873086-100873086 on Build GRCh38
rs911774459 Chr.1: 100873051-100873051 on Build GRCh38
rs576058058 Chr.1: 100872902-100872902 on Build GRCh38
rs117273279 Chr.1: 100872817-100872817 on Build GRCh38
rs577955733 Chr.1: 100872976-100872976 on Build GRCh38
rs1016621507 Chr.1: 100873228-100873228 on Build GRCh38
rs951303393 Chr.1: 100873160-100873160 on Build GRCh38
rs3188491 Chr.1: 100872926-100872926 on Build GRCh38
rs752052073 Chr.1: 100873195-100873195 on Build GRCh38
rs543174391 Chr.1: 100873134-100873134 on Build GRCh38
rs540519782 Chr.1: 100873025-100873025 on Build GRCh38
rs756810480 Chr.1: 100873236-100873236 on Build GRCh38
rs531785762 Chr.1: 100873232-100873232 on Build GRCh38

View Full Product Details