Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117869494-117870072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP2
Entrez Gene ID
54834
Gene Name
ganglioside induced differentiation associated protein 2
Gene Aliases
MACROD3
UniGene
Hs.310809
Gene Chromosome Location
Chr.1: 117860227-117930042 on Build GRCh38
Transcripts
NM_017686.3 XM_005270959.2 XM_011541623.2 XM_017001531.1 XM_017001532.1

SNP Detail

SNP ID Chromosome Location
rs185525560 Chr.1: 117869908-117869908 on Build GRCh38
rs145261191 Chr.1: 117869745-117869745 on Build GRCh38
rs926737071 Chr.1: 117869953-117869953 on Build GRCh38
rs912211772 Chr.1: 117870033-117870034 on Build GRCh38
rs759718780 Chr.1: 117869721-117869721 on Build GRCh38
rs970186119 Chr.1: 117869534-117869534 on Build GRCh38
rs964249648 Chr.1: 117869546-117869546 on Build GRCh38
rs774289618 Chr.1: 117869693-117869693 on Build GRCh38
rs149176485 Chr.1: 117870030-117870030 on Build GRCh38
rs937217253 Chr.1: 117869700-117869700 on Build GRCh38
rs370753517 Chr.1: 117869981-117869981 on Build GRCh38
rs377708186 Chr.1: 117869801-117869801 on Build GRCh38
rs576514064 Chr.1: 117869708-117869708 on Build GRCh38
rs527448957 Chr.1: 117869866-117869866 on Build GRCh38
rs900913504 Chr.1: 117869871-117869871 on Build GRCh38
rs113584373 Chr.1: 117869773-117869773 on Build GRCh38
rs560292157 Chr.1: 117869857-117869857 on Build GRCh38
rs751047089 Chr.1: 117869674-117869674 on Build GRCh38
rs141306023 Chr.1: 117869701-117869701 on Build GRCh38
rs1018865918 Chr.1: 117869544-117869544 on Build GRCh38
rs942467293 Chr.1: 117869748-117869748 on Build GRCh38
rs572485080 Chr.1: 117869654-117869654 on Build GRCh38
rs988571242 Chr.1: 117869610-117869610 on Build GRCh38
rs1032578523 Chr.1: 117869909-117869909 on Build GRCh38
rs1005858578 Chr.1: 117869920-117869920 on Build GRCh38
rs781635745 Chr.1: 117870042-117870042 on Build GRCh38
rs928650057 Chr.1: 117869633-117869633 on Build GRCh38
rs956678906 Chr.1: 117869600-117869600 on Build GRCh38
rs780881239 Chr.1: 117869859-117869859 on Build GRCh38
rs958510941 Chr.1: 117869972-117869972 on Build GRCh38
rs890844754 Chr.1: 117869830-117869830 on Build GRCh38
rs960098747 Chr.1: 117869957-117869957 on Build GRCh38
rs956962562 Chr.1: 117869910-117869910 on Build GRCh38
rs562790994 Chr.1: 117869750-117869750 on Build GRCh38
rs980117840 Chr.1: 117869549-117869549 on Build GRCh38
rs764149844 Chr.1: 117869769-117869780 on Build GRCh38
rs973894562 Chr.1: 117869977-117869977 on Build GRCh38
rs370796920 Chr.1: 117869636-117869636 on Build GRCh38
rs913193366 Chr.1: 117870024-117870024 on Build GRCh38
rs566990406 Chr.1: 117869918-117869918 on Build GRCh38
rs147690093 Chr.1: 117869924-117869924 on Build GRCh38
rs919865008 Chr.1: 117869702-117869702 on Build GRCh38
rs189233404 Chr.1: 117869946-117869946 on Build GRCh38
rs979678023 Chr.1: 117869560-117869560 on Build GRCh38
rs114205518 Chr.1: 117869692-117869692 on Build GRCh38
rs41276590 Chr.1: 117869901-117869901 on Build GRCh38
rs1032629383 Chr.1: 117869580-117869580 on Build GRCh38
rs757969986 Chr.1: 117870032-117870032 on Build GRCh38
rs979382809 Chr.1: 117870040-117870040 on Build GRCh38
rs41276592 Chr.1: 117869942-117869942 on Build GRCh38
rs112858106 Chr.1: 117869821-117869821 on Build GRCh38
rs773374888 Chr.1: 117869605-117869605 on Build GRCh38
rs533249616 Chr.1: 117869781-117869781 on Build GRCh38
rs988771424 Chr.1: 117869683-117869683 on Build GRCh38
rs749488156 Chr.1: 117869615-117869615 on Build GRCh38
rs899994026 Chr.1: 117869860-117869860 on Build GRCh38
rs753479888 Chr.1: 117869896-117869896 on Build GRCh38
rs550296179 Chr.1: 117869831-117869831 on Build GRCh38
rs969595166 Chr.1: 117869926-117869926 on Build GRCh38

View Full Product Details