Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155725337-155725896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAP3
Entrez Gene ID
7818
Gene Name
death associated protein 3
Gene Aliases
DAP-3, MRP-S29, MRPS29, bMRP-10
UniGene
Hs.516746
Gene Chromosome Location
Chr.1: 155688104-155739010 on Build GRCh38
Transcripts
NM_001199849.1 NM_001199850.1 NM_001199851.1 NM_004632.3 NM_033657.2 XM_005245480.2 XM_005245481.2 XM_005245482.4 XM_011509972.2 XM_017002289.1 XM_017002290.1 XM_017002291.1 XM_017002292.1 XM_017002293.1 XM_017002294.1 XM_017002295.1 XM_017002296.1 XM_017002297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3418124 c.368G>A p.R123Q Chr.1: 155725479-155725479 on Build GRCh38
896971 c.272T>C p.V91A Chr.1: 155725383-155725383 on Build GRCh38
1600928 c.275A>T p.K92M Chr.1: 155725386-155725386 on Build GRCh38
3788793 c.358C>G p.P120A Chr.1: 155725469-155725469 on Build GRCh38
1491772 c.333C>T p.Y111Y Chr.1: 155725444-155725444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375071367 Chr.1: 155725596-155725596 on Build GRCh38
rs765382125 Chr.1: 155725503-155725503 on Build GRCh38
rs754195688 Chr.1: 155725702-155725702 on Build GRCh38
rs755321949 Chr.1: 155725404-155725404 on Build GRCh38
rs758651594 Chr.1: 155725506-155725506 on Build GRCh38
rs145734270 Chr.1: 155725691-155725691 on Build GRCh38
rs916908988 Chr.1: 155725478-155725478 on Build GRCh38
rs1014686164 Chr.1: 155725580-155725580 on Build GRCh38
rs144412256 Chr.1: 155725594-155725594 on Build GRCh38
rs1015484502 Chr.1: 155725845-155725845 on Build GRCh38
rs925790546 Chr.1: 155725361-155725361 on Build GRCh38
rs1027761159 Chr.1: 155725617-155725617 on Build GRCh38
rs11264406 Chr.1: 155725789-155725789 on Build GRCh38
rs994250679 Chr.1: 155725584-155725584 on Build GRCh38
rs1018297157 Chr.1: 155725627-155725627 on Build GRCh38
rs373116379 Chr.1: 155725529-155725529 on Build GRCh38
rs567143323 Chr.1: 155725482-155725482 on Build GRCh38
rs370581706 Chr.1: 155725585-155725585 on Build GRCh38
rs747307421 Chr.1: 155725393-155725393 on Build GRCh38
rs150789035 Chr.1: 155725619-155725619 on Build GRCh38
rs534413248 Chr.1: 155725806-155725806 on Build GRCh38
rs755374880 Chr.1: 155725509-155725509 on Build GRCh38
rs1050290008 Chr.1: 155725365-155725365 on Build GRCh38
rs1018266222 Chr.1: 155725795-155725795 on Build GRCh38
rs200957340 Chr.1: 155725369-155725369 on Build GRCh38
rs560966685 Chr.1: 155725638-155725638 on Build GRCh38
rs368403908 Chr.1: 155725642-155725642 on Build GRCh38
rs1032962713 Chr.1: 155725862-155725862 on Build GRCh38
rs548758207 Chr.1: 155725788-155725788 on Build GRCh38
rs548531604 Chr.1: 155725470-155725470 on Build GRCh38
rs777337438 Chr.1: 155725521-155725521 on Build GRCh38
rs758501463 Chr.1: 155725374-155725374 on Build GRCh38
rs552560079 Chr.1: 155725859-155725859 on Build GRCh38
rs190366665 Chr.1: 155725779-155725779 on Build GRCh38
rs545948218 Chr.1: 155725796-155725796 on Build GRCh38
rs780480654 Chr.1: 155725377-155725377 on Build GRCh38
rs772546005 Chr.1: 155725442-155725442 on Build GRCh38
rs141049384 Chr.1: 155725465-155725465 on Build GRCh38
rs948033090 Chr.1: 155725741-155725741 on Build GRCh38
rs750594124 Chr.1: 155725589-155725589 on Build GRCh38
rs1000103489 Chr.1: 155725817-155725817 on Build GRCh38
rs911758384 Chr.1: 155725447-155725448 on Build GRCh38
rs941597083 Chr.1: 155725721-155725721 on Build GRCh38
rs893846428 Chr.1: 155725738-155725738 on Build GRCh38
rs374539256 Chr.1: 155725787-155725787 on Build GRCh38
rs757463758 Chr.1: 155725368-155725368 on Build GRCh38
rs748621976 Chr.1: 155725433-155725433 on Build GRCh38
rs1038619367 Chr.1: 155725733-155725733 on Build GRCh38
rs138952726 Chr.1: 155725477-155725477 on Build GRCh38
rs926979699 Chr.1: 155725708-155725708 on Build GRCh38
rs554102059 Chr.1: 155725357-155725357 on Build GRCh38
rs944551010 Chr.1: 155725649-155725649 on Build GRCh38
rs542793313 Chr.1: 155725628-155725628 on Build GRCh38
rs781494315 Chr.1: 155725510-155725510 on Build GRCh38
rs576997692 Chr.1: 155725629-155725629 on Build GRCh38
rs949832786 Chr.1: 155725692-155725692 on Build GRCh38
rs769374598 Chr.1: 155725440-155725440 on Build GRCh38
rs748979582 Chr.1: 155725445-155725445 on Build GRCh38
rs748673513 Chr.1: 155725514-155725514 on Build GRCh38
rs766646097 Chr.1: 155725507-155725507 on Build GRCh38
rs756517211 Chr.1: 155725515-155725515 on Build GRCh38
rs147691190 Chr.1: 155725460-155725460 on Build GRCh38
rs149766534 Chr.1: 155725683-155725683 on Build GRCh38
rs1023885817 Chr.1: 155725548-155725548 on Build GRCh38
rs113946783 Chr.1: 155725653-155725653 on Build GRCh38
rs140463523 Chr.1: 155725508-155725508 on Build GRCh38
rs1005770776 Chr.1: 155725557-155725557 on Build GRCh38
rs750690878 Chr.1: 155725371-155725371 on Build GRCh38
rs574621333 Chr.1: 155725625-155725625 on Build GRCh38
rs770465048 Chr.1: 155725536-155725536 on Build GRCh38
rs897310636 Chr.1: 155725767-155725767 on Build GRCh38
rs760491767 Chr.1: 155725502-155725502 on Build GRCh38
rs369172189 Chr.1: 155725520-155725520 on Build GRCh38
rs370909543 Chr.1: 155725648-155725648 on Build GRCh38
rs1038477840 Chr.1: 155725562-155725562 on Build GRCh38
rs12754819 Chr.1: 155725505-155725505 on Build GRCh38
rs574083836 Chr.1: 155725417-155725417 on Build GRCh38
rs3738589 Chr.1: 155725679-155725679 on Build GRCh38
rs376655902 Chr.1: 155725571-155725571 on Build GRCh38
rs781700081 Chr.1: 155725430-155725430 on Build GRCh38
rs906739559 Chr.1: 155725570-155725570 on Build GRCh38

View Full Product Details