Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155319632-155320227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDPS
Entrez Gene ID
2224
Gene Name
farnesyl diphosphate synthase
Gene Aliases
FPPS, FPS, POROK9
UniGene
Hs.335918
Gene Chromosome Location
Chr.1: 155308748-155320666 on Build GRCh38
Transcripts
NM_001135821.1 NM_001135822.1 NM_001242824.1 NM_001242825.1 NM_002004.3 XM_005244962.1 XM_005244963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4561757 c.898G>A p.E300K Chr.1: 155319662-155319662 on Build GRCh38
6121464 c.964G>C p.G322R Chr.1: 155319833-155319833 on Build GRCh38
1747796 c.912C>G p.F304L Chr.1: 155319676-155319676 on Build GRCh38
6662992 c.1059G>T p.K353N Chr.1: 155319928-155319928 on Build GRCh38
6662991 c.1001G>A p.S334N Chr.1: 155319870-155319870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054391105 Chr.1: 155319749-155319749 on Build GRCh38
rs909077922 Chr.1: 155319782-155319782 on Build GRCh38
rs754425677 Chr.1: 155320168-155320168 on Build GRCh38
rs767089569 Chr.1: 155319932-155319932 on Build GRCh38
rs906824796 Chr.1: 155320119-155320119 on Build GRCh38
rs897478180 Chr.1: 155320201-155320201 on Build GRCh38
rs773800168 Chr.1: 155319916-155319916 on Build GRCh38
rs748054065 Chr.1: 155319696-155319696 on Build GRCh38
rs759228894 Chr.1: 155319758-155319758 on Build GRCh38
rs752359619 Chr.1: 155319938-155319938 on Build GRCh38
rs758401183 Chr.1: 155319824-155319824 on Build GRCh38
rs755941790 Chr.1: 155319776-155319776 on Build GRCh38
rs767177758 Chr.1: 155319763-155319763 on Build GRCh38
rs147738837 Chr.1: 155319685-155319685 on Build GRCh38
rs371872906 Chr.1: 155319832-155319832 on Build GRCh38
rs772202497 Chr.1: 155319716-155319716 on Build GRCh38
rs754756693 Chr.1: 155319683-155319683 on Build GRCh38
rs777796449 Chr.1: 155319708-155319708 on Build GRCh38
rs183159854 Chr.1: 155320129-155320129 on Build GRCh38
rs953257548 Chr.1: 155320031-155320031 on Build GRCh38
rs753684541 Chr.1: 155319978-155319978 on Build GRCh38
rs760369267 Chr.1: 155319960-155319960 on Build GRCh38
rs374794832 Chr.1: 155320023-155320023 on Build GRCh38
rs762294350 Chr.1: 155319731-155319731 on Build GRCh38
rs575100393 Chr.1: 155320055-155320055 on Build GRCh38
rs765626045 Chr.1: 155319736-155319736 on Build GRCh38
rs536916396 Chr.1: 155319855-155319855 on Build GRCh38
rs539881168 Chr.1: 155319700-155319700 on Build GRCh38
rs764824041 Chr.1: 155320122-155320122 on Build GRCh38
rs760296516 Chr.1: 155319771-155319771 on Build GRCh38
rs542471474 Chr.1: 155320077-155320077 on Build GRCh38
rs770398620 Chr.1: 155319915-155319915 on Build GRCh38
rs546317941 Chr.1: 155319699-155319699 on Build GRCh38
rs1012803914 Chr.1: 155319765-155319765 on Build GRCh38
rs753868548 Chr.1: 155319822-155319822 on Build GRCh38
rs141139771 Chr.1: 155320051-155320051 on Build GRCh38
rs764012544 Chr.1: 155319963-155319963 on Build GRCh38
rs916011914 Chr.1: 155319753-155319753 on Build GRCh38
rs76468446 Chr.1: 155320114-155320114 on Build GRCh38
rs986341042 Chr.1: 155320040-155320040 on Build GRCh38
rs534198189 Chr.1: 155320011-155320011 on Build GRCh38
rs564760041 Chr.1: 155320128-155320128 on Build GRCh38
rs200432999 Chr.1: 155319896-155319896 on Build GRCh38
rs781599688 Chr.1: 155319837-155319837 on Build GRCh38
rs1023900192 Chr.1: 155319809-155319809 on Build GRCh38
rs146928362 Chr.1: 155319835-155319835 on Build GRCh38
rs372588531 Chr.1: 155319724-155319724 on Build GRCh38
rs147759901 Chr.1: 155320048-155320048 on Build GRCh38
rs567094112 Chr.1: 155319914-155319914 on Build GRCh38
rs941865357 Chr.1: 155319815-155319815 on Build GRCh38
rs762312349 Chr.1: 155319752-155319752 on Build GRCh38
rs144949010 Chr.1: 155320047-155320047 on Build GRCh38
rs1045604413 Chr.1: 155320118-155320118 on Build GRCh38
rs747298940 Chr.1: 155319720-155319720 on Build GRCh38
rs554838568 Chr.1: 155320080-155320080 on Build GRCh38
rs745867995 Chr.1: 155319715-155319715 on Build GRCh38
rs370400053 Chr.1: 155320064-155320065 on Build GRCh38
rs759075926 Chr.1: 155319919-155319919 on Build GRCh38
rs992805873 Chr.1: 155319739-155319739 on Build GRCh38
rs781033064 Chr.1: 155319694-155319694 on Build GRCh38
rs759220845 Chr.1: 155320064-155320064 on Build GRCh38
rs773777378 Chr.1: 155319738-155319738 on Build GRCh38
rs770629251 Chr.1: 155319756-155319757 on Build GRCh38
rs773847307 Chr.1: 155319980-155319982 on Build GRCh38
rs747214912 Chr.1: 155319831-155319831 on Build GRCh38
rs751226316 Chr.1: 155319659-155319659 on Build GRCh38
rs11264361 Chr.1: 155319754-155319754 on Build GRCh38
rs544343798 Chr.1: 155320177-155320177 on Build GRCh38
rs201526312 Chr.1: 155319816-155319816 on Build GRCh38
rs543749231 Chr.1: 155319770-155319770 on Build GRCh38
rs766244906 Chr.1: 155319846-155319846 on Build GRCh38
rs973166263 Chr.1: 155319991-155319991 on Build GRCh38
rs143140546 Chr.1: 155319902-155319902 on Build GRCh38
rs368286561 Chr.1: 155319718-155319718 on Build GRCh38
rs201016550 Chr.1: 155319825-155319825 on Build GRCh38
rs191093924 Chr.1: 155320090-155320090 on Build GRCh38
rs544368647 Chr.1: 155320062-155320063 on Build GRCh38
rs748399701 Chr.1: 155319850-155319850 on Build GRCh38

View Full Product Details