Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 229366701-229367267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNER
Entrez Gene ID
92737
Gene Name
delta/notch like EGF repeat containing
Gene Aliases
UNQ26, bet
UniGene
Hs.234074
Gene Chromosome Location
Chr.2: 229357629-229714570 on Build GRCh38
Transcripts
NM_139072.3 XM_005246950.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5928259 c.1954C>T p.L652F Chr.2: 229367021-229367021 on Build GRCh38
6442140 c.1871C>A p.S624Y Chr.2: 229367104-229367104 on Build GRCh38
7250397 c.2093G>T p.R698L Chr.2: 229366882-229366882 on Build GRCh38
1692119 c.1891C>T p.L631F Chr.2: 229367084-229367084 on Build GRCh38
1692120 c.1866G>A p.W622* Chr.2: 229367109-229367109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528331788 Chr.2: 229366964-229366964 on Build GRCh38
rs751453763 Chr.2: 229366846-229366846 on Build GRCh38
rs139514662 Chr.2: 229366759-229366759 on Build GRCh38
rs571518557 Chr.2: 229367027-229367027 on Build GRCh38
rs372749536 Chr.2: 229367168-229367168 on Build GRCh38
rs770592104 Chr.2: 229367163-229367163 on Build GRCh38
rs777629340 Chr.2: 229367147-229367147 on Build GRCh38
rs776566467 Chr.2: 229367011-229367011 on Build GRCh38
rs1043859821 Chr.2: 229367126-229367126 on Build GRCh38
rs758068063 Chr.2: 229367141-229367141 on Build GRCh38
rs755411230 Chr.2: 229367062-229367062 on Build GRCh38
rs1044280014 Chr.2: 229366952-229366952 on Build GRCh38
rs150035332 Chr.2: 229366910-229366910 on Build GRCh38
rs772826749 Chr.2: 229366857-229366857 on Build GRCh38
rs777343605 Chr.2: 229367069-229367069 on Build GRCh38
rs553161857 Chr.2: 229367127-229367127 on Build GRCh38
rs112975208 Chr.2: 229366854-229366854 on Build GRCh38
rs141125901 Chr.2: 229367001-229367001 on Build GRCh38
rs780209701 Chr.2: 229367214-229367214 on Build GRCh38
rs1019649993 Chr.2: 229367182-229367182 on Build GRCh38
rs370587377 Chr.2: 229367000-229367000 on Build GRCh38
rs752912056 Chr.2: 229366917-229366917 on Build GRCh38
rs760704438 Chr.2: 229367007-229367007 on Build GRCh38
rs780401095 Chr.2: 229366978-229366978 on Build GRCh38
rs866256730 Chr.2: 229367109-229367109 on Build GRCh38
rs770173487 Chr.2: 229366883-229366883 on Build GRCh38
rs147670807 Chr.2: 229367194-229367194 on Build GRCh38
rs750171761 Chr.2: 229366855-229366855 on Build GRCh38
rs775160268 Chr.2: 229366830-229366830 on Build GRCh38
rs771341328 Chr.2: 229366867-229366867 on Build GRCh38
rs767646633 Chr.2: 229367030-229367030 on Build GRCh38
rs141811504 Chr.2: 229367033-229367033 on Build GRCh38
rs901477630 Chr.2: 229367203-229367203 on Build GRCh38
rs111775882 Chr.2: 229366790-229366790 on Build GRCh38
rs1007808308 Chr.2: 229367180-229367180 on Build GRCh38
rs549129843 Chr.2: 229366925-229366925 on Build GRCh38
rs775072382 Chr.2: 229366884-229366884 on Build GRCh38
rs776762113 Chr.2: 229367080-229367080 on Build GRCh38
rs759360449 Chr.2: 229367081-229367081 on Build GRCh38
rs969679545 Chr.2: 229367188-229367188 on Build GRCh38
rs746587370 Chr.2: 229367076-229367076 on Build GRCh38
rs778014620 Chr.2: 229366864-229366864 on Build GRCh38
rs770702460 Chr.2: 229367028-229367028 on Build GRCh38
rs762406084 Chr.2: 229366842-229366842 on Build GRCh38
rs761443379 Chr.2: 229366897-229366897 on Build GRCh38
rs769410792 Chr.2: 229366828-229366828 on Build GRCh38
rs770496301 Chr.2: 229366752-229366752 on Build GRCh38
rs993574120 Chr.2: 229367240-229367240 on Build GRCh38
rs779351483 Chr.2: 229367118-229367118 on Build GRCh38
rs145368755 Chr.2: 229366916-229366916 on Build GRCh38
rs769650188 Chr.2: 229367082-229367082 on Build GRCh38
rs747487250 Chr.2: 229366865-229366865 on Build GRCh38
rs201490102 Chr.2: 229366974-229366974 on Build GRCh38
rs777202282 Chr.2: 229367216-229367216 on Build GRCh38
rs761660347 Chr.2: 229366853-229366853 on Build GRCh38
rs767698174 Chr.2: 229367092-229367092 on Build GRCh38
rs143853534 Chr.2: 229366882-229366882 on Build GRCh38
rs145890820 Chr.2: 229366986-229366986 on Build GRCh38
rs146098212 Chr.2: 229367091-229367091 on Build GRCh38
rs948597951 Chr.2: 229367037-229367037 on Build GRCh38
rs780595412 Chr.2: 229367049-229367049 on Build GRCh38
rs778522656 Chr.2: 229366895-229366895 on Build GRCh38
rs781223791 Chr.2: 229367166-229367166 on Build GRCh38
rs2302769 Chr.2: 229367173-229367173 on Build GRCh38
rs752534359 Chr.2: 229367211-229367211 on Build GRCh38
rs751449773 Chr.2: 229366987-229366987 on Build GRCh38
rs758506865 Chr.2: 229366859-229366859 on Build GRCh38
rs546911349 Chr.2: 229366983-229366983 on Build GRCh38
rs1019207998 Chr.2: 229366789-229366789 on Build GRCh38
rs148580590 Chr.2: 229367102-229367102 on Build GRCh38
rs770982597 Chr.2: 229367079-229367079 on Build GRCh38
rs751581618 Chr.2: 229366926-229366926 on Build GRCh38
rs767334705 Chr.2: 229366900-229366900 on Build GRCh38
rs763615331 Chr.2: 229366844-229366844 on Build GRCh38
rs146422547 Chr.2: 229366928-229366929 on Build GRCh38
rs898996837 Chr.2: 229366733-229366733 on Build GRCh38
rs147002066 Chr.2: 229367112-229367112 on Build GRCh38
rs749718515 Chr.2: 229366979-229366979 on Build GRCh38
rs774375660 Chr.2: 229367087-229367087 on Build GRCh38
rs768087498 Chr.2: 229366894-229366894 on Build GRCh38
rs1023954926 Chr.2: 229367183-229367183 on Build GRCh38
rs760016453 Chr.2: 229366825-229366825 on Build GRCh38
rs111685443 Chr.2: 229366881-229366881 on Build GRCh38
rs373612462 Chr.2: 229366858-229366858 on Build GRCh38
rs200069865 Chr.2: 229367068-229367068 on Build GRCh38
rs149348216 Chr.2: 229367096-229367096 on Build GRCh38
rs764187705 Chr.2: 229367025-229367025 on Build GRCh38
rs561051341 Chr.2: 229366928-229366928 on Build GRCh38
rs538564713 Chr.2: 229367067-229367067 on Build GRCh38
rs778391300 Chr.2: 229367064-229367064 on Build GRCh38
rs142994637 Chr.2: 229367103-229367103 on Build GRCh38
rs748951182 Chr.2: 229366892-229366892 on Build GRCh38
rs367716187 Chr.2: 229366908-229366908 on Build GRCh38
rs138706796 Chr.2: 229367034-229367034 on Build GRCh38
rs746499540 Chr.2: 229367151-229367151 on Build GRCh38
rs766213399 Chr.2: 229367008-229367008 on Build GRCh38
rs530602445 Chr.2: 229366924-229366924 on Build GRCh38
rs749630300 Chr.2: 229367060-229367060 on Build GRCh38
rs60699589 Chr.2: 229366756-229366756 on Build GRCh38
rs781507293 Chr.2: 229366932-229366932 on Build GRCh38
rs200724319 Chr.2: 229366915-229366915 on Build GRCh38
rs74488906 Chr.2: 229366791-229366791 on Build GRCh38
rs762238880 Chr.2: 229366736-229366736 on Build GRCh38

View Full Product Details