Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33585817-33586386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DONSON
Entrez Gene ID
29980
Gene Name
downstream neighbor of SON
Gene Aliases
B17, C21orf60
UniGene
Hs.436341
Gene Chromosome Location
Chr.21: 33577551-33588708 on Build GRCh38
Transcripts
NM_017613.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
350254 c.531G>A p.L177L Chr.21: 33586053-33586053 on Build GRCh38
3972758 c.482C>T p.T161M Chr.21: 33586102-33586102 on Build GRCh38
4926482 c.436C>A p.P146T Chr.21: 33586148-33586148 on Build GRCh38
3841821 c.505C>G p.Q169E Chr.21: 33586079-33586079 on Build GRCh38
3550416 c.421T>G p.S141A Chr.21: 33586163-33586163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751159231 Chr.21: 33586210-33586210 on Build GRCh38
rs927236850 Chr.21: 33586323-33586323 on Build GRCh38
rs774855346 Chr.21: 33586021-33586021 on Build GRCh38
rs754454018 Chr.21: 33586135-33586135 on Build GRCh38
rs920184198 Chr.21: 33586256-33586256 on Build GRCh38
rs974293696 Chr.21: 33586253-33586253 on Build GRCh38
rs555750312 Chr.21: 33586131-33586131 on Build GRCh38
rs755729661 Chr.21: 33586232-33586232 on Build GRCh38
rs559494341 Chr.21: 33586355-33586355 on Build GRCh38
rs965498360 Chr.21: 33585911-33585911 on Build GRCh38
rs574419195 Chr.21: 33586244-33586244 on Build GRCh38
rs750284399 Chr.21: 33585954-33585954 on Build GRCh38
rs377656968 Chr.21: 33585945-33585945 on Build GRCh38
rs376262017 Chr.21: 33586192-33586192 on Build GRCh38
rs199655790 Chr.21: 33585971-33585971 on Build GRCh38
rs1000384402 Chr.21: 33585841-33585841 on Build GRCh38
rs765127652 Chr.21: 33586190-33586190 on Build GRCh38
rs2276225 Chr.21: 33586299-33586299 on Build GRCh38
rs1042163743 Chr.21: 33586365-33586365 on Build GRCh38
rs766100630 Chr.21: 33586202-33586202 on Build GRCh38
rs753740715 Chr.21: 33586100-33586100 on Build GRCh38
rs972872027 Chr.21: 33586252-33586252 on Build GRCh38
rs918128188 Chr.21: 33586324-33586324 on Build GRCh38
rs776559036 Chr.21: 33586171-33586171 on Build GRCh38
rs746496033 Chr.21: 33585965-33585965 on Build GRCh38
rs908520235 Chr.21: 33586219-33586219 on Build GRCh38
rs755496540 Chr.21: 33586283-33586283 on Build GRCh38
rs542882053 Chr.21: 33586352-33586352 on Build GRCh38
rs779730426 Chr.21: 33585964-33585964 on Build GRCh38
rs753360310 Chr.21: 33586321-33586321 on Build GRCh38
rs779615686 Chr.21: 33586147-33586147 on Build GRCh38
rs746207457 Chr.21: 33586150-33586150 on Build GRCh38
rs761927338 Chr.21: 33586173-33586173 on Build GRCh38
rs752235448 Chr.21: 33586230-33586230 on Build GRCh38
rs377360381 Chr.21: 33586111-33586111 on Build GRCh38
rs560205499 Chr.21: 33585918-33585918 on Build GRCh38
rs1025328588 Chr.21: 33586257-33586257 on Build GRCh38
rs992703018 Chr.21: 33586327-33586327 on Build GRCh38
rs772406088 Chr.21: 33586047-33586047 on Build GRCh38
rs764091077 Chr.21: 33586078-33586078 on Build GRCh38
rs1029166032 Chr.21: 33586151-33586151 on Build GRCh38
rs371685464 Chr.21: 33586166-33586166 on Build GRCh38
rs758031611 Chr.21: 33585959-33585959 on Build GRCh38
rs147415181 Chr.21: 33586037-33586037 on Build GRCh38
rs112757200 Chr.21: 33586349-33586349 on Build GRCh38
rs767260621 Chr.21: 33586229-33586229 on Build GRCh38
rs760755463 Chr.21: 33586076-33586076 on Build GRCh38
rs35144167 Chr.21: 33585881-33585882 on Build GRCh38
rs895936669 Chr.21: 33586086-33586086 on Build GRCh38
rs771356652 Chr.21: 33585994-33585994 on Build GRCh38
rs762688764 Chr.21: 33586201-33586201 on Build GRCh38
rs527241707 Chr.21: 33586244-33586244 on Build GRCh38
rs953517829 Chr.21: 33585888-33585888 on Build GRCh38
rs139553109 Chr.21: 33586142-33586142 on Build GRCh38
rs1057365152 Chr.21: 33586082-33586082 on Build GRCh38
rs920068195 Chr.21: 33585860-33585861 on Build GRCh38
rs757016491 Chr.21: 33585929-33585929 on Build GRCh38
rs754583646 Chr.21: 33586212-33586212 on Build GRCh38
rs761797700 Chr.21: 33586193-33586196 on Build GRCh38
rs1041741921 Chr.21: 33586102-33586102 on Build GRCh38
rs560220669 Chr.21: 33586330-33586330 on Build GRCh38
rs778294070 Chr.21: 33585985-33585985 on Build GRCh38
rs918230324 Chr.21: 33585941-33585941 on Build GRCh38
rs912273739 Chr.21: 33585908-33585908 on Build GRCh38
rs778681259 Chr.21: 33585935-33585935 on Build GRCh38
rs747762582 Chr.21: 33586144-33586144 on Build GRCh38
rs751194578 Chr.21: 33586134-33586134 on Build GRCh38
rs775719667 Chr.21: 33586161-33586161 on Build GRCh38
rs1029133196 Chr.21: 33585862-33585862 on Build GRCh38
rs937235660 Chr.21: 33586325-33586325 on Build GRCh38
rs368596012 Chr.21: 33586158-33586158 on Build GRCh38
rs780645262 Chr.21: 33586148-33586148 on Build GRCh38
rs751559041 Chr.21: 33586183-33586184 on Build GRCh38
rs985183664 Chr.21: 33585891-33585891 on Build GRCh38
rs953548902 Chr.21: 33586164-33586164 on Build GRCh38
rs544457425 Chr.21: 33586128-33586128 on Build GRCh38
rs762653453 Chr.21: 33585935-33585936 on Build GRCh38
rs544797310 Chr.21: 33586319-33586319 on Build GRCh38
rs1010722195 Chr.21: 33586090-33586090 on Build GRCh38
rs756689989 Chr.21: 33586065-33586065 on Build GRCh38
rs773163935 Chr.21: 33586200-33586200 on Build GRCh38
rs761731134 Chr.21: 33586101-33586101 on Build GRCh38
rs763917165 Chr.21: 33585959-33585960 on Build GRCh38
rs374188850 Chr.21: 33586132-33586132 on Build GRCh38
rs952323145 Chr.21: 33586278-33586278 on Build GRCh38

View Full Product Details