Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36070951-36071524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR1
Entrez Gene ID
873
Gene Name
carbonyl reductase 1
Gene Aliases
CBR, SDR21C1, hCBR1
UniGene
Hs.88778
Gene Chromosome Location
Chr.21: 36069924-36073177 on Build GRCh38
Transcripts
NM_001286789.1 NM_001757.3 NR_040084.1

Gene Symbol
LOC100133286
Entrez Gene ID
100133286
Gene Name
uncharacterized LOC100133286
Gene Aliases
-
UniGene
Hs.728809
Gene Chromosome Location
Chr.21: 36069642-36126640 on Build GRCh38
Transcripts
NM_001286789.1 NM_001757.3 NR_040084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5974225 c.359A>G p.D120G Chr.21: 36071019-36071019 on Build GRCh38
478553 c.324A>G p.E108E Chr.21: 36070984-36070984 on Build GRCh38
1413977 c.366C>T p.C122C Chr.21: 36071026-36071026 on Build GRCh38
5392867 c.390A>G p.K130K Chr.21: 36071050-36071050 on Build GRCh38
2844384 c.330G>A p.T110T Chr.21: 36070990-36070990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047971351 Chr.21: 36071391-36071391 on Build GRCh38
rs375784875 Chr.21: 36071024-36071024 on Build GRCh38
rs563909657 Chr.21: 36071155-36071155 on Build GRCh38
rs779948695 Chr.21: 36071190-36071190 on Build GRCh38
rs928620563 Chr.21: 36071293-36071294 on Build GRCh38
rs966667547 Chr.21: 36071307-36071307 on Build GRCh38
rs1032046144 Chr.21: 36071270-36071270 on Build GRCh38
rs757911931 Chr.21: 36071036-36071036 on Build GRCh38
rs759348201 Chr.21: 36071013-36071013 on Build GRCh38
rs200192508 Chr.21: 36070989-36070989 on Build GRCh38
rs778239233 Chr.21: 36071063-36071063 on Build GRCh38
rs953773304 Chr.21: 36071394-36071394 on Build GRCh38
rs747107090 Chr.21: 36071195-36071195 on Build GRCh38
rs753663954 Chr.21: 36071007-36071007 on Build GRCh38
rs754253094 Chr.21: 36071166-36071166 on Build GRCh38
rs946491141 Chr.21: 36071119-36071119 on Build GRCh38
rs761599903 Chr.21: 36071121-36071121 on Build GRCh38
rs751141827 Chr.21: 36071153-36071153 on Build GRCh38
rs776354083 Chr.21: 36071114-36071114 on Build GRCh38
rs778116397 Chr.21: 36070980-36070980 on Build GRCh38
rs530322864 Chr.21: 36071184-36071184 on Build GRCh38
rs970189549 Chr.21: 36071473-36071473 on Build GRCh38
rs746020945 Chr.21: 36071209-36071209 on Build GRCh38
rs760444957 Chr.21: 36071005-36071005 on Build GRCh38
rs1002191556 Chr.21: 36071189-36071189 on Build GRCh38
rs537530000 Chr.21: 36071266-36071266 on Build GRCh38
rs764885114 Chr.21: 36071022-36071022 on Build GRCh38
rs1049201766 Chr.21: 36071325-36071325 on Build GRCh38
rs190160512 Chr.21: 36071361-36071361 on Build GRCh38
rs2156406 Chr.21: 36071373-36071373 on Build GRCh38
rs542811393 Chr.21: 36071247-36071248 on Build GRCh38
rs929451642 Chr.21: 36071321-36071321 on Build GRCh38
rs545432870 Chr.21: 36071090-36071090 on Build GRCh38
rs763855338 Chr.21: 36071189-36071189 on Build GRCh38
rs557204729 Chr.21: 36070995-36070995 on Build GRCh38
rs1005696 Chr.21: 36071182-36071182 on Build GRCh38
rs768515405 Chr.21: 36071198-36071198 on Build GRCh38
rs192717129 Chr.21: 36071224-36071224 on Build GRCh38
rs986435269 Chr.21: 36071038-36071038 on Build GRCh38
rs3787726 Chr.21: 36071364-36071364 on Build GRCh38
rs970476399 Chr.21: 36071291-36071291 on Build GRCh38
rs1005695 Chr.21: 36071267-36071267 on Build GRCh38
rs765126870 Chr.21: 36071019-36071020 on Build GRCh38
rs966322447 Chr.21: 36071440-36071440 on Build GRCh38
rs118126898 Chr.21: 36071289-36071289 on Build GRCh38
rs570206837 Chr.21: 36071227-36071227 on Build GRCh38
rs938044398 Chr.21: 36071338-36071338 on Build GRCh38
rs902390226 Chr.21: 36071380-36071380 on Build GRCh38
rs765843052 Chr.21: 36071150-36071150 on Build GRCh38
rs781715120 Chr.21: 36071046-36071046 on Build GRCh38
rs897925767 Chr.21: 36071146-36071146 on Build GRCh38
rs774437766 Chr.21: 36070985-36070985 on Build GRCh38
rs757494065 Chr.21: 36071079-36071079 on Build GRCh38
rs762680340 Chr.21: 36071182-36071183 on Build GRCh38
rs772888706 Chr.21: 36071247-36071247 on Build GRCh38
rs753192675 Chr.21: 36071049-36071049 on Build GRCh38
rs749558223 Chr.21: 36070981-36070981 on Build GRCh38
rs749690236 Chr.21: 36071070-36071070 on Build GRCh38
rs892296070 Chr.21: 36071110-36071110 on Build GRCh38
rs763917846 Chr.21: 36071006-36071006 on Build GRCh38
rs1055504587 Chr.21: 36071340-36071340 on Build GRCh38
rs758678387 Chr.21: 36071186-36071186 on Build GRCh38
rs532865639 Chr.21: 36071160-36071160 on Build GRCh38
rs761407628 Chr.21: 36071015-36071015 on Build GRCh38
rs572763511 Chr.21: 36071480-36071480 on Build GRCh38
rs41557318 Chr.21: 36071051-36071051 on Build GRCh38
rs1006571287 Chr.21: 36071402-36071402 on Build GRCh38
rs565597795 Chr.21: 36071296-36071296 on Build GRCh38
rs764936035 Chr.21: 36071128-36071128 on Build GRCh38
rs943096906 Chr.21: 36071362-36071362 on Build GRCh38
rs981114222 Chr.21: 36070990-36070990 on Build GRCh38
rs775491534 Chr.21: 36071051-36071052 on Build GRCh38
rs976286176 Chr.21: 36071316-36071316 on Build GRCh38
rs1040104215 Chr.21: 36071363-36071363 on Build GRCh38
rs113888409 Chr.21: 36071439-36071439 on Build GRCh38
rs547794915 Chr.21: 36071170-36071170 on Build GRCh38
rs981494506 Chr.21: 36071292-36071292 on Build GRCh38
rs775330867 Chr.21: 36071233-36071233 on Build GRCh38
rs775470312 Chr.21: 36071088-36071088 on Build GRCh38
rs769526133 Chr.21: 36071244-36071244 on Build GRCh38
rs1021170847 Chr.21: 36071258-36071258 on Build GRCh38
rs1032078254 Chr.21: 36071388-36071388 on Build GRCh38
rs762501525 Chr.21: 36071149-36071149 on Build GRCh38
rs772044820 Chr.21: 36070997-36070997 on Build GRCh38
rs772797612 Chr.21: 36071138-36071138 on Build GRCh38
rs150365005 Chr.21: 36071002-36071002 on Build GRCh38
rs1023541379 Chr.21: 36071469-36071469 on Build GRCh38
rs772097439 Chr.21: 36071083-36071083 on Build GRCh38
rs745950206 Chr.21: 36071082-36071082 on Build GRCh38
rs759028268 Chr.21: 36071016-36071016 on Build GRCh38
rs993749760 Chr.21: 36071237-36071237 on Build GRCh38
rs559624284 Chr.21: 36071252-36071253 on Build GRCh38
rs138094996 Chr.21: 36071328-36071328 on Build GRCh38
rs1000643873 Chr.21: 36071386-36071386 on Build GRCh38
rs1035686450 Chr.21: 36071497-36071497 on Build GRCh38
rs1006360426 Chr.21: 36071444-36071444 on Build GRCh38
rs919495098 Chr.21: 36071500-36071500 on Build GRCh38
rs919386702 Chr.21: 36071320-36071320 on Build GRCh38
rs1005694 Chr.21: 36071360-36071360 on Build GRCh38
rs779074269 Chr.21: 36071080-36071080 on Build GRCh38
rs1018611683 Chr.21: 36071204-36071204 on Build GRCh38
rs768603380 Chr.21: 36071113-36071113 on Build GRCh38
rs747156853 Chr.21: 36071057-36071057 on Build GRCh38
rs184719814 Chr.21: 36071285-36071285 on Build GRCh38

View Full Product Details