Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38483876-38484475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX17
Entrez Gene ID
10521
Gene Name
DEAD-box helicase 17
Gene Aliases
P72, RH70
UniGene
Hs.528305
Gene Chromosome Location
Chr.22: 38483438-38506340 on Build GRCh38
Transcripts
NM_001098504.1 NM_006386.4

SNP Detail

SNP ID Chromosome Location
rs773412087 Chr.22: 38484030-38484031 on Build GRCh38
rs144704720 Chr.22: 38484293-38484293 on Build GRCh38
rs928984436 Chr.22: 38484292-38484293 on Build GRCh38
rs1055956967 Chr.22: 38483962-38483962 on Build GRCh38
rs901646026 Chr.22: 38484393-38484393 on Build GRCh38
rs1017532524 Chr.22: 38484030-38484030 on Build GRCh38
rs760792240 Chr.22: 38484245-38484246 on Build GRCh38
rs936341792 Chr.22: 38484412-38484412 on Build GRCh38
rs958769717 Chr.22: 38484359-38484359 on Build GRCh38
rs5845381 Chr.22: 38484093-38484093 on Build GRCh38
rs956823624 Chr.22: 38483945-38483945 on Build GRCh38
rs981406611 Chr.22: 38484349-38484349 on Build GRCh38
rs916154830 Chr.22: 38484112-38484112 on Build GRCh38
rs748423557 Chr.22: 38483974-38483974 on Build GRCh38
rs1048785447 Chr.22: 38484167-38484167 on Build GRCh38
rs560323426 Chr.22: 38483904-38483904 on Build GRCh38
rs763121 Chr.22: 38483935-38483935 on Build GRCh38
rs775407022 Chr.22: 38484075-38484077 on Build GRCh38
rs1007888114 Chr.22: 38484156-38484156 on Build GRCh38
rs912809401 Chr.22: 38484385-38484385 on Build GRCh38
rs892278435 Chr.22: 38484293-38484293 on Build GRCh38
rs946119459 Chr.22: 38483920-38483920 on Build GRCh38
rs916038719 Chr.22: 38484264-38484268 on Build GRCh38
rs398101758 Chr.22: 38484253-38484253 on Build GRCh38
rs1047270013 Chr.22: 38484105-38484105 on Build GRCh38
rs1042280403 Chr.22: 38483923-38483923 on Build GRCh38
rs1033391396 Chr.22: 38484415-38484415 on Build GRCh38
rs760956277 Chr.22: 38484053-38484054 on Build GRCh38
rs1030087830 Chr.22: 38484246-38484247 on Build GRCh38
rs755816148 Chr.22: 38484170-38484170 on Build GRCh38
rs993238985 Chr.22: 38484014-38484014 on Build GRCh38
rs1056011483 Chr.22: 38484106-38484106 on Build GRCh38
rs564130574 Chr.22: 38484083-38484083 on Build GRCh38
rs545943315 Chr.22: 38484036-38484036 on Build GRCh38
rs190443601 Chr.22: 38484386-38484386 on Build GRCh38
rs1007394253 Chr.22: 38484171-38484171 on Build GRCh38
rs1013872287 Chr.22: 38483979-38483979 on Build GRCh38
rs1039425468 Chr.22: 38484178-38484178 on Build GRCh38
rs887544848 Chr.22: 38484373-38484373 on Build GRCh38
rs998860055 Chr.22: 38484409-38484409 on Build GRCh38
rs543257541 Chr.22: 38484042-38484042 on Build GRCh38
rs112341627 Chr.22: 38484097-38484097 on Build GRCh38
rs1045949696 Chr.22: 38484273-38484273 on Build GRCh38
rs561654924 Chr.22: 38484245-38484245 on Build GRCh38
rs559363645 Chr.22: 38484372-38484372 on Build GRCh38
rs950267691 Chr.22: 38484147-38484147 on Build GRCh38
rs964076759 Chr.22: 38484032-38484032 on Build GRCh38
rs1009819720 Chr.22: 38484307-38484307 on Build GRCh38
rs1045392826 Chr.22: 38484151-38484151 on Build GRCh38
rs1017417628 Chr.22: 38484179-38484180 on Build GRCh38
rs200707579 Chr.22: 38484308-38484308 on Build GRCh38
rs1024042028 Chr.22: 38483985-38483985 on Build GRCh38
rs567588807 Chr.22: 38484336-38484336 on Build GRCh38
rs927374814 Chr.22: 38484026-38484026 on Build GRCh38
rs775575639 Chr.22: 38484403-38484403 on Build GRCh38
rs1049319183 Chr.22: 38484297-38484297 on Build GRCh38
rs971087171 Chr.22: 38484325-38484325 on Build GRCh38
rs181066892 Chr.22: 38484441-38484441 on Build GRCh38
rs977931528 Chr.22: 38484249-38484249 on Build GRCh38
rs73157136 Chr.22: 38484021-38484021 on Build GRCh38
rs1000643673 Chr.22: 38484247-38484247 on Build GRCh38
rs977502307 Chr.22: 38484084-38484084 on Build GRCh38
rs149118579 Chr.22: 38484251-38484251 on Build GRCh38
rs868475148 Chr.22: 38484242-38484242 on Build GRCh38
rs201070119 Chr.22: 38484244-38484245 on Build GRCh38
rs398101757 Chr.22: 38484094-38484094 on Build GRCh38
rs779511117 Chr.22: 38484289-38484289 on Build GRCh38
rs146941567 Chr.22: 38484017-38484017 on Build GRCh38
rs552200609 Chr.22: 38484006-38484009 on Build GRCh38
rs891379234 Chr.22: 38484144-38484144 on Build GRCh38
rs532894019 Chr.22: 38484407-38484407 on Build GRCh38
rs11538316 Chr.22: 38484426-38484426 on Build GRCh38
rs930154371 Chr.22: 38484088-38484088 on Build GRCh38
rs988049436 Chr.22: 38483968-38483968 on Build GRCh38
rs534776956 Chr.22: 38483976-38483976 on Build GRCh38
rs550993723 Chr.22: 38484238-38484238 on Build GRCh38
rs922559300 Chr.22: 38484051-38484051 on Build GRCh38
rs974103913 Chr.22: 38484368-38484368 on Build GRCh38
rs950316966 Chr.22: 38484269-38484273 on Build GRCh38
rs1034337282 Chr.22: 38484353-38484353 on Build GRCh38
rs1032522475 Chr.22: 38484279-38484279 on Build GRCh38
rs944294583 Chr.22: 38484399-38484399 on Build GRCh38
rs5845383 Chr.22: 38484245-38484245 on Build GRCh38
rs897462095 Chr.22: 38484154-38484155 on Build GRCh38
rs373847642 Chr.22: 38483912-38483912 on Build GRCh38
rs918941650 Chr.22: 38484292-38484292 on Build GRCh38
rs778118866 Chr.22: 38484283-38484283 on Build GRCh38
rs547439489 Chr.22: 38484326-38484326 on Build GRCh38
rs983388061 Chr.22: 38484376-38484376 on Build GRCh38
rs576283704 Chr.22: 38484087-38484087 on Build GRCh38
rs930125450 Chr.22: 38484374-38484374 on Build GRCh38
rs3216849 Chr.22: 38484097-38484097 on Build GRCh38
rs749439645 Chr.22: 38484294-38484294 on Build GRCh38
rs967883711 Chr.22: 38484080-38484080 on Build GRCh38

View Full Product Details