Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39264005-39264550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CR1
Entrez Gene ID
1524
Gene Name
C-X3-C motif chemokine receptor 1
Gene Aliases
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
UniGene
Hs.78913
Gene Chromosome Location
Chr.3: 39263494-39281735 on Build GRCh38
Transcripts
NM_001171171.1 NM_001171172.1 NM_001171174.1 NM_001337.3

SNP Detail

SNP ID Chromosome Location
rs953584844 Chr.3: 39264092-39264092 on Build GRCh38
rs916459896 Chr.3: 39264467-39264467 on Build GRCh38
rs765057979 Chr.3: 39264028-39264028 on Build GRCh38
rs532615161 Chr.3: 39264338-39264338 on Build GRCh38
rs965472145 Chr.3: 39264347-39264347 on Build GRCh38
rs936561101 Chr.3: 39264163-39264163 on Build GRCh38
rs746650329 Chr.3: 39264523-39264523 on Build GRCh38
rs868594594 Chr.3: 39264438-39264438 on Build GRCh38
rs978137748 Chr.3: 39264160-39264160 on Build GRCh38
rs185538043 Chr.3: 39264225-39264225 on Build GRCh38
rs973654102 Chr.3: 39264032-39264032 on Build GRCh38
rs916417126 Chr.3: 39264191-39264191 on Build GRCh38
rs941523078 Chr.3: 39264239-39264239 on Build GRCh38
rs1022382819 Chr.3: 39264434-39264434 on Build GRCh38
rs1049894524 Chr.3: 39264198-39264198 on Build GRCh38
rs550687298 Chr.3: 39264454-39264454 on Build GRCh38
rs903720925 Chr.3: 39264439-39264439 on Build GRCh38
rs976137666 Chr.3: 39264155-39264155 on Build GRCh38
rs989390364 Chr.3: 39264458-39264458 on Build GRCh38
rs1036487133 Chr.3: 39264516-39264516 on Build GRCh38
rs897388826 Chr.3: 39264337-39264337 on Build GRCh38
rs115326610 Chr.3: 39264447-39264447 on Build GRCh38
rs71534233 Chr.3: 39264186-39264187 on Build GRCh38
rs1053729254 Chr.3: 39264187-39264187 on Build GRCh38
rs376154355 Chr.3: 39264279-39264279 on Build GRCh38
rs767783481 Chr.3: 39264341-39264341 on Build GRCh38
rs1012299007 Chr.3: 39264432-39264432 on Build GRCh38
rs546816109 Chr.3: 39264114-39264114 on Build GRCh38
rs756240450 Chr.3: 39264416-39264416 on Build GRCh38
rs368646004 Chr.3: 39264161-39264161 on Build GRCh38
rs1009441627 Chr.3: 39264323-39264323 on Build GRCh38
rs985167394 Chr.3: 39264109-39264109 on Build GRCh38
rs927845365 Chr.3: 39264493-39264493 on Build GRCh38
rs995046107 Chr.3: 39264300-39264300 on Build GRCh38
rs55870737 Chr.3: 39264366-39264366 on Build GRCh38
rs147642545 Chr.3: 39264437-39264437 on Build GRCh38
rs1037321084 Chr.3: 39264257-39264257 on Build GRCh38
rs869257685 Chr.3: 39264187-39264228 on Build GRCh38
rs574775684 Chr.3: 39264278-39264278 on Build GRCh38
rs965160417 Chr.3: 39264158-39264158 on Build GRCh38
rs939385012 Chr.3: 39264339-39264339 on Build GRCh38
rs965767737 Chr.3: 39264145-39264145 on Build GRCh38
rs560799610 Chr.3: 39264291-39264291 on Build GRCh38
rs1004979316 Chr.3: 39264240-39264240 on Build GRCh38
rs761999825 Chr.3: 39264211-39264211 on Build GRCh38
rs940752788 Chr.3: 39264502-39264502 on Build GRCh38
rs538792233 Chr.3: 39264096-39264096 on Build GRCh38
rs114770245 Chr.3: 39264197-39264197 on Build GRCh38
rs182434611 Chr.3: 39264113-39264113 on Build GRCh38
rs1046625581 Chr.3: 39264490-39264490 on Build GRCh38
rs947969080 Chr.3: 39264468-39264468 on Build GRCh38
rs750429036 Chr.3: 39264344-39264344 on Build GRCh38
rs1056449211 Chr.3: 39264358-39264358 on Build GRCh38
rs953638909 Chr.3: 39264314-39264314 on Build GRCh38
rs1029233094 Chr.3: 39264312-39264312 on Build GRCh38
rs79566019 Chr.3: 39264126-39264126 on Build GRCh38
rs754432710 Chr.3: 39264462-39264462 on Build GRCh38
rs114764201 Chr.3: 39264049-39264049 on Build GRCh38

View Full Product Details