Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10726382-10726758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1C2
Entrez Gene ID
245973
Gene Name
ATPase H+ transporting V1 subunit C2
Gene Aliases
ATP6C2, VMA5
UniGene
Hs.580464
Gene Chromosome Location
Chr.2: 10720974-10785110 on Build GRCh38
Transcripts
NM_001039362.1 NM_144583.3 XM_011510339.2 XM_011510340.2 XM_011510341.2 XM_017003745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
345210 c.186C>T p.T62T Chr.2: 10726558-10726558 on Build GRCh38
3565043 c.146C>T p.S49F Chr.2: 10726518-10726518 on Build GRCh38
345211 c.186C>T p.T62T Chr.2: 10726558-10726558 on Build GRCh38
3565044 c.146C>T p.S49F Chr.2: 10726518-10726518 on Build GRCh38
5973329 c.180C>T p.L60L Chr.2: 10726552-10726552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763444135 Chr.2: 10726479-10726479 on Build GRCh38
rs998816215 Chr.2: 10726537-10726537 on Build GRCh38
rs910585160 Chr.2: 10726451-10726451 on Build GRCh38
rs199712045 Chr.2: 10726477-10726477 on Build GRCh38
rs775182175 Chr.2: 10726717-10726717 on Build GRCh38
rs116376282 Chr.2: 10726475-10726475 on Build GRCh38
rs375061582 Chr.2: 10726593-10726593 on Build GRCh38
rs894200899 Chr.2: 10726519-10726519 on Build GRCh38
rs905187086 Chr.2: 10726711-10726711 on Build GRCh38
rs768154866 Chr.2: 10726595-10726595 on Build GRCh38
rs575889877 Chr.2: 10726594-10726594 on Build GRCh38
rs758730321 Chr.2: 10726518-10726518 on Build GRCh38
rs986271480 Chr.2: 10726449-10726449 on Build GRCh38
rs867071617 Chr.2: 10726540-10726540 on Build GRCh38
rs373910621 Chr.2: 10726491-10726491 on Build GRCh38
rs755638200 Chr.2: 10726673-10726673 on Build GRCh38
rs368938552 Chr.2: 10726618-10726618 on Build GRCh38
rs748192171 Chr.2: 10726552-10726552 on Build GRCh38
rs1032883322 Chr.2: 10726432-10726432 on Build GRCh38
rs765681984 Chr.2: 10726510-10726510 on Build GRCh38
rs934463092 Chr.2: 10726666-10726666 on Build GRCh38
rs766500946 Chr.2: 10726540-10726542 on Build GRCh38
rs117482977 Chr.2: 10726702-10726702 on Build GRCh38
rs763281382 Chr.2: 10726615-10726615 on Build GRCh38
rs745727972 Chr.2: 10726577-10726577 on Build GRCh38
rs780203954 Chr.2: 10726523-10726523 on Build GRCh38
rs778967006 Chr.2: 10726574-10726574 on Build GRCh38
rs976144885 Chr.2: 10726629-10726629 on Build GRCh38
rs762042840 Chr.2: 10726442-10726442 on Build GRCh38
rs771043793 Chr.2: 10726553-10726553 on Build GRCh38
rs141081826 Chr.2: 10726476-10726476 on Build GRCh38
rs773537361 Chr.2: 10726451-10726452 on Build GRCh38
rs896641222 Chr.2: 10726730-10726730 on Build GRCh38
rs372197178 Chr.2: 10726601-10726601 on Build GRCh38
rs115321601 Chr.2: 10726528-10726528 on Build GRCh38
rs200067399 Chr.2: 10726453-10726453 on Build GRCh38
rs1013893593 Chr.2: 10726524-10726524 on Build GRCh38
rs762338262 Chr.2: 10726507-10726507 on Build GRCh38
rs919294661 Chr.2: 10726677-10726677 on Build GRCh38
rs975078677 Chr.2: 10726520-10726520 on Build GRCh38
rs967244495 Chr.2: 10726404-10726404 on Build GRCh38
rs371894347 Chr.2: 10726579-10726579 on Build GRCh38
rs376785108 Chr.2: 10726605-10726605 on Build GRCh38
rs1017209920 Chr.2: 10726734-10726734 on Build GRCh38
rs1003663358 Chr.2: 10726712-10726712 on Build GRCh38
rs371578415 Chr.2: 10726435-10726435 on Build GRCh38
rs949832956 Chr.2: 10726739-10726739 on Build GRCh38
rs781334191 Chr.2: 10726551-10726551 on Build GRCh38
rs766775983 Chr.2: 10726617-10726617 on Build GRCh38
rs957423817 Chr.2: 10726444-10726444 on Build GRCh38
rs1023658001 Chr.2: 10726706-10726706 on Build GRCh38
rs183059615 Chr.2: 10726501-10726501 on Build GRCh38
rs1055679599 Chr.2: 10726725-10726725 on Build GRCh38
rs1054305083 Chr.2: 10726698-10726698 on Build GRCh38
rs775207898 Chr.2: 10726483-10726483 on Build GRCh38
rs566877820 Chr.2: 10726465-10726465 on Build GRCh38
rs750835309 Chr.2: 10726512-10726512 on Build GRCh38
rs201965618 Chr.2: 10726550-10726550 on Build GRCh38
rs573540830 Chr.2: 10726716-10726716 on Build GRCh38
rs771666410 Chr.2: 10726482-10726482 on Build GRCh38
rs967003799 Chr.2: 10726529-10726529 on Build GRCh38
rs150280505 Chr.2: 10726718-10726718 on Build GRCh38
rs543303933 Chr.2: 10726612-10726612 on Build GRCh38
rs903061696 Chr.2: 10726652-10726652 on Build GRCh38
rs977290546 Chr.2: 10726433-10726433 on Build GRCh38
rs776139960 Chr.2: 10726596-10726596 on Build GRCh38
rs188491104 Chr.2: 10726502-10726502 on Build GRCh38
rs775090046 Chr.2: 10726592-10726592 on Build GRCh38
rs749249093 Chr.2: 10726468-10726468 on Build GRCh38

View Full Product Details