Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48376357-48376926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN2
Entrez Gene ID
3344
Gene Name
forkhead box N2
Gene Aliases
HTLF
UniGene
Hs.468478
Gene Chromosome Location
Chr.2: 48314640-48379295 on Build GRCh38
Transcripts
NM_002158.3 XM_005264282.2 XM_005264283.2 XM_006712001.3 XM_006712002.3 XM_011532808.2 XM_017003956.1 XM_017003957.1 XM_017003958.1 XM_017003959.1

SNP Detail

SNP ID Chromosome Location
rs565619450 Chr.2: 48376773-48376774 on Build GRCh38
rs941700944 Chr.2: 48376615-48376615 on Build GRCh38
rs375924866 Chr.2: 48376781-48376782 on Build GRCh38
rs560314200 Chr.2: 48376580-48376580 on Build GRCh38
rs185959136 Chr.2: 48376676-48376676 on Build GRCh38
rs976240034 Chr.2: 48376585-48376585 on Build GRCh38
rs939063690 Chr.2: 48376880-48376880 on Build GRCh38
rs140433382 Chr.2: 48376523-48376523 on Build GRCh38
rs555635102 Chr.2: 48376717-48376717 on Build GRCh38
rs948898687 Chr.2: 48376752-48376752 on Build GRCh38
rs908959922 Chr.2: 48376842-48376842 on Build GRCh38
rs186326613 Chr.2: 48376766-48376766 on Build GRCh38
rs758611196 Chr.2: 48376859-48376859 on Build GRCh38
rs953212364 Chr.2: 48376590-48376590 on Build GRCh38
rs1013429754 Chr.2: 48376467-48376467 on Build GRCh38
rs950983644 Chr.2: 48376818-48376818 on Build GRCh38
rs927714680 Chr.2: 48376799-48376799 on Build GRCh38
rs549033105 Chr.2: 48376559-48376560 on Build GRCh38
rs977419161 Chr.2: 48376643-48376643 on Build GRCh38
rs556228989 Chr.2: 48376886-48376886 on Build GRCh38
rs916167141 Chr.2: 48376735-48376735 on Build GRCh38
rs180999996 Chr.2: 48376723-48376723 on Build GRCh38
rs35454093 Chr.2: 48376576-48376576 on Build GRCh38
rs547460152 Chr.2: 48376598-48376598 on Build GRCh38
rs752985661 Chr.2: 48376638-48376638 on Build GRCh38
rs948913328 Chr.2: 48376451-48376451 on Build GRCh38
rs537728072 Chr.2: 48376821-48376821 on Build GRCh38
rs916521572 Chr.2: 48376634-48376636 on Build GRCh38
rs115448144 Chr.2: 48376519-48376519 on Build GRCh38
rs879366920 Chr.2: 48376522-48376523 on Build GRCh38
rs1027996856 Chr.2: 48376583-48376583 on Build GRCh38
rs964944627 Chr.2: 48376759-48376759 on Build GRCh38
rs949257548 Chr.2: 48376667-48376667 on Build GRCh38
rs767233272 Chr.2: 48376651-48376651 on Build GRCh38
rs765511651 Chr.2: 48376630-48376630 on Build GRCh38
rs946763943 Chr.2: 48376874-48376887 on Build GRCh38
rs983605750 Chr.2: 48376825-48376825 on Build GRCh38
rs1055121194 Chr.2: 48376410-48376410 on Build GRCh38
rs963139069 Chr.2: 48376857-48376857 on Build GRCh38
rs571689900 Chr.2: 48376518-48376518 on Build GRCh38
rs990388790 Chr.2: 48376732-48376732 on Build GRCh38
rs1055081810 Chr.2: 48376417-48376417 on Build GRCh38
rs906316943 Chr.2: 48376493-48376493 on Build GRCh38
rs985984153 Chr.2: 48376606-48376606 on Build GRCh38
rs144376062 Chr.2: 48376633-48376633 on Build GRCh38
rs893185386 Chr.2: 48376726-48376730 on Build GRCh38
rs190729513 Chr.2: 48376721-48376721 on Build GRCh38
rs879214560 Chr.2: 48376562-48376562 on Build GRCh38
rs1046330914 Chr.2: 48376673-48376673 on Build GRCh38
rs902366482 Chr.2: 48376709-48376709 on Build GRCh38
rs994729738 Chr.2: 48376568-48376568 on Build GRCh38
rs1053645685 Chr.2: 48376724-48376724 on Build GRCh38
rs113546529 Chr.2: 48376505-48376505 on Build GRCh38
rs908955513 Chr.2: 48376613-48376613 on Build GRCh38
rs778938125 Chr.2: 48376483-48376483 on Build GRCh38
rs374917031 Chr.2: 48376515-48376515 on Build GRCh38
rs1044769387 Chr.2: 48376476-48376476 on Build GRCh38
rs757248461 Chr.2: 48376554-48376554 on Build GRCh38
rs894611786 Chr.2: 48376423-48376423 on Build GRCh38
rs897460530 Chr.2: 48376894-48376894 on Build GRCh38
rs553382606 Chr.2: 48376511-48376511 on Build GRCh38
rs1017534766 Chr.2: 48376746-48376746 on Build GRCh38
rs941704424 Chr.2: 48376414-48376414 on Build GRCh38
rs994927415 Chr.2: 48376901-48376901 on Build GRCh38
rs924598531 Chr.2: 48376705-48376705 on Build GRCh38

View Full Product Details