Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70160493-70160861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf42
Entrez Gene ID
54980
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
-
UniGene
Hs.720049
Gene Chromosome Location
Chr.2: 70149885-70191071 on Build GRCh38
Transcripts
NM_017880.1 XM_005264389.2 XM_005264390.2 XM_005264391.2 XM_011532937.2 XM_017004398.1 XM_017004399.1 XM_017004400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4464897 c.1358C>T p.P453L Chr.2: 70160783-70160783 on Build GRCh38
3910521 c.1371C>T p.T457T Chr.2: 70160770-70160770 on Build GRCh38
5814213 c.1496T>G p.L499R Chr.2: 70160645-70160645 on Build GRCh38
6402778 c.1391G>T p.R464L Chr.2: 70160750-70160750 on Build GRCh38
1022495 c.1382T>A p.I461N Chr.2: 70160759-70160759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368144536 Chr.2: 70160815-70160815 on Build GRCh38
rs770059802 Chr.2: 70160807-70160808 on Build GRCh38
rs138062525 Chr.2: 70160689-70160689 on Build GRCh38
rs150172838 Chr.2: 70160685-70160685 on Build GRCh38
rs200580709 Chr.2: 70160658-70160658 on Build GRCh38
rs976285430 Chr.2: 70160763-70160763 on Build GRCh38
rs143487383 Chr.2: 70160686-70160686 on Build GRCh38
rs546866294 Chr.2: 70160570-70160570 on Build GRCh38
rs762525659 Chr.2: 70160612-70160612 on Build GRCh38
rs183733229 Chr.2: 70160657-70160657 on Build GRCh38
rs1002018871 Chr.2: 70160680-70160680 on Build GRCh38
rs769897997 Chr.2: 70160684-70160684 on Build GRCh38
rs996046674 Chr.2: 70160756-70160756 on Build GRCh38
rs760716205 Chr.2: 70160630-70160630 on Build GRCh38
rs752714230 Chr.2: 70160709-70160709 on Build GRCh38
rs755379857 Chr.2: 70160700-70160700 on Build GRCh38
rs201536144 Chr.2: 70160792-70160792 on Build GRCh38
rs755981442 Chr.2: 70160768-70160768 on Build GRCh38
rs773427484 Chr.2: 70160808-70160808 on Build GRCh38
rs578224129 Chr.2: 70160567-70160567 on Build GRCh38
rs754345567 Chr.2: 70160698-70160698 on Build GRCh38
rs559507367 Chr.2: 70160679-70160679 on Build GRCh38
rs748277376 Chr.2: 70160579-70160579 on Build GRCh38
rs552906288 Chr.2: 70160769-70160769 on Build GRCh38
rs943404887 Chr.2: 70160563-70160563 on Build GRCh38
rs979692827 Chr.2: 70160744-70160744 on Build GRCh38
rs944066431 Chr.2: 70160572-70160572 on Build GRCh38
rs749309237 Chr.2: 70160781-70160781 on Build GRCh38
rs563802318 Chr.2: 70160592-70160592 on Build GRCh38
rs775176259 Chr.2: 70160597-70160597 on Build GRCh38
rs368402264 Chr.2: 70160687-70160687 on Build GRCh38
rs1040468938 Chr.2: 70160565-70160565 on Build GRCh38
rs1014659699 Chr.2: 70160725-70160725 on Build GRCh38
rs763474631 Chr.2: 70160618-70160618 on Build GRCh38
rs148563389 Chr.2: 70160655-70160655 on Build GRCh38
rs199935818 Chr.2: 70160750-70160750 on Build GRCh38
rs372345601 Chr.2: 70160787-70160787 on Build GRCh38
rs375469438 Chr.2: 70160582-70160582 on Build GRCh38
rs765416025 Chr.2: 70160706-70160706 on Build GRCh38
rs886989681 Chr.2: 70160608-70160608 on Build GRCh38
rs192357786 Chr.2: 70160581-70160581 on Build GRCh38
rs780517544 Chr.2: 70160660-70160660 on Build GRCh38
rs752412946 Chr.2: 70160765-70160765 on Build GRCh38
rs191212969 Chr.2: 70160738-70160738 on Build GRCh38
rs578262819 Chr.2: 70160836-70160836 on Build GRCh38
rs35340595 Chr.2: 70160681-70160681 on Build GRCh38
rs779218955 Chr.2: 70160577-70160577 on Build GRCh38
rs751232964 Chr.2: 70160619-70160619 on Build GRCh38
rs1005487028 Chr.2: 70160638-70160638 on Build GRCh38
rs762269994 Chr.2: 70160594-70160597 on Build GRCh38
rs202118682 Chr.2: 70160814-70160814 on Build GRCh38
rs146789788 Chr.2: 70160682-70160682 on Build GRCh38
rs747590323 Chr.2: 70160806-70160806 on Build GRCh38
rs755115145 Chr.2: 70160667-70160667 on Build GRCh38
rs943498172 Chr.2: 70160757-70160757 on Build GRCh38
rs558480162 Chr.2: 70160831-70160831 on Build GRCh38
rs770765145 Chr.2: 70160580-70160580 on Build GRCh38
rs947198678 Chr.2: 70160591-70160591 on Build GRCh38
rs142861699 Chr.2: 70160664-70160664 on Build GRCh38
rs114745930 Chr.2: 70160690-70160690 on Build GRCh38
rs6722591 Chr.2: 70160822-70160822 on Build GRCh38
rs761316823 Chr.2: 70160642-70160642 on Build GRCh38
rs550388807 Chr.2: 70160691-70160691 on Build GRCh38
rs371180319 Chr.2: 70160826-70160826 on Build GRCh38

View Full Product Details