Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110637751-110638295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
UniGene
Hs.469649
Gene Chromosome Location
Chr.2: 110637698-110678107 on Build GRCh38
Transcripts
NM_001278616.1 NM_001278617.1 NM_004336.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5887664 c.3193A>G p.K1065E Chr.2: 110638029-110638029 on Build GRCh38
6423696 c.3109A>T p.N1037Y Chr.2: 110638113-110638113 on Build GRCh38
4084165 c.3222T>G p.I1074M Chr.2: 110638000-110638000 on Build GRCh38
6399371 c.3208C>T p.R1070C Chr.2: 110638014-110638014 on Build GRCh38
6087416 c.3162G>A p.L1054L Chr.2: 110638060-110638060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026865593 Chr.2: 110637954-110637954 on Build GRCh38
rs748999502 Chr.2: 110637977-110637977 on Build GRCh38
rs375754310 Chr.2: 110637921-110637921 on Build GRCh38
rs372788700 Chr.2: 110637978-110637978 on Build GRCh38
rs55839851 Chr.2: 110638091-110638091 on Build GRCh38
rs761394321 Chr.2: 110637980-110637980 on Build GRCh38
rs181020604 Chr.2: 110638101-110638101 on Build GRCh38
rs1009345099 Chr.2: 110638107-110638107 on Build GRCh38
rs779965899 Chr.2: 110637972-110637972 on Build GRCh38
rs751673537 Chr.2: 110638031-110638031 on Build GRCh38
rs1018010189 Chr.2: 110637869-110637869 on Build GRCh38
rs765636832 Chr.2: 110638013-110638013 on Build GRCh38
rs758757177 Chr.2: 110638201-110638201 on Build GRCh38
rs1001829222 Chr.2: 110638211-110638211 on Build GRCh38
rs763226529 Chr.2: 110638020-110638020 on Build GRCh38
rs757215922 Chr.2: 110637960-110637960 on Build GRCh38
rs748697577 Chr.2: 110638136-110638136 on Build GRCh38
rs201215781 Chr.2: 110638159-110638159 on Build GRCh38
rs368415851 Chr.2: 110637923-110637923 on Build GRCh38
rs199887786 Chr.2: 110638130-110638130 on Build GRCh38
rs751652650 Chr.2: 110637957-110637957 on Build GRCh38
rs781202575 Chr.2: 110637961-110637961 on Build GRCh38
rs908709416 Chr.2: 110637865-110637865 on Build GRCh38
rs116334746 Chr.2: 110638220-110638220 on Build GRCh38
rs991198254 Chr.2: 110638177-110638177 on Build GRCh38
rs768074987 Chr.2: 110638107-110638108 on Build GRCh38
rs528194626 Chr.2: 110638133-110638133 on Build GRCh38
rs767456937 Chr.2: 110638056-110638056 on Build GRCh38
rs587776642 Chr.2: 110638153-110638153 on Build GRCh38
rs938414105 Chr.2: 110638162-110638162 on Build GRCh38
rs963345687 Chr.2: 110637891-110637891 on Build GRCh38
rs757380682 Chr.2: 110638041-110638041 on Build GRCh38
rs974004871 Chr.2: 110637922-110637922 on Build GRCh38
rs760001777 Chr.2: 110637999-110637999 on Build GRCh38
rs890527649 Chr.2: 110637817-110637817 on Build GRCh38
rs143604436 Chr.2: 110638210-110638210 on Build GRCh38
rs777222739 Chr.2: 110637994-110637994 on Build GRCh38
rs1007597594 Chr.2: 110637830-110637830 on Build GRCh38
rs756570200 Chr.2: 110637796-110637796 on Build GRCh38
rs769743072 Chr.2: 110637969-110637969 on Build GRCh38
rs528710907 Chr.2: 110637891-110637891 on Build GRCh38
rs866762120 Chr.2: 110638047-110638047 on Build GRCh38
rs1022475028 Chr.2: 110638142-110638142 on Build GRCh38
rs191292475 Chr.2: 110638014-110638014 on Build GRCh38
rs866272996 Chr.2: 110637946-110637946 on Build GRCh38
rs536675264 Chr.2: 110638252-110638252 on Build GRCh38
rs992296947 Chr.2: 110637853-110637853 on Build GRCh38
rs771429383 Chr.2: 110637991-110637991 on Build GRCh38
rs199818479 Chr.2: 110638026-110638026 on Build GRCh38
rs777886671 Chr.2: 110637943-110637943 on Build GRCh38
rs756101035 Chr.2: 110638097-110638097 on Build GRCh38
rs76458371 Chr.2: 110638212-110638212 on Build GRCh38
rs781651172 Chr.2: 110638132-110638132 on Build GRCh38
rs745745049 Chr.2: 110637966-110637966 on Build GRCh38
rs1030423503 Chr.2: 110637787-110637787 on Build GRCh38

View Full Product Details