Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206162179-206162631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1B2
Entrez Gene ID
1933
Gene Name
eukaryotic translation elongation factor 1 beta 2
Gene Aliases
EEF1B, EEF1B1, EF1B
UniGene
Hs.421608
Gene Chromosome Location
Chr.2: 206159594-206162929 on Build GRCh38
Transcripts
NM_001037663.1 NM_001959.3 NM_021121.3 NR_002590.1

Gene Symbol
SNORA41
Entrez Gene ID
619569
Gene Name
small nucleolar RNA, H/ACA box 41
Gene Aliases
ACA41, SNORA41A
UniGene
Hs.742407
Gene Chromosome Location
Chr.2: 206162228-206162359 on Build GRCh38
Transcripts
NM_001037663.1 NM_001959.3 NM_021121.3 NR_002590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260642 c.398-1G>T p.? Chr.2: 206162488-206162488 on Build GRCh38
1015306 c.468G>A p.A156A Chr.2: 206162559-206162559 on Build GRCh38
5777691 c.475G>A p.E159K Chr.2: 206162566-206162566 on Build GRCh38
1015307 c.498A>C p.Q166H Chr.2: 206162589-206162589 on Build GRCh38
6156457 c.451G>A p.D151N Chr.2: 206162542-206162542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911730164 Chr.2: 206162587-206162587 on Build GRCh38
rs753393059 Chr.2: 206162604-206162604 on Build GRCh38
rs768480936 Chr.2: 206162227-206162227 on Build GRCh38
rs759689793 Chr.2: 206162561-206162561 on Build GRCh38
rs35841485 Chr.2: 206162312-206162313 on Build GRCh38
rs774061053 Chr.2: 206162398-206162398 on Build GRCh38
rs751363521 Chr.2: 206162308-206162308 on Build GRCh38
rs768605716 Chr.2: 206162390-206162390 on Build GRCh38
rs774742946 Chr.2: 206162441-206162442 on Build GRCh38
rs760588035 Chr.2: 206162312-206162315 on Build GRCh38
rs769022552 Chr.2: 206162239-206162239 on Build GRCh38
rs1004150430 Chr.2: 206162413-206162413 on Build GRCh38
rs768520410 Chr.2: 206162315-206162317 on Build GRCh38
rs751944979 Chr.2: 206162584-206162584 on Build GRCh38
rs377381799 Chr.2: 206162590-206162590 on Build GRCh38
rs368216880 Chr.2: 206162234-206162234 on Build GRCh38
rs766564190 Chr.2: 206162558-206162558 on Build GRCh38
rs759926959 Chr.2: 206162513-206162514 on Build GRCh38
rs764623938 Chr.2: 206162582-206162582 on Build GRCh38
rs889571248 Chr.2: 206162492-206162492 on Build GRCh38
rs745597653 Chr.2: 206162210-206162211 on Build GRCh38
rs147791783 Chr.2: 206162559-206162559 on Build GRCh38
rs749585121 Chr.2: 206162301-206162301 on Build GRCh38
rs781193837 Chr.2: 206162204-206162204 on Build GRCh38
rs774378420 Chr.2: 206162499-206162499 on Build GRCh38
rs756662684 Chr.2: 206162218-206162218 on Build GRCh38
rs199531068 Chr.2: 206162456-206162456 on Build GRCh38
rs758644125 Chr.2: 206162347-206162347 on Build GRCh38
rs760874509 Chr.2: 206162441-206162441 on Build GRCh38
rs753100752 Chr.2: 206162472-206162472 on Build GRCh38
rs762922135 Chr.2: 206162290-206162290 on Build GRCh38
rs756190698 Chr.2: 206162330-206162330 on Build GRCh38
rs748811604 Chr.2: 206162494-206162494 on Build GRCh38
rs755489934 Chr.2: 206162460-206162460 on Build GRCh38
rs778229183 Chr.2: 206162477-206162477 on Build GRCh38
rs181533336 Chr.2: 206162475-206162475 on Build GRCh38
rs761690328 Chr.2: 206162403-206162403 on Build GRCh38
rs771886327 Chr.2: 206162296-206162297 on Build GRCh38
rs564396403 Chr.2: 206162523-206162523 on Build GRCh38
rs375707487 Chr.2: 206162448-206162448 on Build GRCh38
rs750469854 Chr.2: 206162215-206162215 on Build GRCh38
rs941260635 Chr.2: 206162381-206162381 on Build GRCh38
rs1035207797 Chr.2: 206162437-206162437 on Build GRCh38
rs765160465 Chr.2: 206162562-206162562 on Build GRCh38
rs766553196 Chr.2: 206162351-206162397 on Build GRCh38
rs760925547 Chr.2: 206162554-206162554 on Build GRCh38
rs779876137 Chr.2: 206162598-206162598 on Build GRCh38
rs763003764 Chr.2: 206162335-206162335 on Build GRCh38
rs768745279 Chr.2: 206162312-206162312 on Build GRCh38
rs777073785 Chr.2: 206162300-206162304 on Build GRCh38
rs758821800 Chr.2: 206162575-206162575 on Build GRCh38
rs1042659403 Chr.2: 206162351-206162351 on Build GRCh38
rs775123295 Chr.2: 206162370-206162370 on Build GRCh38
rs767176998 Chr.2: 206162407-206162407 on Build GRCh38
rs762085449 Chr.2: 206162434-206162434 on Build GRCh38
rs773339771 Chr.2: 206162328-206162329 on Build GRCh38
rs745442409 Chr.2: 206162355-206162355 on Build GRCh38
rs775569178 Chr.2: 206162283-206162283 on Build GRCh38
rs771959213 Chr.2: 206162515-206162515 on Build GRCh38
rs774570428 Chr.2: 206162328-206162328 on Build GRCh38
rs374517025 Chr.2: 206162341-206162341 on Build GRCh38
rs576509470 Chr.2: 206162267-206162267 on Build GRCh38
rs893814080 Chr.2: 206162323-206162323 on Build GRCh38
rs372481081 Chr.2: 206162279-206162279 on Build GRCh38
rs902664690 Chr.2: 206162379-206162379 on Build GRCh38
rs745576823 Chr.2: 206162483-206162483 on Build GRCh38
rs369521473 Chr.2: 206162259-206162259 on Build GRCh38
rs2359156 Chr.2: 206162574-206162574 on Build GRCh38
rs780051329 Chr.2: 206162336-206162336 on Build GRCh38
rs376795945 Chr.2: 206162281-206162281 on Build GRCh38
rs974834268 Chr.2: 206162596-206162596 on Build GRCh38
rs779753319 Chr.2: 206162493-206162493 on Build GRCh38
rs769438741 Chr.2: 206162369-206162369 on Build GRCh38
rs748954510 Chr.2: 206162303-206162306 on Build GRCh38
rs747977038 Chr.2: 206162500-206162500 on Build GRCh38
rs369574321 Chr.2: 206162339-206162339 on Build GRCh38
rs562321830 Chr.2: 206162353-206162353 on Build GRCh38
rs767755310 Chr.2: 206162311-206162311 on Build GRCh38
rs773436421 Chr.2: 206162440-206162440 on Build GRCh38
rs776057247 Chr.2: 206162273-206162273 on Build GRCh38
rs754402478 Chr.2: 206162337-206162337 on Build GRCh38
rs200624747 Chr.2: 206162467-206162467 on Build GRCh38
rs755827613 Chr.2: 206162484-206162484 on Build GRCh38
rs768003462 Chr.2: 206162497-206162497 on Build GRCh38
rs200814967 Chr.2: 206162566-206162566 on Build GRCh38
rs749302392 Chr.2: 206162388-206162388 on Build GRCh38

View Full Product Details