Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73715497-73716070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMSAN1
Entrez Gene ID
91748
Gene Name
ELM2 and Myb/SANT domain containing 1
Gene Aliases
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541
UniGene
Hs.594157
Gene Chromosome Location
Chr.14: 73715122-73790285 on Build GRCh38
Transcripts
NM_001043318.2 NM_194278.3 XM_005268204.1 XM_005268205.1 XM_005268206.1

SNP Detail

SNP ID Chromosome Location
rs571838914 Chr.14: 73715881-73715881 on Build GRCh38
rs879090632 Chr.14: 73715775-73715775 on Build GRCh38
rs752176404 Chr.14: 73715771-73715771 on Build GRCh38
rs186833385 Chr.14: 73715930-73715930 on Build GRCh38
rs372924077 Chr.14: 73715672-73715672 on Build GRCh38
rs976329830 Chr.14: 73715523-73715523 on Build GRCh38
rs191276191 Chr.14: 73715548-73715548 on Build GRCh38
rs555699509 Chr.14: 73715546-73715546 on Build GRCh38
rs951519528 Chr.14: 73715952-73715952 on Build GRCh38
rs962198153 Chr.14: 73715855-73715855 on Build GRCh38
rs756886703 Chr.14: 73715884-73715884 on Build GRCh38
rs1017306321 Chr.14: 73715915-73715915 on Build GRCh38
rs1015141384 Chr.14: 73715805-73715805 on Build GRCh38
rs192370394 Chr.14: 73715934-73715934 on Build GRCh38
rs890089261 Chr.14: 73715539-73715539 on Build GRCh38
rs765596746 Chr.14: 73715589-73715589 on Build GRCh38
rs1007256201 Chr.14: 73715554-73715554 on Build GRCh38
rs572412894 Chr.14: 73715637-73715637 on Build GRCh38
rs75154871 Chr.14: 73715875-73715875 on Build GRCh38
rs933639429 Chr.14: 73715577-73715577 on Build GRCh38
rs572828050 Chr.14: 73716031-73716031 on Build GRCh38
rs963138458 Chr.14: 73715603-73715603 on Build GRCh38
rs150370323 Chr.14: 73716027-73716027 on Build GRCh38
rs750760526 Chr.14: 73715666-73715666 on Build GRCh38
rs1056375097 Chr.14: 73715747-73715747 on Build GRCh38
rs528708826 Chr.14: 73715535-73715535 on Build GRCh38
rs890402145 Chr.14: 73715900-73715900 on Build GRCh38
rs1017753899 Chr.14: 73715578-73715578 on Build GRCh38
rs140371962 Chr.14: 73715776-73715776 on Build GRCh38
rs1012208892 Chr.14: 73715767-73715770 on Build GRCh38
rs112581359 Chr.14: 73715974-73715974 on Build GRCh38
rs1008274567 Chr.14: 73715871-73715871 on Build GRCh38
rs367993961 Chr.14: 73715889-73715890 on Build GRCh38
rs112283607 Chr.14: 73715592-73715592 on Build GRCh38
rs935287714 Chr.14: 73715723-73715723 on Build GRCh38
rs1006292832 Chr.14: 73715803-73715803 on Build GRCh38
rs947487622 Chr.14: 73715853-73715853 on Build GRCh38
rs547240400 Chr.14: 73715594-73715594 on Build GRCh38
rs887838215 Chr.14: 73715781-73715781 on Build GRCh38
rs1036332691 Chr.14: 73715976-73715976 on Build GRCh38
rs188061175 Chr.14: 73716033-73716033 on Build GRCh38
rs991890611 Chr.14: 73716009-73716009 on Build GRCh38
rs1007635705 Chr.14: 73715904-73715904 on Build GRCh38
rs1043566993 Chr.14: 73715685-73715685 on Build GRCh38
rs546958019 Chr.14: 73715989-73715989 on Build GRCh38
rs974206465 Chr.14: 73715617-73715617 on Build GRCh38
rs910952374 Chr.14: 73715638-73715638 on Build GRCh38
rs1045097145 Chr.14: 73715995-73715995 on Build GRCh38
rs182677962 Chr.14: 73715830-73715830 on Build GRCh38
rs567939293 Chr.14: 73715888-73715889 on Build GRCh38
rs902334651 Chr.14: 73715700-73715700 on Build GRCh38
rs923808708 Chr.14: 73715971-73715971 on Build GRCh38
rs551249082 Chr.14: 73715982-73715982 on Build GRCh38
rs533240421 Chr.14: 73715827-73715827 on Build GRCh38
rs138105201 Chr.14: 73716040-73716040 on Build GRCh38
rs985618497 Chr.14: 73715614-73715614 on Build GRCh38
rs1027078451 Chr.14: 73715621-73715621 on Build GRCh38
rs1043169860 Chr.14: 73715862-73715862 on Build GRCh38
rs978817078 Chr.14: 73716044-73716044 on Build GRCh38
rs763461457 Chr.14: 73715710-73715710 on Build GRCh38
rs1052104066 Chr.14: 73715890-73715890 on Build GRCh38
rs983417591 Chr.14: 73715694-73715694 on Build GRCh38
rs1026040700 Chr.14: 73715931-73715931 on Build GRCh38
rs73297453 Chr.14: 73715754-73715754 on Build GRCh38
rs74572727 Chr.14: 73715925-73715925 on Build GRCh38
rs922295224 Chr.14: 73715531-73715531 on Build GRCh38
rs948062108 Chr.14: 73715992-73715992 on Build GRCh38
rs946566941 Chr.14: 73715684-73715684 on Build GRCh38
rs960355915 Chr.14: 73715978-73715978 on Build GRCh38
rs887775734 Chr.14: 73716014-73716015 on Build GRCh38

View Full Product Details