Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 89161990-89162529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN3
Entrez Gene ID
1112
Gene Name
forkhead box N3
Gene Aliases
C14orf116, CHES1, PRO1635
UniGene
Hs.434286
Gene Chromosome Location
Chr.14: 89156172-89619150 on Build GRCh38
Transcripts
NM_001085471.1 NM_005197.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4961315 c.1439G>A p.G480E Chr.14: 89162448-89162448 on Build GRCh38
6666429 c.1406A>C p.*469S Chr.14: 89162415-89162415 on Build GRCh38
4465894 c.1338C>T p.S446S Chr.14: 89162483-89162483 on Build GRCh38
4150865 c.1421C>A p.T474N Chr.14: 89162466-89162466 on Build GRCh38
4150867 c.1420A>C p.T474P Chr.14: 89162467-89162467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554217030 Chr.14: 89162078-89162078 on Build GRCh38
rs375063809 Chr.14: 89162066-89162066 on Build GRCh38
rs756634006 Chr.14: 89162116-89162116 on Build GRCh38
rs562009009 Chr.14: 89162159-89162159 on Build GRCh38
rs761485419 Chr.14: 89162486-89162486 on Build GRCh38
rs945847341 Chr.14: 89162233-89162233 on Build GRCh38
rs769831252 Chr.14: 89162457-89162457 on Build GRCh38
rs532911597 Chr.14: 89162352-89162352 on Build GRCh38
rs538103366 Chr.14: 89162035-89162035 on Build GRCh38
rs369131306 Chr.14: 89162136-89162136 on Build GRCh38
rs182736587 Chr.14: 89162073-89162073 on Build GRCh38
rs370548969 Chr.14: 89162411-89162411 on Build GRCh38
rs893749602 Chr.14: 89162230-89162248 on Build GRCh38
rs746187182 Chr.14: 89162408-89162410 on Build GRCh38
rs370136184 Chr.14: 89162461-89162461 on Build GRCh38
rs557739226 Chr.14: 89162046-89162046 on Build GRCh38
rs147844828 Chr.14: 89162487-89162487 on Build GRCh38
rs764885051 Chr.14: 89162173-89162173 on Build GRCh38
rs780069905 Chr.14: 89162284-89162284 on Build GRCh38
rs930907117 Chr.14: 89162131-89162131 on Build GRCh38
rs1028994512 Chr.14: 89162025-89162025 on Build GRCh38
rs926965377 Chr.14: 89162169-89162169 on Build GRCh38
rs937108640 Chr.14: 89162186-89162187 on Build GRCh38
rs984226556 Chr.14: 89162143-89162143 on Build GRCh38
rs188906675 Chr.14: 89162130-89162130 on Build GRCh38
rs373751435 Chr.14: 89162443-89162443 on Build GRCh38
rs141894477 Chr.14: 89162391-89162391 on Build GRCh38
rs972323441 Chr.14: 89162092-89162092 on Build GRCh38
rs183189525 Chr.14: 89162181-89162181 on Build GRCh38
rs1041621903 Chr.14: 89162243-89162243 on Build GRCh38
rs372726798 Chr.14: 89162065-89162065 on Build GRCh38
rs750854893 Chr.14: 89162365-89162365 on Build GRCh38
rs748392128 Chr.14: 89162438-89162438 on Build GRCh38
rs187523861 Chr.14: 89162270-89162270 on Build GRCh38
rs193241414 Chr.14: 89162150-89162150 on Build GRCh38
rs987562383 Chr.14: 89162039-89162039 on Build GRCh38
rs776533234 Chr.14: 89162450-89162450 on Build GRCh38
rs753923069 Chr.14: 89162471-89162471 on Build GRCh38
rs375570570 Chr.14: 89162460-89162460 on Build GRCh38
rs1065101 Chr.14: 89162304-89162304 on Build GRCh38
rs1000112008 Chr.14: 89162283-89162283 on Build GRCh38
rs763609278 Chr.14: 89162463-89162463 on Build GRCh38
rs550889391 Chr.14: 89162322-89162322 on Build GRCh38
rs1000567456 Chr.14: 89162022-89162022 on Build GRCh38
rs779207955 Chr.14: 89162425-89162425 on Build GRCh38
rs577936374 Chr.14: 89162060-89162060 on Build GRCh38
rs1056858152 Chr.14: 89162204-89162204 on Build GRCh38
rs889105394 Chr.14: 89162379-89162379 on Build GRCh38
rs759373581 Chr.14: 89162451-89162451 on Build GRCh38
rs539248302 Chr.14: 89162193-89162193 on Build GRCh38
rs751164977 Chr.14: 89162465-89162465 on Build GRCh38
rs1142538 Chr.14: 89162303-89162303 on Build GRCh38
rs904490892 Chr.14: 89162275-89162275 on Build GRCh38
rs563939958 Chr.14: 89162348-89162348 on Build GRCh38
rs756684594 Chr.14: 89162384-89162384 on Build GRCh38
rs1029439384 Chr.14: 89162299-89162299 on Build GRCh38
rs767808648 Chr.14: 89162020-89162020 on Build GRCh38

View Full Product Details