Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103558483-103558964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG5
Entrez Gene ID
9529
Gene Name
BCL2 associated athanogene 5
Gene Aliases
BAG-5
UniGene
Hs.5443
Gene Chromosome Location
Chr.14: 103556544-103562814 on Build GRCh38
Transcripts
NM_001015048.2 NM_001015049.2 NM_004873.3

SNP Detail

SNP ID Chromosome Location
rs559305464 Chr.14: 103558597-103558597 on Build GRCh38
rs544058555 Chr.14: 103558527-103558527 on Build GRCh38
rs1013124822 Chr.14: 103558710-103558710 on Build GRCh38
rs893722499 Chr.14: 103558722-103558722 on Build GRCh38
rs138438048 Chr.14: 103558592-103558592 on Build GRCh38
rs998799511 Chr.14: 103558699-103558699 on Build GRCh38
rs911576537 Chr.14: 103558538-103558538 on Build GRCh38
rs186854575 Chr.14: 103558902-103558902 on Build GRCh38
rs754488016 Chr.14: 103558697-103558697 on Build GRCh38
rs952280552 Chr.14: 103558833-103558833 on Build GRCh38
rs748817236 Chr.14: 103558728-103558728 on Build GRCh38
rs1055974512 Chr.14: 103558624-103558624 on Build GRCh38
rs962941958 Chr.14: 103558845-103558845 on Build GRCh38
rs950244957 Chr.14: 103558879-103558879 on Build GRCh38
rs924682357 Chr.14: 103558921-103558921 on Build GRCh38
rs756751225 Chr.14: 103558504-103558504 on Build GRCh38
rs756917001 Chr.14: 103558817-103558817 on Build GRCh38
rs934675836 Chr.14: 103558690-103558690 on Build GRCh38
rs145061566 Chr.14: 103558844-103558844 on Build GRCh38
rs930835984 Chr.14: 103558619-103558619 on Build GRCh38
rs142926576 Chr.14: 103558829-103558829 on Build GRCh38
rs940329394 Chr.14: 103558563-103558563 on Build GRCh38
rs961768600 Chr.14: 103558821-103558821 on Build GRCh38
rs894600929 Chr.14: 103558629-103558629 on Build GRCh38
rs533016766 Chr.14: 103558640-103558640 on Build GRCh38
rs781580681 Chr.14: 103558826-103558826 on Build GRCh38
rs916099496 Chr.14: 103558868-103558868 on Build GRCh38
rs77386676 Chr.14: 103558713-103558713 on Build GRCh38
rs1028835743 Chr.14: 103558767-103558767 on Build GRCh38
rs573721371 Chr.14: 103558567-103558567 on Build GRCh38
rs975612303 Chr.14: 103558593-103558593 on Build GRCh38
rs953160976 Chr.14: 103558520-103558520 on Build GRCh38
rs752618806 Chr.14: 103558624-103558625 on Build GRCh38
rs950175102 Chr.14: 103558632-103558632 on Build GRCh38
rs953306847 Chr.14: 103558801-103558801 on Build GRCh38
rs181151039 Chr.14: 103558693-103558693 on Build GRCh38
rs143809081 Chr.14: 103558530-103558530 on Build GRCh38
rs903164262 Chr.14: 103558659-103558659 on Build GRCh38
rs921441398 Chr.14: 103558922-103558922 on Build GRCh38
rs550796774 Chr.14: 103558816-103558816 on Build GRCh38
rs934178630 Chr.14: 103558942-103558942 on Build GRCh38
rs971720048 Chr.14: 103558828-103558828 on Build GRCh38
rs551297675 Chr.14: 103558696-103558696 on Build GRCh38
rs1028740414 Chr.14: 103558517-103558517 on Build GRCh38
rs191224625 Chr.14: 103558604-103558604 on Build GRCh38

View Full Product Details