Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 25678823-25679417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10A
Entrez Gene ID
57194
Gene Name
ATPase phospholipid transporting 10A (putative)
Gene Aliases
ATP10C, ATPVA, ATPVC
UniGene
Hs.638517
Gene Chromosome Location
Chr.15: 25672241-25865144 on Build GRCh38
Transcripts
NM_024490.3 XM_005268261.4 XM_011521826.2 XM_011521828.2 XM_011521829.2 XM_011521830.2 XM_017022436.1 XM_017022437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2156834 c.4482T>A p.S1494S Chr.15: 25679359-25679359 on Build GRCh38
6340075 c.4440A>T p.S1480S Chr.15: 25679401-25679401 on Build GRCh38
5765223 c.4449A>G p.Q1483Q Chr.15: 25679392-25679392 on Build GRCh38
4664127 c.4491G>A p.R1497R Chr.15: 25679350-25679350 on Build GRCh38
7248352 c.4469T>A p.L1490H Chr.15: 25679372-25679372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543922246 Chr.15: 25678853-25678853 on Build GRCh38
rs993520196 Chr.15: 25678865-25678865 on Build GRCh38
rs140854556 Chr.15: 25679366-25679366 on Build GRCh38
rs529960150 Chr.15: 25679111-25679111 on Build GRCh38
rs746568259 Chr.15: 25679328-25679328 on Build GRCh38
rs910443545 Chr.15: 25679101-25679101 on Build GRCh38
rs199722193 Chr.15: 25679351-25679351 on Build GRCh38
rs549742970 Chr.15: 25679234-25679234 on Build GRCh38
rs749763099 Chr.15: 25679349-25679349 on Build GRCh38
rs770460921 Chr.15: 25679329-25679329 on Build GRCh38
rs1007800484 Chr.15: 25679142-25679142 on Build GRCh38
rs188895255 Chr.15: 25679027-25679027 on Build GRCh38
rs564930977 Chr.15: 25678962-25678962 on Build GRCh38
rs758138600 Chr.15: 25679324-25679324 on Build GRCh38
rs1038169662 Chr.15: 25679056-25679056 on Build GRCh38
rs957989640 Chr.15: 25678991-25678991 on Build GRCh38
rs760720206 Chr.15: 25679383-25679383 on Build GRCh38
rs942109865 Chr.15: 25679055-25679055 on Build GRCh38
rs775235581 Chr.15: 25678918-25678918 on Build GRCh38
rs753932918 Chr.15: 25679385-25679385 on Build GRCh38
rs978970513 Chr.15: 25679198-25679198 on Build GRCh38
rs1052114086 Chr.15: 25678966-25678966 on Build GRCh38
rs559502916 Chr.15: 25679136-25679136 on Build GRCh38
rs573928015 Chr.15: 25679011-25679011 on Build GRCh38
rs987764440 Chr.15: 25679288-25679288 on Build GRCh38
rs560655210 Chr.15: 25678916-25678916 on Build GRCh38
rs543040579 Chr.15: 25679108-25679108 on Build GRCh38
rs896109188 Chr.15: 25678891-25678891 on Build GRCh38
rs1032477836 Chr.15: 25679257-25679257 on Build GRCh38
rs751202180 Chr.15: 25679393-25679393 on Build GRCh38
rs929377847 Chr.15: 25679215-25679216 on Build GRCh38
rs920690716 Chr.15: 25679208-25679208 on Build GRCh38
rs769110069 Chr.15: 25679350-25679350 on Build GRCh38
rs755580136 Chr.15: 25679193-25679193 on Build GRCh38
rs759571053 Chr.15: 25679388-25679388 on Build GRCh38
rs560815557 Chr.15: 25679085-25679085 on Build GRCh38
rs540838161 Chr.15: 25679075-25679075 on Build GRCh38
rs752493136 Chr.15: 25679319-25679319 on Build GRCh38
rs933270298 Chr.15: 25679068-25679068 on Build GRCh38
rs145625538 Chr.15: 25678956-25678956 on Build GRCh38
rs763457276 Chr.15: 25679093-25679093 on Build GRCh38
rs890705556 Chr.15: 25679139-25679139 on Build GRCh38
rs897924841 Chr.15: 25679066-25679066 on Build GRCh38
rs140549327 Chr.15: 25678879-25678879 on Build GRCh38
rs917804088 Chr.15: 25679012-25679012 on Build GRCh38
rs754863958 Chr.15: 25679311-25679311 on Build GRCh38
rs376349144 Chr.15: 25678972-25678972 on Build GRCh38
rs778885111 Chr.15: 25679313-25679313 on Build GRCh38
rs956451662 Chr.15: 25679272-25679272 on Build GRCh38
rs753795165 Chr.15: 25679307-25679307 on Build GRCh38
rs777417710 Chr.15: 25679326-25679326 on Build GRCh38
rs549238076 Chr.15: 25679028-25679028 on Build GRCh38
rs963822782 Chr.15: 25678921-25678921 on Build GRCh38
rs1017759453 Chr.15: 25679145-25679145 on Build GRCh38
rs765210750 Chr.15: 25679389-25679389 on Build GRCh38
rs1047700 Chr.15: 25679392-25679392 on Build GRCh38
rs150127969 Chr.15: 25679376-25679376 on Build GRCh38
rs949283410 Chr.15: 25679017-25679017 on Build GRCh38
rs752546336 Chr.15: 25679391-25679391 on Build GRCh38
rs201645433 Chr.15: 25679352-25679352 on Build GRCh38
rs759692595 Chr.15: 25678924-25678924 on Build GRCh38
rs2628634 Chr.15: 25679262-25679262 on Build GRCh38
rs557534584 Chr.15: 25678897-25678897 on Build GRCh38

View Full Product Details