Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21258446-21259014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYM
Entrez Gene ID
1428
Gene Name
crystallin mu
Gene Aliases
DFNA40, THBP
UniGene
Hs.924
Gene Chromosome Location
Chr.16: 21258518-21303136 on Build GRCh38
Transcripts
NM_001888.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1376608 c.924T>G p.D308E Chr.16: 21258802-21258802 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs74595324 Chr.16: 21258668-21258668 on Build GRCh38
rs766111782 Chr.16: 21258887-21258887 on Build GRCh38
rs765085844 Chr.16: 21258797-21258797 on Build GRCh38
rs996800416 Chr.16: 21258992-21258992 on Build GRCh38
rs200789314 Chr.16: 21258859-21258859 on Build GRCh38
rs140779001 Chr.16: 21258783-21258783 on Build GRCh38
rs776123039 Chr.16: 21258769-21258769 on Build GRCh38
rs777654819 Chr.16: 21258824-21258824 on Build GRCh38
rs750046675 Chr.16: 21258877-21258877 on Build GRCh38
rs995345639 Chr.16: 21258562-21258562 on Build GRCh38
rs1011317314 Chr.16: 21258595-21258595 on Build GRCh38
rs759971841 Chr.16: 21258764-21258764 on Build GRCh38
rs34601843 Chr.16: 21258768-21258769 on Build GRCh38
rs104894509 Chr.16: 21258781-21258781 on Build GRCh38
rs370571927 Chr.16: 21258869-21258869 on Build GRCh38
rs902820298 Chr.16: 21258940-21258940 on Build GRCh38
rs750253275 Chr.16: 21258808-21258808 on Build GRCh38
rs1027716208 Chr.16: 21258576-21258576 on Build GRCh38
rs533525035 Chr.16: 21258841-21258841 on Build GRCh38
rs1036963608 Chr.16: 21258543-21258543 on Build GRCh38
rs947120311 Chr.16: 21258899-21258899 on Build GRCh38
rs1029049249 Chr.16: 21258778-21258778 on Build GRCh38
rs956541600 Chr.16: 21258782-21258782 on Build GRCh38
rs551983588 Chr.16: 21258725-21258725 on Build GRCh38
rs554519464 Chr.16: 21258969-21258969 on Build GRCh38
rs749526724 Chr.16: 21258827-21258827 on Build GRCh38
rs773464316 Chr.16: 21258693-21258693 on Build GRCh38
rs758220013 Chr.16: 21258884-21258884 on Build GRCh38
rs104894512 Chr.16: 21258785-21258785 on Build GRCh38
rs201156670 Chr.16: 21258774-21258774 on Build GRCh38
rs751510353 Chr.16: 21258894-21258894 on Build GRCh38
rs774509583 Chr.16: 21258759-21258759 on Build GRCh38
rs776176258 Chr.16: 21258856-21258856 on Build GRCh38
rs753552998 Chr.16: 21258780-21258780 on Build GRCh38
rs931596422 Chr.16: 21258472-21258472 on Build GRCh38
rs749420695 Chr.16: 21258740-21258740 on Build GRCh38
rs749834549 Chr.16: 21258775-21258779 on Build GRCh38
rs771337760 Chr.16: 21258743-21258743 on Build GRCh38
rs76286936 Chr.16: 21258960-21258960 on Build GRCh38
rs374011065 Chr.16: 21258875-21258875 on Build GRCh38
rs775938865 Chr.16: 21258870-21258870 on Build GRCh38
rs897982156 Chr.16: 21258556-21258556 on Build GRCh38
rs727502945 Chr.16: 21258819-21258819 on Build GRCh38
rs1012952557 Chr.16: 21258958-21258958 on Build GRCh38
rs958961447 Chr.16: 21258591-21258591 on Build GRCh38
rs10726 Chr.16: 21258690-21258690 on Build GRCh38
rs531341396 Chr.16: 21258510-21258510 on Build GRCh38
rs756206837 Chr.16: 21258820-21258820 on Build GRCh38
rs984911894 Chr.16: 21258795-21258795 on Build GRCh38
rs35946835 Chr.16: 21258688-21258688 on Build GRCh38
rs142217769 Chr.16: 21258684-21258684 on Build GRCh38
rs551248689 Chr.16: 21258577-21258577 on Build GRCh38
rs994977575 Chr.16: 21258881-21258881 on Build GRCh38
rs1049099211 Chr.16: 21258469-21258469 on Build GRCh38
rs963889665 Chr.16: 21258694-21258694 on Build GRCh38
rs567279229 Chr.16: 21258951-21258951 on Build GRCh38
rs911544434 Chr.16: 21258852-21258852 on Build GRCh38
rs757990110 Chr.16: 21258794-21258795 on Build GRCh38
rs902873224 Chr.16: 21258674-21258674 on Build GRCh38
rs758176818 Chr.16: 21258809-21258809 on Build GRCh38
rs780122021 Chr.16: 21258859-21258859 on Build GRCh38
rs890743819 Chr.16: 21258499-21258499 on Build GRCh38
rs377417888 Chr.16: 21258485-21258485 on Build GRCh38
rs892888016 Chr.16: 21258557-21258557 on Build GRCh38
rs780006425 Chr.16: 21258811-21258811 on Build GRCh38
rs868460924 Chr.16: 21258644-21258644 on Build GRCh38
rs1012836206 Chr.16: 21258600-21258600 on Build GRCh38
rs186774352 Chr.16: 21258501-21258501 on Build GRCh38
rs768192471 Chr.16: 21258765-21258765 on Build GRCh38
rs764942368 Chr.16: 21258971-21258971 on Build GRCh38
rs760305239 Chr.16: 21258770-21258770 on Build GRCh38
rs771122602 Chr.16: 21258828-21258828 on Build GRCh38
rs1041888691 Chr.16: 21258932-21258932 on Build GRCh38
rs772536686 Chr.16: 21258842-21258842 on Build GRCh38
rs774615199 Chr.16: 21258836-21258836 on Build GRCh38

View Full Product Details