Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19564734-19565016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf62
Entrez Gene ID
57020
Gene Name
chromosome 16 open reading frame 62
Gene Aliases
-
UniGene
Hs.654964
Gene Chromosome Location
Chr.16: 19555415-19701163 on Build GRCh38
Transcripts
NM_001300743.1 NM_020314.5 XM_005255435.3 XM_006721065.3 XM_011545896.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6326180 c.288C>T p.H96H Chr.16: 19564854-19564854 on Build GRCh38
4647886 c.323C>T p.S108L Chr.16: 19564889-19564889 on Build GRCh38
4647885 c.56C>T p.S19L Chr.16: 19564889-19564889 on Build GRCh38
5828351 c.299G>A p.R100K Chr.16: 19564865-19564865 on Build GRCh38
1608994 c.381A>G p.I127M Chr.16: 19564947-19564947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778699468 Chr.16: 19564895-19564895 on Build GRCh38
rs373404204 Chr.16: 19564965-19564965 on Build GRCh38
rs749450774 Chr.16: 19564759-19564760 on Build GRCh38
rs775946795 Chr.16: 19564924-19564924 on Build GRCh38
rs751801555 Chr.16: 19564973-19564973 on Build GRCh38
rs767644712 Chr.16: 19564977-19564977 on Build GRCh38
rs752667328 Chr.16: 19564982-19564982 on Build GRCh38
rs150643142 Chr.16: 19564911-19564911 on Build GRCh38
rs750280149 Chr.16: 19564988-19564988 on Build GRCh38
rs746570672 Chr.16: 19564917-19564917 on Build GRCh38
rs765711055 Chr.16: 19564814-19564814 on Build GRCh38
rs542636522 Chr.16: 19564775-19564775 on Build GRCh38
rs369566134 Chr.16: 19564810-19564813 on Build GRCh38
rs770385859 Chr.16: 19564931-19564931 on Build GRCh38
rs777638235 Chr.16: 19564854-19564854 on Build GRCh38
rs58497407 Chr.16: 19564839-19564839 on Build GRCh38
rs1010306107 Chr.16: 19564786-19564786 on Build GRCh38
rs895959804 Chr.16: 19564770-19564770 on Build GRCh38
rs751533830 Chr.16: 19564919-19564919 on Build GRCh38
rs753813652 Chr.16: 19564863-19564863 on Build GRCh38
rs750769446 Chr.16: 19564815-19564815 on Build GRCh38
rs554913587 Chr.16: 19564812-19564812 on Build GRCh38
rs757124401 Chr.16: 19564894-19564894 on Build GRCh38
rs766676140 Chr.16: 19564838-19564838 on Build GRCh38
rs17854969 Chr.16: 19564928-19564928 on Build GRCh38
rs969165849 Chr.16: 19564869-19564869 on Build GRCh38
rs771763824 Chr.16: 19564907-19564907 on Build GRCh38
rs755071858 Chr.16: 19564842-19564842 on Build GRCh38
rs527257312 Chr.16: 19564987-19564987 on Build GRCh38
rs1018659631 Chr.16: 19564918-19564918 on Build GRCh38
rs762433487 Chr.16: 19564926-19564926 on Build GRCh38
rs763385215 Chr.16: 19564960-19564960 on Build GRCh38
rs763300364 Chr.16: 19564821-19564821 on Build GRCh38
rs773753882 Chr.16: 19564945-19564945 on Build GRCh38
rs766566976 Chr.16: 19564966-19564966 on Build GRCh38
rs771212191 Chr.16: 19564860-19564860 on Build GRCh38
rs759610640 Chr.16: 19564974-19564974 on Build GRCh38
rs955803707 Chr.16: 19564935-19564935 on Build GRCh38

View Full Product Details