Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7454195-7454718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB1
Entrez Gene ID
1140
Gene Name
cholinergic receptor nicotinic beta 1 subunit
Gene Aliases
ACHRB, CHRNB, CMS1D, CMS2A, CMS2C, SCCMS
UniGene
Hs.330386
Gene Chromosome Location
Chr.17: 7445087-7457613 on Build GRCh38
Transcripts
NM_000747.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628758 c.991A>G p.N331D Chr.17: 7454467-7454467 on Build GRCh38
4510796 c.849C>T p.A283A Chr.17: 7454325-7454325 on Build GRCh38
7274347 c.956T>C p.V319A Chr.17: 7454432-7454432 on Build GRCh38
1721706 c.871_873delCTG p.L294delL Chr.17: 7454347-7454349 on Build GRCh38
4672280 c.840A>G p.S280S Chr.17: 7454316-7454316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750396330 Chr.17: 7454439-7454439 on Build GRCh38
rs771545907 Chr.17: 7454409-7454409 on Build GRCh38
rs140527233 Chr.17: 7454621-7454621 on Build GRCh38
rs886041490 Chr.17: 7454395-7454395 on Build GRCh38
rs372471189 Chr.17: 7454567-7454567 on Build GRCh38
rs1024392634 Chr.17: 7454541-7454541 on Build GRCh38
rs757194127 Chr.17: 7454317-7454317 on Build GRCh38
rs778746961 Chr.17: 7454318-7454318 on Build GRCh38
rs369677810 Chr.17: 7454481-7454481 on Build GRCh38
rs151303014 Chr.17: 7454540-7454540 on Build GRCh38
rs1052747193 Chr.17: 7454583-7454583 on Build GRCh38
rs761535567 Chr.17: 7454435-7454435 on Build GRCh38
rs779086505 Chr.17: 7454563-7454563 on Build GRCh38
rs372910299 Chr.17: 7454390-7454390 on Build GRCh38
rs1021959339 Chr.17: 7454607-7454607 on Build GRCh38
rs762732871 Chr.17: 7454534-7454534 on Build GRCh38
rs775264960 Chr.17: 7454412-7454412 on Build GRCh38
rs781190969 Chr.17: 7454448-7454448 on Build GRCh38
rs922345288 Chr.17: 7454684-7454684 on Build GRCh38
rs189153402 Chr.17: 7454603-7454603 on Build GRCh38
rs945086307 Chr.17: 7454374-7454374 on Build GRCh38
rs137852810 Chr.17: 7454341-7454341 on Build GRCh38
rs956543933 Chr.17: 7454353-7454353 on Build GRCh38
rs201913823 Chr.17: 7454515-7454515 on Build GRCh38
rs752778673 Chr.17: 7454455-7454455 on Build GRCh38
rs140169734 Chr.17: 7454377-7454377 on Build GRCh38
rs143871421 Chr.17: 7454529-7454529 on Build GRCh38
rs775070102 Chr.17: 7454491-7454491 on Build GRCh38
rs758930059 Chr.17: 7454568-7454568 on Build GRCh38
rs375505960 Chr.17: 7454414-7454414 on Build GRCh38
rs200491626 Chr.17: 7454430-7454430 on Build GRCh38
rs759057934 Chr.17: 7454400-7454400 on Build GRCh38
rs969121499 Chr.17: 7454424-7454424 on Build GRCh38
rs112895222 Chr.17: 7454277-7454277 on Build GRCh38
rs756239699 Chr.17: 7454547-7454547 on Build GRCh38
rs567864992 Chr.17: 7454266-7454266 on Build GRCh38
rs534048413 Chr.17: 7454668-7454668 on Build GRCh38
rs180908813 Chr.17: 7454255-7454255 on Build GRCh38
rs768374343 Chr.17: 7454514-7454514 on Build GRCh38
rs76001008 Chr.17: 7454288-7454288 on Build GRCh38
rs966401975 Chr.17: 7454643-7454643 on Build GRCh38
rs746417364 Chr.17: 7454480-7454480 on Build GRCh38
rs4796418 Chr.17: 7454681-7454681 on Build GRCh38
rs148217194 Chr.17: 7454383-7454383 on Build GRCh38
rs868298067 Chr.17: 7454524-7454524 on Build GRCh38
rs773696133 Chr.17: 7454256-7454256 on Build GRCh38
rs748428224 Chr.17: 7454371-7454371 on Build GRCh38
rs763393615 Chr.17: 7454258-7454258 on Build GRCh38
rs781475180 Chr.17: 7454364-7454364 on Build GRCh38
rs753695361 Chr.17: 7454316-7454316 on Build GRCh38
rs756960602 Chr.17: 7454355-7454358 on Build GRCh38
rs373928636 Chr.17: 7454246-7454246 on Build GRCh38
rs137852811 Chr.17: 7454329-7454329 on Build GRCh38
rs777942954 Chr.17: 7454549-7454549 on Build GRCh38
rs755060667 Chr.17: 7454350-7454350 on Build GRCh38
rs769813899 Chr.17: 7454273-7454273 on Build GRCh38
rs188543727 Chr.17: 7454238-7454238 on Build GRCh38
rs764035317 Chr.17: 7454293-7454293 on Build GRCh38
rs760292772 Chr.17: 7454503-7454503 on Build GRCh38
rs755073927 Chr.17: 7454445-7454445 on Build GRCh38
rs77319126 Chr.17: 7454544-7454544 on Build GRCh38
rs371190837 Chr.17: 7454281-7454281 on Build GRCh38
rs758993114 Chr.17: 7454271-7454271 on Build GRCh38
rs368084123 Chr.17: 7454623-7454623 on Build GRCh38
rs536204964 Chr.17: 7454532-7454534 on Build GRCh38
rs750248584 Chr.17: 7454532-7454532 on Build GRCh38
rs865958957 Chr.17: 7454458-7454458 on Build GRCh38
rs998494409 Chr.17: 7454319-7454320 on Build GRCh38
rs117168441 Chr.17: 7454379-7454379 on Build GRCh38
rs371142002 Chr.17: 7454516-7454516 on Build GRCh38
rs752775617 Chr.17: 7454615-7454615 on Build GRCh38
rs961734949 Chr.17: 7454230-7454230 on Build GRCh38
rs780391043 Chr.17: 7454358-7454358 on Build GRCh38
rs927431484 Chr.17: 7454469-7454469 on Build GRCh38
rs753428978 Chr.17: 7454329-7454334 on Build GRCh38
rs201129045 Chr.17: 7454323-7454323 on Build GRCh38
rs759680486 Chr.17: 7454591-7454591 on Build GRCh38
rs989527059 Chr.17: 7454326-7454326 on Build GRCh38
rs756292892 Chr.17: 7454468-7454468 on Build GRCh38
rs578007406 Chr.17: 7454555-7454555 on Build GRCh38
rs770017785 Chr.17: 7454375-7454375 on Build GRCh38
rs779467380 Chr.17: 7454479-7454479 on Build GRCh38
rs763839239 Chr.17: 7454423-7454423 on Build GRCh38
rs749634279 Chr.17: 7454471-7454471 on Build GRCh38
rs1024444772 Chr.17: 7454252-7454252 on Build GRCh38
rs753646298 Chr.17: 7454431-7454431 on Build GRCh38
rs7210231 Chr.17: 7454545-7454545 on Build GRCh38
rs774773039 Chr.17: 7454385-7454385 on Build GRCh38
rs145826933 Chr.17: 7454328-7454328 on Build GRCh38
rs771242993 Chr.17: 7454477-7454477 on Build GRCh38
rs780728641 Chr.17: 7454572-7454572 on Build GRCh38
rs202089107 Chr.17: 7454262-7454262 on Build GRCh38

View Full Product Details