Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 35772095-35772661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK1
Entrez Gene ID
9201
Gene Name
doublecortin like kinase 1
Gene Aliases
CL1, CLICK1, DCAMKL1, DCDC3A, DCLK
UniGene
Hs.507755
Gene Chromosome Location
Chr.13: 35768652-36132020 on Build GRCh38
Transcripts
NM_001195415.1 NM_001195416.1 NM_004734.4 XM_005266592.2 XM_006719893.2 XM_017020847.1

SNP Detail

SNP ID Chromosome Location
rs551510870 Chr.13: 35772435-35772435 on Build GRCh38
rs567074283 Chr.13: 35772539-35772539 on Build GRCh38
rs912393016 Chr.13: 35772496-35772496 on Build GRCh38
rs543029016 Chr.13: 35772508-35772508 on Build GRCh38
rs992895168 Chr.13: 35772307-35772307 on Build GRCh38
rs940416174 Chr.13: 35772388-35772388 on Build GRCh38
rs529516593 Chr.13: 35772337-35772337 on Build GRCh38
rs1001750785 Chr.13: 35772329-35772329 on Build GRCh38
rs552812226 Chr.13: 35772578-35772578 on Build GRCh38
rs953947893 Chr.13: 35772492-35772492 on Build GRCh38
rs968988869 Chr.13: 35772278-35772278 on Build GRCh38
rs1029415104 Chr.13: 35772467-35772467 on Build GRCh38
rs563375916 Chr.13: 35772191-35772191 on Build GRCh38
rs1010261817 Chr.13: 35772401-35772401 on Build GRCh38
rs752508333 Chr.13: 35772483-35772483 on Build GRCh38
rs965236765 Chr.13: 35772516-35772516 on Build GRCh38
rs570116666 Chr.13: 35772440-35772440 on Build GRCh38
rs762870219 Chr.13: 35772592-35772592 on Build GRCh38
rs560813804 Chr.13: 35772179-35772179 on Build GRCh38
rs918235192 Chr.13: 35772274-35772274 on Build GRCh38
rs956313278 Chr.13: 35772185-35772185 on Build GRCh38
rs57621661 Chr.13: 35772312-35772312 on Build GRCh38
rs548764679 Chr.13: 35772512-35772512 on Build GRCh38
rs904571449 Chr.13: 35772639-35772639 on Build GRCh38
rs960565694 Chr.13: 35772636-35772636 on Build GRCh38
rs1008604177 Chr.13: 35772140-35772140 on Build GRCh38
rs890335077 Chr.13: 35772507-35772507 on Build GRCh38
rs77735904 Chr.13: 35772553-35772553 on Build GRCh38
rs9943927 Chr.13: 35772414-35772414 on Build GRCh38
rs1021689902 Chr.13: 35772543-35772543 on Build GRCh38
rs1010725333 Chr.13: 35772542-35772542 on Build GRCh38
rs185371565 Chr.13: 35772589-35772589 on Build GRCh38
rs760904440 Chr.13: 35772442-35772442 on Build GRCh38
rs544022242 Chr.13: 35772262-35772262 on Build GRCh38
rs371071845 Chr.13: 35772227-35772227 on Build GRCh38
rs34230126 Chr.13: 35772289-35772290 on Build GRCh38
rs895968057 Chr.13: 35772586-35772587 on Build GRCh38
rs775514054 Chr.13: 35772265-35772265 on Build GRCh38
rs1057192125 Chr.13: 35772332-35772332 on Build GRCh38
rs896020285 Chr.13: 35772369-35772369 on Build GRCh38
rs764993164 Chr.13: 35772476-35772476 on Build GRCh38
rs968774661 Chr.13: 35772179-35772179 on Build GRCh38
rs1020232369 Chr.13: 35772158-35772158 on Build GRCh38
rs943766345 Chr.13: 35772245-35772245 on Build GRCh38
rs1019706933 Chr.13: 35772160-35772160 on Build GRCh38
rs937595201 Chr.13: 35772577-35772577 on Build GRCh38
rs927507580 Chr.13: 35772289-35772289 on Build GRCh38
rs181178100 Chr.13: 35772284-35772284 on Build GRCh38
rs931071199 Chr.13: 35772547-35772547 on Build GRCh38
rs946193658 Chr.13: 35772600-35772600 on Build GRCh38
rs548071106 Chr.13: 35772413-35772413 on Build GRCh38
rs918321029 Chr.13: 35772546-35772546 on Build GRCh38
rs150326507 Chr.13: 35772124-35772124 on Build GRCh38
rs57984726 Chr.13: 35772293-35772294 on Build GRCh38
rs985413321 Chr.13: 35772496-35772496 on Build GRCh38
rs145881215 Chr.13: 35772443-35772443 on Build GRCh38
rs954111381 Chr.13: 35772216-35772216 on Build GRCh38
rs960082382 Chr.13: 35772119-35772119 on Build GRCh38
rs1021784792 Chr.13: 35772260-35772260 on Build GRCh38
rs140690595 Chr.13: 35772415-35772415 on Build GRCh38
rs1008264830 Chr.13: 35772527-35772527 on Build GRCh38
rs764196871 Chr.13: 35772290-35772290 on Build GRCh38
rs1020032961 Chr.13: 35772538-35772538 on Build GRCh38
rs926957467 Chr.13: 35772571-35772571 on Build GRCh38

View Full Product Details