Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43068848-43069158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC15
Entrez Gene ID
29103
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1, HSD18, MCJ
UniGene
Hs.438830
Gene Chromosome Location
Chr.13: 43023226-43109170 on Build GRCh38
Transcripts
NM_013238.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
416357 c.177G>A p.R59R Chr.13: 43068946-43068946 on Build GRCh38
4047708 c.175C>T p.R59W Chr.13: 43068944-43068944 on Build GRCh38
1941204 c.176G>A p.R59Q Chr.13: 43068945-43068945 on Build GRCh38
4047710 c.206C>G p.T69R Chr.13: 43068975-43068975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935511679 Chr.13: 43069124-43069125 on Build GRCh38
rs977658385 Chr.13: 43068920-43068920 on Build GRCh38
rs780341477 Chr.13: 43068900-43068900 on Build GRCh38
rs765899911 Chr.13: 43068932-43068932 on Build GRCh38
rs563092086 Chr.13: 43068873-43068873 on Build GRCh38
rs192280089 Chr.13: 43069079-43069079 on Build GRCh38
rs1021720571 Chr.13: 43069067-43069067 on Build GRCh38
rs61733777 Chr.13: 43068990-43068990 on Build GRCh38
rs1052618639 Chr.13: 43068893-43068893 on Build GRCh38
rs774780407 Chr.13: 43068923-43068923 on Build GRCh38
rs768781912 Chr.13: 43068903-43068903 on Build GRCh38
rs752317033 Chr.13: 43068944-43068944 on Build GRCh38
rs148028737 Chr.13: 43068937-43068937 on Build GRCh38
rs188210172 Chr.13: 43068875-43068875 on Build GRCh38
rs763438611 Chr.13: 43069054-43069055 on Build GRCh38
rs370540312 Chr.13: 43068924-43068924 on Build GRCh38
rs756941914 Chr.13: 43069035-43069035 on Build GRCh38
rs141721055 Chr.13: 43068966-43068966 on Build GRCh38
rs368420201 Chr.13: 43068975-43068975 on Build GRCh38
rs777426623 Chr.13: 43068894-43068895 on Build GRCh38
rs201392334 Chr.13: 43068983-43068983 on Build GRCh38
rs762570627 Chr.13: 43069019-43069019 on Build GRCh38
rs758408975 Chr.13: 43068945-43068945 on Build GRCh38
rs116641501 Chr.13: 43068952-43068952 on Build GRCh38
rs999023580 Chr.13: 43069084-43069084 on Build GRCh38
rs771333561 Chr.13: 43068994-43068994 on Build GRCh38
rs542678080 Chr.13: 43068930-43068930 on Build GRCh38
rs142686061 Chr.13: 43069023-43069023 on Build GRCh38
rs768717251 Chr.13: 43068933-43068933 on Build GRCh38
rs564710643 Chr.13: 43069015-43069015 on Build GRCh38
rs550268649 Chr.13: 43069051-43069051 on Build GRCh38
rs757102808 Chr.13: 43068956-43068956 on Build GRCh38
rs776092175 Chr.13: 43069011-43069011 on Build GRCh38
rs773667685 Chr.13: 43069052-43069053 on Build GRCh38
rs1021787410 Chr.13: 43068906-43068906 on Build GRCh38
rs768272650 Chr.13: 43068934-43068934 on Build GRCh38
rs747739713 Chr.13: 43068999-43069001 on Build GRCh38
rs755248581 Chr.13: 43068968-43068968 on Build GRCh38
rs774495616 Chr.13: 43068986-43068988 on Build GRCh38
rs759215985 Chr.13: 43069014-43069014 on Build GRCh38
rs1052181867 Chr.13: 43069129-43069129 on Build GRCh38
rs748266523 Chr.13: 43068982-43068982 on Build GRCh38
rs968930390 Chr.13: 43068914-43068914 on Build GRCh38
rs749284979 Chr.13: 43068902-43068902 on Build GRCh38
rs780071020 Chr.13: 43068961-43068961 on Build GRCh38
rs769588361 Chr.13: 43069010-43069011 on Build GRCh38
rs1010641382 Chr.13: 43069053-43069053 on Build GRCh38
rs758906339 Chr.13: 43068936-43068936 on Build GRCh38

View Full Product Details