Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14594359-14594865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC17A
Entrez Gene ID
388512
Gene Name
C-type lectin domain family 17 member A
Gene Aliases
-
UniGene
Hs.567933
Gene Chromosome Location
Chr.19: 14580777-14612035 on Build GRCh38
Transcripts
NM_001204118.1 NM_207390.3 NR_109784.1 NR_109785.1 XM_017026785.1 XM_017026786.1 XM_017026787.1 XM_017026788.1 XM_017026789.1 XM_017026790.1 XM_017026791.1 XM_017026792.1 XM_017026793.1 XM_017026794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630147 c.373G>A p.A125T Chr.19: 14594770-14594770 on Build GRCh38
5224975 c.290T>C p.L97P Chr.19: 14594499-14594499 on Build GRCh38
6443127 c.345_346GG>AA p.E116K Chr.19: 14594636-14594637 on Build GRCh38
5043015 c.311-1G>T p.? Chr.19: 14594631-14594631 on Build GRCh38
5195493 c.288C>T p.T96T Chr.19: 14594497-14594497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367815850 Chr.19: 14594773-14594773 on Build GRCh38
rs371430849 Chr.19: 14594543-14594543 on Build GRCh38
rs757111898 Chr.19: 14594741-14594741 on Build GRCh38
rs770888771 Chr.19: 14594555-14594555 on Build GRCh38
rs756279350 Chr.19: 14594677-14594677 on Build GRCh38
rs778637159 Chr.19: 14594507-14594507 on Build GRCh38
rs544779902 Chr.19: 14594837-14594837 on Build GRCh38
rs376222790 Chr.19: 14594667-14594667 on Build GRCh38
rs894402646 Chr.19: 14594760-14594760 on Build GRCh38
rs777280967 Chr.19: 14594393-14594393 on Build GRCh38
rs956147066 Chr.19: 14594597-14594597 on Build GRCh38
rs1055714844 Chr.19: 14594436-14594436 on Build GRCh38
rs767290093 Chr.19: 14594752-14594752 on Build GRCh38
rs755048797 Chr.19: 14594770-14594770 on Build GRCh38
rs773427024 Chr.19: 14594781-14594781 on Build GRCh38
rs138054531 Chr.19: 14594671-14594671 on Build GRCh38
rs752452950 Chr.19: 14594649-14594649 on Build GRCh38
rs375465288 Chr.19: 14594765-14594765 on Build GRCh38
rs762408076 Chr.19: 14594722-14594722 on Build GRCh38
rs537474682 Chr.19: 14594791-14594791 on Build GRCh38
rs370861432 Chr.19: 14594695-14594695 on Build GRCh38
rs114453977 Chr.19: 14594504-14594504 on Build GRCh38
rs775852314 Chr.19: 14594571-14594571 on Build GRCh38
rs1005503011 Chr.19: 14594424-14594424 on Build GRCh38
rs372350704 Chr.19: 14594475-14594475 on Build GRCh38
rs765200414 Chr.19: 14594734-14594734 on Build GRCh38
rs78269670 Chr.19: 14594762-14594762 on Build GRCh38
rs370387834 Chr.19: 14594829-14594829 on Build GRCh38
rs780005106 Chr.19: 14594769-14594769 on Build GRCh38
rs751146435 Chr.19: 14594726-14594726 on Build GRCh38
rs762390484 Chr.19: 14594484-14594484 on Build GRCh38
rs888097127 Chr.19: 14594403-14594403 on Build GRCh38
rs369929625 Chr.19: 14594564-14594564 on Build GRCh38
rs762457015 Chr.19: 14594815-14594815 on Build GRCh38
rs369115211 Chr.19: 14594670-14594670 on Build GRCh38
rs1024528992 Chr.19: 14594469-14594469 on Build GRCh38
rs764365528 Chr.19: 14594468-14594468 on Build GRCh38
rs772622288 Chr.19: 14594776-14594776 on Build GRCh38
rs763915824 Chr.19: 14594662-14594662 on Build GRCh38
rs367981721 Chr.19: 14594404-14594404 on Build GRCh38
rs779979349 Chr.19: 14594679-14594679 on Build GRCh38
rs562083233 Chr.19: 14594612-14594612 on Build GRCh38
rs752221536 Chr.19: 14594516-14594516 on Build GRCh38
rs894342403 Chr.19: 14594454-14594454 on Build GRCh38
rs753426690 Chr.19: 14594494-14594494 on Build GRCh38
rs747589482 Chr.19: 14594782-14594782 on Build GRCh38
rs35049360 Chr.19: 14594697-14594697 on Build GRCh38
rs34335847 Chr.19: 14594687-14594687 on Build GRCh38
rs376153824 Chr.19: 14594699-14594699 on Build GRCh38
rs199917925 Chr.19: 14594551-14594551 on Build GRCh38
rs774830203 Chr.19: 14594714-14594714 on Build GRCh38
rs774828809 Chr.19: 14594595-14594595 on Build GRCh38
rs1050436410 Chr.19: 14594565-14594565 on Build GRCh38
rs540582324 Chr.19: 14594609-14594609 on Build GRCh38
rs373832997 Chr.19: 14594569-14594569 on Build GRCh38
rs764851867 Chr.19: 14594713-14594713 on Build GRCh38
rs761073515 Chr.19: 14594702-14594702 on Build GRCh38
rs777139189 Chr.19: 14594797-14594797 on Build GRCh38
rs748533514 Chr.19: 14594696-14594696 on Build GRCh38
rs779277712 Chr.19: 14594772-14594772 on Build GRCh38
rs557103156 Chr.19: 14594394-14594394 on Build GRCh38
rs762225243 Chr.19: 14594610-14594610 on Build GRCh38
rs903283652 Chr.19: 14594489-14594489 on Build GRCh38
rs757963370 Chr.19: 14594653-14594653 on Build GRCh38
rs763381765 Chr.19: 14594458-14594458 on Build GRCh38
rs761436170 Chr.19: 14594830-14594830 on Build GRCh38
rs533142602 Chr.19: 14594694-14594694 on Build GRCh38
rs539835683 Chr.19: 14594725-14594725 on Build GRCh38
rs781061314 Chr.19: 14594561-14594561 on Build GRCh38
rs1033001890 Chr.19: 14594511-14594511 on Build GRCh38
rs764647865 Chr.19: 14594621-14594621 on Build GRCh38
rs904736450 Chr.19: 14594566-14594566 on Build GRCh38
rs759406532 Chr.19: 14594649-14594649 on Build GRCh38
rs759088253 Chr.19: 14594615-14594615 on Build GRCh38
rs776690695 Chr.19: 14594446-14594446 on Build GRCh38
rs766030282 Chr.19: 14594490-14594490 on Build GRCh38
rs777531272 Chr.19: 14594549-14594549 on Build GRCh38

View Full Product Details