Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35778177-35778721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP33
Entrez Gene ID
115703
Gene Name
Rho GTPase activating protein 33
Gene Aliases
NOMA-GAP, SNX26, TCGAP
UniGene
Hs.515364
Gene Chromosome Location
Chr.19: 35775515-35788822 on Build GRCh38
Transcripts
NM_001172630.1 NM_052948.3 XM_005258488.1 XM_006722999.1 XM_011526416.1 XM_011526417.1 XM_011526418.1 XM_011526419.1 XM_011526420.1 XM_011526421.2 XM_017026238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026192 c.429T>A p.L143L Chr.19: 35778568-35778568 on Build GRCh38
1680899 c.338C>T p.P113L Chr.19: 35778477-35778477 on Build GRCh38
6479064 c.286G>T p.E96* Chr.19: 35778322-35778322 on Build GRCh38
2154502 c.296T>G p.L99R Chr.19: 35778332-35778332 on Build GRCh38
295871 c.264C>T p.T88T Chr.19: 35778354-35778354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776829480 Chr.19: 35778339-35778339 on Build GRCh38
rs189367785 Chr.19: 35778205-35778205 on Build GRCh38
rs747378107 Chr.19: 35778415-35778415 on Build GRCh38
rs775801416 Chr.19: 35778343-35778343 on Build GRCh38
rs138067561 Chr.19: 35778261-35778261 on Build GRCh38
rs764837225 Chr.19: 35778478-35778478 on Build GRCh38
rs750869355 Chr.19: 35778570-35778570 on Build GRCh38
rs777517321 Chr.19: 35778661-35778661 on Build GRCh38
rs1038681371 Chr.19: 35778390-35778390 on Build GRCh38
rs748317877 Chr.19: 35778500-35778501 on Build GRCh38
rs747883670 Chr.19: 35778533-35778533 on Build GRCh38
rs763947024 Chr.19: 35778355-35778355 on Build GRCh38
rs926824343 Chr.19: 35778582-35778589 on Build GRCh38
rs903876848 Chr.19: 35778489-35778489 on Build GRCh38
rs753343608 Chr.19: 35778467-35778467 on Build GRCh38
rs915914207 Chr.19: 35778266-35778266 on Build GRCh38
rs765905938 Chr.19: 35778460-35778460 on Build GRCh38
rs201533373 Chr.19: 35778506-35778506 on Build GRCh38
rs762589982 Chr.19: 35778352-35778352 on Build GRCh38
rs770099536 Chr.19: 35778341-35778341 on Build GRCh38
rs35902249 Chr.19: 35778573-35778573 on Build GRCh38
rs748362553 Chr.19: 35778641-35778641 on Build GRCh38
rs76260948 Chr.19: 35778700-35778700 on Build GRCh38
rs747177293 Chr.19: 35778623-35778623 on Build GRCh38
rs764894828 Chr.19: 35778559-35778559 on Build GRCh38
rs914077313 Chr.19: 35778693-35778693 on Build GRCh38
rs112865072 Chr.19: 35778511-35778511 on Build GRCh38
rs771620059 Chr.19: 35778334-35778334 on Build GRCh38
rs768808973 Chr.19: 35778438-35778438 on Build GRCh38
rs375128182 Chr.19: 35778368-35778368 on Build GRCh38
rs567996125 Chr.19: 35778674-35778674 on Build GRCh38
rs146045855 Chr.19: 35778544-35778544 on Build GRCh38
rs376004154 Chr.19: 35778386-35778386 on Build GRCh38
rs199996590 Chr.19: 35778373-35778373 on Build GRCh38
rs755736461 Chr.19: 35778309-35778309 on Build GRCh38
rs765960551 Chr.19: 35778378-35778378 on Build GRCh38
rs754633637 Chr.19: 35778468-35778468 on Build GRCh38
rs531034753 Chr.19: 35778410-35778410 on Build GRCh38
rs146328719 Chr.19: 35778340-35778340 on Build GRCh38
rs541372672 Chr.19: 35778258-35778258 on Build GRCh38
rs751370190 Chr.19: 35778359-35778359 on Build GRCh38
rs745891592 Chr.19: 35778426-35778426 on Build GRCh38
rs530975098 Chr.19: 35778216-35778216 on Build GRCh38
rs781740762 Chr.19: 35778494-35778494 on Build GRCh38
rs747542545 Chr.19: 35778329-35778329 on Build GRCh38
rs1045367847 Chr.19: 35778367-35778367 on Build GRCh38
rs773095605 Chr.19: 35778602-35778603 on Build GRCh38
rs142070691 Chr.19: 35778327-35778327 on Build GRCh38
rs749514446 Chr.19: 35778433-35778433 on Build GRCh38
rs556912669 Chr.19: 35778561-35778561 on Build GRCh38
rs375297737 Chr.19: 35778540-35778540 on Build GRCh38
rs772496264 Chr.19: 35778326-35778326 on Build GRCh38
rs180862527 Chr.19: 35778214-35778214 on Build GRCh38
rs369008398 Chr.19: 35778412-35778412 on Build GRCh38
rs746094199 Chr.19: 35778581-35778581 on Build GRCh38
rs759027074 Chr.19: 35778677-35778677 on Build GRCh38
rs967965417 Chr.19: 35778255-35778255 on Build GRCh38
rs372223573 Chr.19: 35778555-35778555 on Build GRCh38
rs777911177 Chr.19: 35778542-35778542 on Build GRCh38
rs142645416 Chr.19: 35778541-35778541 on Build GRCh38
rs533445422 Chr.19: 35778668-35778668 on Build GRCh38
rs779715955 Chr.19: 35778312-35778312 on Build GRCh38
rs751521287 Chr.19: 35778295-35778295 on Build GRCh38
rs774164767 Chr.19: 35778443-35778443 on Build GRCh38
rs757807292 Chr.19: 35778418-35778418 on Build GRCh38
rs770886534 Chr.19: 35778562-35778562 on Build GRCh38
rs770006381 Chr.19: 35778430-35778430 on Build GRCh38
rs1030557491 Chr.19: 35778202-35778202 on Build GRCh38
rs771861946 Chr.19: 35778539-35778539 on Build GRCh38
rs754651050 Chr.19: 35778408-35778408 on Build GRCh38
rs939593144 Chr.19: 35778664-35778664 on Build GRCh38
rs966583560 Chr.19: 35778204-35778204 on Build GRCh38
rs754208248 Chr.19: 35778698-35778698 on Build GRCh38
rs570481273 Chr.19: 35778432-35778432 on Build GRCh38
rs753909509 Chr.19: 35778580-35778580 on Build GRCh38
rs534667335 Chr.19: 35778450-35778450 on Build GRCh38
rs900187203 Chr.19: 35778401-35778401 on Build GRCh38
rs997815864 Chr.19: 35778434-35778434 on Build GRCh38
rs201365669 Chr.19: 35778298-35778298 on Build GRCh38
rs760108110 Chr.19: 35778451-35778451 on Build GRCh38
rs780457520 Chr.19: 35778574-35778574 on Build GRCh38
rs780668839 Chr.19: 35778299-35778299 on Build GRCh38
rs762674251 Chr.19: 35778701-35778701 on Build GRCh38
rs918973373 Chr.19: 35778307-35778307 on Build GRCh38
rs371607047 Chr.19: 35778374-35778374 on Build GRCh38
rs371671563 Chr.19: 35778232-35778232 on Build GRCh38
rs374221923 Chr.19: 35778503-35778503 on Build GRCh38
rs769190833 Chr.19: 35778679-35778679 on Build GRCh38
rs772293323 Chr.19: 35778574-35778575 on Build GRCh38
rs748414370 Chr.19: 35778317-35778317 on Build GRCh38
rs370786881 Chr.19: 35778516-35778516 on Build GRCh38
rs202132781 Chr.19: 35778528-35778528 on Build GRCh38
rs138284683 Chr.19: 35778493-35778493 on Build GRCh38
rs761630819 Chr.19: 35778444-35778444 on Build GRCh38
rs767981943 Chr.19: 35778569-35778569 on Build GRCh38
rs528663110 Chr.19: 35778675-35778675 on Build GRCh38
rs780159840 Chr.19: 35778505-35778505 on Build GRCh38
rs781772311 Chr.19: 35778419-35778419 on Build GRCh38
rs369373966 Chr.19: 35778372-35778372 on Build GRCh38
rs746374775 Chr.19: 35778495-35778495 on Build GRCh38
rs779043047 Chr.19: 35778606-35778606 on Build GRCh38
rs148799554 Chr.19: 35778547-35778547 on Build GRCh38
rs774463935 Chr.19: 35778529-35778529 on Build GRCh38
rs749985450 Chr.19: 35778300-35778300 on Build GRCh38

View Full Product Details