Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32597980-32598499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD27
Entrez Gene ID
84079
Gene Name
ankyrin repeat domain 27
Gene Aliases
PP12899, VARP
UniGene
Hs.59236
Gene Chromosome Location
Chr.19: 32597001-32675215 on Build GRCh38
Transcripts
NM_032139.2 XM_006723412.2 XM_017027349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6890808 c.3094G>A p.G1032S Chr.19: 32598204-32598204 on Build GRCh38
6600572 c.3058C>T p.R1020W Chr.19: 32598240-32598240 on Build GRCh38
3532178 c.3098C>T p.P1033L Chr.19: 32598200-32598200 on Build GRCh38
6297163 c.2955C>A p.N985K Chr.19: 32598343-32598343 on Build GRCh38
6600571 c.3080C>T p.A1027V Chr.19: 32598218-32598218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149558100 Chr.19: 32598252-32598252 on Build GRCh38
rs375258009 Chr.19: 32598297-32598297 on Build GRCh38
rs150349860 Chr.19: 32598200-32598200 on Build GRCh38
rs923896719 Chr.19: 32598198-32598198 on Build GRCh38
rs767625710 Chr.19: 32598154-32598154 on Build GRCh38
rs575861049 Chr.19: 32598365-32598365 on Build GRCh38
rs748771025 Chr.19: 32598228-32598228 on Build GRCh38
rs773156520 Chr.19: 32598305-32598305 on Build GRCh38
rs780541563 Chr.19: 32598283-32598283 on Build GRCh38
rs761812840 Chr.19: 32598413-32598413 on Build GRCh38
rs758564943 Chr.19: 32598176-32598176 on Build GRCh38
rs138242852 Chr.19: 32598249-32598249 on Build GRCh38
rs371160650 Chr.19: 32598324-32598324 on Build GRCh38
rs771175856 Chr.19: 32598062-32598062 on Build GRCh38
rs761632048 Chr.19: 32598153-32598153 on Build GRCh38
rs766294847 Chr.19: 32598315-32598315 on Build GRCh38
rs761754846 Chr.19: 32598239-32598239 on Build GRCh38
rs769729680 Chr.19: 32598381-32598386 on Build GRCh38
rs146818631 Chr.19: 32598213-32598213 on Build GRCh38
rs748814631 Chr.19: 32598285-32598285 on Build GRCh38
rs890122360 Chr.19: 32598439-32598439 on Build GRCh38
rs376452284 Chr.19: 32598214-32598214 on Build GRCh38
rs1001082750 Chr.19: 32598367-32598367 on Build GRCh38
rs774883566 Chr.19: 32598219-32598219 on Build GRCh38
rs928911418 Chr.19: 32598090-32598090 on Build GRCh38
rs781414796 Chr.19: 32598284-32598285 on Build GRCh38
rs529414162 Chr.19: 32598152-32598152 on Build GRCh38
rs759548763 Chr.19: 32598407-32598407 on Build GRCh38
rs776473820 Chr.19: 32598405-32598405 on Build GRCh38
rs141702275 Chr.19: 32598395-32598395 on Build GRCh38
rs958127664 Chr.19: 32598256-32598256 on Build GRCh38
rs770124619 Chr.19: 32598386-32598386 on Build GRCh38
rs764129016 Chr.19: 32598327-32598327 on Build GRCh38
rs948759892 Chr.19: 32598087-32598087 on Build GRCh38
rs780479884 Chr.19: 32598375-32598375 on Build GRCh38
rs746267119 Chr.19: 32598378-32598378 on Build GRCh38
rs745908469 Chr.19: 32598355-32598356 on Build GRCh38
rs781567816 Chr.19: 32598272-32598272 on Build GRCh38
rs1035995730 Chr.19: 32598282-32598282 on Build GRCh38
rs747837816 Chr.19: 32598298-32598298 on Build GRCh38
rs17754651 Chr.19: 32598082-32598082 on Build GRCh38
rs768195635 Chr.19: 32598229-32598229 on Build GRCh38
rs368439993 Chr.19: 32598312-32598312 on Build GRCh38
rs777906137 Chr.19: 32598199-32598199 on Build GRCh38
rs538870613 Chr.19: 32598280-32598280 on Build GRCh38
rs145648811 Chr.19: 32598248-32598248 on Build GRCh38
rs370994601 Chr.19: 32598260-32598260 on Build GRCh38
rs758502126 Chr.19: 32598253-32598253 on Build GRCh38
rs767392918 Chr.19: 32598240-32598240 on Build GRCh38
rs201881809 Chr.19: 32598037-32598037 on Build GRCh38
rs1030888987 Chr.19: 32598466-32598466 on Build GRCh38
rs776619083 Chr.19: 32598320-32598320 on Build GRCh38
rs752776051 Chr.19: 32598175-32598175 on Build GRCh38
rs770088624 Chr.19: 32598390-32598390 on Build GRCh38
rs768264042 Chr.19: 32598287-32598287 on Build GRCh38
rs200475662 Chr.19: 32598353-32598353 on Build GRCh38
rs769669461 Chr.19: 32598409-32598409 on Build GRCh38
rs191023196 Chr.19: 32598404-32598404 on Build GRCh38
rs763751965 Chr.19: 32598144-32598144 on Build GRCh38
rs115207613 Chr.19: 32598410-32598410 on Build GRCh38
rs764358078 Chr.19: 32598255-32598255 on Build GRCh38
rs746193393 Chr.19: 32598273-32598273 on Build GRCh38
rs760990041 Chr.19: 32598425-32598425 on Build GRCh38
rs751760120 Chr.19: 32598259-32598259 on Build GRCh38
rs770293349 Chr.19: 32598218-32598218 on Build GRCh38
rs566768915 Chr.19: 32598081-32598081 on Build GRCh38
rs765267249 Chr.19: 32598174-32598174 on Build GRCh38
rs775688641 Chr.19: 32598417-32598418 on Build GRCh38
rs775248678 Chr.19: 32598133-32598133 on Build GRCh38
rs754343994 Chr.19: 32598376-32598376 on Build GRCh38
rs750368832 Chr.19: 32598169-32598169 on Build GRCh38
rs558664447 Chr.19: 32598129-32598129 on Build GRCh38
rs897582168 Chr.19: 32598043-32598043 on Build GRCh38
rs751148398 Chr.19: 32598121-32598121 on Build GRCh38
rs749061431 Chr.19: 32598132-32598134 on Build GRCh38
rs148375756 Chr.19: 32598303-32598303 on Build GRCh38
rs778592813 Chr.19: 32598391-32598391 on Build GRCh38
rs560378357 Chr.19: 32598017-32598017 on Build GRCh38
rs771085324 Chr.19: 32598269-32598270 on Build GRCh38
rs756494193 Chr.19: 32598374-32598374 on Build GRCh38
rs201922550 Chr.19: 32598331-32598331 on Build GRCh38
rs1002545905 Chr.19: 32598067-32598067 on Build GRCh38
rs560316864 Chr.19: 32598432-32598432 on Build GRCh38
rs758421939 Chr.19: 32598097-32598098 on Build GRCh38
rs866581092 Chr.19: 32598197-32598197 on Build GRCh38
rs774137684 Chr.19: 32598230-32598230 on Build GRCh38
rs767790921 Chr.19: 32598421-32598421 on Build GRCh38
rs1044635399 Chr.19: 32598089-32598089 on Build GRCh38
rs549362560 Chr.19: 32598173-32598173 on Build GRCh38
rs771474510 Chr.19: 32598232-32598232 on Build GRCh38
rs536339670 Chr.19: 32598242-32598242 on Build GRCh38
rs747607160 Chr.19: 32598394-32598394 on Build GRCh38
rs755505771 Chr.19: 32598383-32598383 on Build GRCh38
rs140577382 Chr.19: 32598217-32598217 on Build GRCh38
rs1035963109 Chr.19: 32598021-32598021 on Build GRCh38
rs199575514 Chr.19: 32598343-32598343 on Build GRCh38
rs766731146 Chr.19: 32598426-32598426 on Build GRCh38
rs766285403 Chr.19: 32598250-32598250 on Build GRCh38
rs924691203 Chr.19: 32598050-32598050 on Build GRCh38
rs61747397 Chr.19: 32598346-32598346 on Build GRCh38
rs764258917 Chr.19: 32598340-32598340 on Build GRCh38
rs999374236 Chr.19: 32598442-32598442 on Build GRCh38
rs199586465 Chr.19: 32598105-32598105 on Build GRCh38
rs566980782 Chr.19: 32598266-32598266 on Build GRCh38
rs199836316 Chr.19: 32598403-32598403 on Build GRCh38
rs970082923 Chr.19: 32598274-32598274 on Build GRCh38

View Full Product Details