Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49690379-49690978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM5
Entrez Gene ID
199800
Gene Name
adrenomedullin 5 (putative)
Gene Aliases
AM5, C19orf76
UniGene
Hs.311193
Gene Chromosome Location
Chr.19: 49688685-49690990 on Build GRCh38
Transcripts
NM_001101340.1 XM_011526438.2 XM_011526439.2

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
UniGene
Hs.112195
Gene Chromosome Location
Chr.19: 49690672-49713731 on Build GRCh38
Transcripts
NM_001101340.1 XM_011526438.2 XM_011526439.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1395309 c.450G>A p.P150P Chr.19: 49690481-49690481 on Build GRCh38
4650513 c.368C>A p.P123H Chr.19: 49690399-49690399 on Build GRCh38
293515 c.446delC p.P150fs*>4 Chr.19: 49690477-49690477 on Build GRCh38
6233305 c.430G>A p.V144I Chr.19: 49690461-49690461 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1006778291 Chr.19: 49690549-49690549 on Build GRCh38
rs868784646 Chr.19: 49690835-49690835 on Build GRCh38
rs45612533 Chr.19: 49690625-49690625 on Build GRCh38
rs763422570 Chr.19: 49690429-49690429 on Build GRCh38
rs569366603 Chr.19: 49690460-49690460 on Build GRCh38
rs998023739 Chr.19: 49690792-49690792 on Build GRCh38
rs557607237 Chr.19: 49690451-49690451 on Build GRCh38
rs934287214 Chr.19: 49690734-49690734 on Build GRCh38
rs1010641645 Chr.19: 49690546-49690546 on Build GRCh38
rs914086484 Chr.19: 49690771-49690771 on Build GRCh38
rs892618539 Chr.19: 49690541-49690541 on Build GRCh38
rs544988918 Chr.19: 49690790-49690790 on Build GRCh38
rs1043127694 Chr.19: 49690727-49690727 on Build GRCh38
rs530620873 Chr.19: 49690825-49690825 on Build GRCh38
rs1002354651 Chr.19: 49690828-49690828 on Build GRCh38
rs1016744032 Chr.19: 49690826-49690826 on Build GRCh38
rs1026210247 Chr.19: 49690912-49690912 on Build GRCh38
rs1018592230 Chr.19: 49690554-49690554 on Build GRCh38
rs1052624726 Chr.19: 49690752-49690752 on Build GRCh38
rs561088374 Chr.19: 49690876-49690876 on Build GRCh38
rs904876641 Chr.19: 49690586-49690586 on Build GRCh38
rs971956012 Chr.19: 49690631-49690631 on Build GRCh38
rs1030430824 Chr.19: 49690617-49690617 on Build GRCh38
rs750860987 Chr.19: 49690428-49690428 on Build GRCh38
rs1044310127 Chr.19: 49690412-49690412 on Build GRCh38
rs913372810 Chr.19: 49690893-49690893 on Build GRCh38
rs760940091 Chr.19: 49690692-49690692 on Build GRCh38
rs1005313796 Chr.19: 49690633-49690633 on Build GRCh38
rs1034572766 Chr.19: 49690879-49690879 on Build GRCh38
rs967934325 Chr.19: 49690944-49690944 on Build GRCh38
rs555506411 Chr.19: 49690560-49690560 on Build GRCh38
rs1052595048 Chr.19: 49690538-49690538 on Build GRCh38
rs185092749 Chr.19: 49690528-49690528 on Build GRCh38
rs912984996 Chr.19: 49690693-49690693 on Build GRCh38
rs771923362 Chr.19: 49690640-49690640 on Build GRCh38
rs998014186 Chr.19: 49690600-49690600 on Build GRCh38
rs571023491 Chr.19: 49690685-49690685 on Build GRCh38
rs963695414 Chr.19: 49690658-49690658 on Build GRCh38
rs751277553 Chr.19: 49690457-49690457 on Build GRCh38
rs573769801 Chr.19: 49690621-49690621 on Build GRCh38
rs1026180515 Chr.19: 49690681-49690681 on Build GRCh38
rs942692528 Chr.19: 49690403-49690403 on Build GRCh38
rs984221074 Chr.19: 49690691-49690691 on Build GRCh38
rs942340755 Chr.19: 49690740-49690740 on Build GRCh38
rs1040359993 Chr.19: 49690435-49690435 on Build GRCh38
rs542408081 Chr.19: 49690874-49690874 on Build GRCh38
rs764323154 Chr.19: 49690720-49690720 on Build GRCh38
rs552672042 Chr.19: 49690666-49690666 on Build GRCh38
rs1039680265 Chr.19: 49690787-49690787 on Build GRCh38
rs927547920 Chr.19: 49690671-49690671 on Build GRCh38
rs960240186 Chr.19: 49690883-49690883 on Build GRCh38
rs922594034 Chr.19: 49690723-49690723 on Build GRCh38
rs117886565 Chr.19: 49690810-49690810 on Build GRCh38
rs945710239 Chr.19: 49690513-49690513 on Build GRCh38
rs74499447 Chr.19: 49690514-49690514 on Build GRCh38
rs780765086 Chr.19: 49690512-49690512 on Build GRCh38
rs1016240215 Chr.19: 49690645-49690645 on Build GRCh38
rs750994858 Chr.19: 49690886-49690886 on Build GRCh38
rs45492293 Chr.19: 49690690-49690690 on Build GRCh38
rs552855692 Chr.19: 49690575-49690575 on Build GRCh38
rs776677807 Chr.19: 49690729-49690729 on Build GRCh38
rs938836119 Chr.19: 49690437-49690437 on Build GRCh38
rs886539285 Chr.19: 49690824-49690824 on Build GRCh38
rs951860496 Chr.19: 49690921-49690921 on Build GRCh38
rs1056850540 Chr.19: 49690487-49690487 on Build GRCh38
rs765174956 Chr.19: 49690850-49690850 on Build GRCh38
rs945791426 Chr.19: 49690696-49690696 on Build GRCh38
rs978556541 Chr.19: 49690958-49690958 on Build GRCh38

View Full Product Details