Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 72760357-72760794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3
Entrez Gene ID
22894
Gene Name
DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
Gene Aliases
2810028N01Rik, EXOSC11, KIAA1008, RRP44, dis3p
UniGene
Hs.744104
Gene Chromosome Location
Chr.13: 72755402-72782128 on Build GRCh38
Transcripts
NM_001128226.2 NM_001322348.1 NM_001322349.1 NM_014953.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6401744 c.2690A>G p.E897G Chr.13: 72760632-72760632 on Build GRCh38
5021145 c.2735T>C p.M912T Chr.13: 72760587-72760587 on Build GRCh38
3999118 c.2757A>G p.Q919Q Chr.13: 72760565-72760565 on Build GRCh38
3469575 c.2733C>T p.I911I Chr.13: 72760589-72760589 on Build GRCh38
5745184 c.2793+2T>C p.? Chr.13: 72760527-72760527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773944481 Chr.13: 72760685-72760685 on Build GRCh38
rs750764460 Chr.13: 72760553-72760553 on Build GRCh38
rs3759418 Chr.13: 72760713-72760713 on Build GRCh38
rs182271579 Chr.13: 72760524-72760524 on Build GRCh38
rs1031858063 Chr.13: 72760475-72760475 on Build GRCh38
rs995873304 Chr.13: 72760473-72760473 on Build GRCh38
rs999474730 Chr.13: 72760460-72760460 on Build GRCh38
rs1042840108 Chr.13: 72760456-72760456 on Build GRCh38
rs762557093 Chr.13: 72760551-72760551 on Build GRCh38
rs754234020 Chr.13: 72760578-72760578 on Build GRCh38
rs756618632 Chr.13: 72760478-72760478 on Build GRCh38
rs756498907 Chr.13: 72760570-72760570 on Build GRCh38
rs1016853796 Chr.13: 72760556-72760556 on Build GRCh38
rs759171810 Chr.13: 72760596-72760601 on Build GRCh38
rs779087641 Chr.13: 72760607-72760607 on Build GRCh38
rs752210696 Chr.13: 72760544-72760544 on Build GRCh38
rs773092927 Chr.13: 72760509-72760509 on Build GRCh38
rs73215082 Chr.13: 72760757-72760757 on Build GRCh38
rs772453417 Chr.13: 72760505-72760505 on Build GRCh38
rs184942401 Chr.13: 72760646-72760646 on Build GRCh38
rs780675113 Chr.13: 72760419-72760419 on Build GRCh38
rs759408672 Chr.13: 72760664-72760664 on Build GRCh38
rs780594935 Chr.13: 72760571-72760571 on Build GRCh38
rs544937159 Chr.13: 72760433-72760433 on Build GRCh38
rs767088806 Chr.13: 72760750-72760750 on Build GRCh38
rs764672135 Chr.13: 72760539-72760539 on Build GRCh38
rs760436588 Chr.13: 72760513-72760513 on Build GRCh38
rs140769423 Chr.13: 72760748-72760748 on Build GRCh38
rs755557027 Chr.13: 72760589-72760589 on Build GRCh38
rs768913294 Chr.13: 72760501-72760501 on Build GRCh38
rs377414126 Chr.13: 72760564-72760564 on Build GRCh38
rs770871664 Chr.13: 72760517-72760517 on Build GRCh38
rs575285131 Chr.13: 72760552-72760552 on Build GRCh38
rs761265345 Chr.13: 72760461-72760461 on Build GRCh38
rs770697185 Chr.13: 72760645-72760645 on Build GRCh38
rs1043415207 Chr.13: 72760770-72760770 on Build GRCh38
rs763726026 Chr.13: 72760675-72760675 on Build GRCh38
rs766192942 Chr.13: 72760574-72760574 on Build GRCh38
rs895942757 Chr.13: 72760486-72760486 on Build GRCh38
rs768988714 Chr.13: 72760500-72760500 on Build GRCh38
rs755586453 Chr.13: 72760664-72760664 on Build GRCh38
rs746804740 Chr.13: 72760644-72760644 on Build GRCh38
rs749379511 Chr.13: 72760499-72760499 on Build GRCh38
rs200025659 Chr.13: 72760626-72760626 on Build GRCh38
rs780461787 Chr.13: 72760497-72760497 on Build GRCh38
rs761533329 Chr.13: 72760696-72760696 on Build GRCh38
rs933843244 Chr.13: 72760622-72760622 on Build GRCh38
rs142922683 Chr.13: 72760667-72760667 on Build GRCh38
rs770572163 Chr.13: 72760498-72760499 on Build GRCh38
rs375074029 Chr.13: 72760536-72760536 on Build GRCh38
rs904781962 Chr.13: 72760438-72760438 on Build GRCh38
rs201178350 Chr.13: 72760503-72760503 on Build GRCh38
rs961598219 Chr.13: 72760588-72760588 on Build GRCh38
rs367680218 Chr.13: 72760671-72760671 on Build GRCh38
rs766597422 Chr.13: 72760701-72760701 on Build GRCh38
rs769742236 Chr.13: 72760666-72760666 on Build GRCh38
rs776501208 Chr.13: 72760518-72760518 on Build GRCh38
rs891669521 Chr.13: 72760431-72760431 on Build GRCh38
rs540000925 Chr.13: 72760623-72760623 on Build GRCh38
rs961320421 Chr.13: 72760587-72760587 on Build GRCh38
rs748269735 Chr.13: 72760616-72760616 on Build GRCh38
rs1015764647 Chr.13: 72760557-72760557 on Build GRCh38
rs947116155 Chr.13: 72760454-72760454 on Build GRCh38
rs773933088 Chr.13: 72760499-72760501 on Build GRCh38
rs974546124 Chr.13: 72760592-72760592 on Build GRCh38

View Full Product Details