Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130450022-130450523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASS1
Entrez Gene ID
445
Gene Name
argininosuccinate synthase 1
Gene Aliases
ASS, CTLN1
UniGene
Hs.160786
Gene Chromosome Location
Chr.9: 130444707-130501274 on Build GRCh38
Transcripts
NM_000050.4 NM_054012.3 XM_005272200.3 XM_011518705.2 XM_017014729.1

SNP Detail

SNP ID Chromosome Location
rs537034604 Chr.9: 130450406-130450406 on Build GRCh38
rs891087400 Chr.9: 130450449-130450449 on Build GRCh38
rs575191736 Chr.9: 130450306-130450306 on Build GRCh38
rs757321147 Chr.9: 130450501-130450501 on Build GRCh38
rs866262416 Chr.9: 130450248-130450248 on Build GRCh38
rs753782147 Chr.9: 130450149-130450149 on Build GRCh38
rs995658510 Chr.9: 130450484-130450484 on Build GRCh38
rs1004627104 Chr.9: 130450455-130450455 on Build GRCh38
rs974270619 Chr.9: 130450242-130450242 on Build GRCh38
rs140853298 Chr.9: 130450257-130450257 on Build GRCh38
rs920623587 Chr.9: 130450232-130450232 on Build GRCh38
rs892948313 Chr.9: 130450050-130450050 on Build GRCh38
rs4740158 Chr.9: 130450085-130450085 on Build GRCh38
rs1015835982 Chr.9: 130450481-130450481 on Build GRCh38
rs562462987 Chr.9: 130450046-130450046 on Build GRCh38
rs891060988 Chr.9: 130450418-130450418 on Build GRCh38
rs996784512 Chr.9: 130450088-130450088 on Build GRCh38
rs1051142587 Chr.9: 130450439-130450439 on Build GRCh38
rs964795510 Chr.9: 130450171-130450171 on Build GRCh38
rs779206869 Chr.9: 130450108-130450108 on Build GRCh38
rs903528557 Chr.9: 130450368-130450368 on Build GRCh38
rs184403245 Chr.9: 130450320-130450320 on Build GRCh38
rs537889984 Chr.9: 130450327-130450327 on Build GRCh38
rs898784451 Chr.9: 130450482-130450482 on Build GRCh38
rs947214966 Chr.9: 130450388-130450388 on Build GRCh38
rs145212223 Chr.9: 130450307-130450307 on Build GRCh38
rs952101534 Chr.9: 130450276-130450276 on Build GRCh38
rs748675185 Chr.9: 130450195-130450195 on Build GRCh38
rs549216391 Chr.9: 130450309-130450309 on Build GRCh38
rs1011827928 Chr.9: 130450051-130450051 on Build GRCh38
rs932707973 Chr.9: 130450429-130450429 on Build GRCh38
rs1043426640 Chr.9: 130450061-130450061 on Build GRCh38
rs181409069 Chr.9: 130450196-130450196 on Build GRCh38
rs955569922 Chr.9: 130450130-130450130 on Build GRCh38
rs75335697 Chr.9: 130450371-130450371 on Build GRCh38
rs1022721250 Chr.9: 130450275-130450275 on Build GRCh38
rs1043757348 Chr.9: 130450052-130450052 on Build GRCh38
rs1018077926 Chr.9: 130450463-130450463 on Build GRCh38
rs866436174 Chr.9: 130450425-130450425 on Build GRCh38
rs1023169563 Chr.9: 130450493-130450493 on Build GRCh38
rs12004384 Chr.9: 130450341-130450341 on Build GRCh38
rs367775083 Chr.9: 130450062-130450062 on Build GRCh38
rs1056668739 Chr.9: 130450319-130450319 on Build GRCh38
rs375022559 Chr.9: 130450116-130450116 on Build GRCh38
rs1008055758 Chr.9: 130450112-130450113 on Build GRCh38
rs575951958 Chr.9: 130450049-130450049 on Build GRCh38
rs1043829391 Chr.9: 130450363-130450363 on Build GRCh38
rs533545223 Chr.9: 130450169-130450169 on Build GRCh38

View Full Product Details