Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15604349-15604933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL5
Entrez Gene ID
26234
Gene Name
F-box and leucine rich repeat protein 5
Gene Aliases
FBL4, FBL5, FLR1
UniGene
Hs.643433
Gene Chromosome Location
Chr.4: 15604384-15681545 on Build GRCh38
Transcripts
NM_001193534.1 NM_001193535.1 NM_012161.3 NR_036464.1

SNP Detail

SNP ID Chromosome Location
rs916401515 Chr.4: 15604657-15604657 on Build GRCh38
rs763653371 Chr.4: 15604541-15604542 on Build GRCh38
rs527424043 Chr.4: 15604785-15604785 on Build GRCh38
rs947879110 Chr.4: 15604743-15604743 on Build GRCh38
rs73239167 Chr.4: 15604400-15604400 on Build GRCh38
rs893663843 Chr.4: 15604814-15604814 on Build GRCh38
rs894845064 Chr.4: 15604484-15604484 on Build GRCh38
rs916235722 Chr.4: 15604742-15604742 on Build GRCh38
rs935218469 Chr.4: 15604768-15604768 on Build GRCh38
rs77079212 Chr.4: 15604797-15604797 on Build GRCh38
rs999292553 Chr.4: 15604545-15604545 on Build GRCh38
rs754602303 Chr.4: 15604559-15604559 on Build GRCh38
rs571653978 Chr.4: 15604491-15604491 on Build GRCh38
rs576554381 Chr.4: 15604722-15604722 on Build GRCh38
rs1054760826 Chr.4: 15604438-15604438 on Build GRCh38
rs542940123 Chr.4: 15604653-15604653 on Build GRCh38
rs374614879 Chr.4: 15604879-15604879 on Build GRCh38
rs1055319795 Chr.4: 15604727-15604727 on Build GRCh38
rs748050858 Chr.4: 15604593-15604593 on Build GRCh38
rs1008336340 Chr.4: 15604818-15604818 on Build GRCh38
rs941784332 Chr.4: 15604405-15604405 on Build GRCh38
rs974734263 Chr.4: 15604511-15604511 on Build GRCh38
rs1032150751 Chr.4: 15604547-15604547 on Build GRCh38
rs772818017 Chr.4: 15604642-15604642 on Build GRCh38
rs547328672 Chr.4: 15604503-15604503 on Build GRCh38
rs941788577 Chr.4: 15604716-15604716 on Build GRCh38
rs564955147 Chr.4: 15604782-15604782 on Build GRCh38
rs951627129 Chr.4: 15604533-15604533 on Build GRCh38
rs558202805 Chr.4: 15604486-15604486 on Build GRCh38
rs902337862 Chr.4: 15604537-15604537 on Build GRCh38
rs183153441 Chr.4: 15604815-15604815 on Build GRCh38
rs779424047 Chr.4: 15604856-15604856 on Build GRCh38
rs569865668 Chr.4: 15604450-15604450 on Build GRCh38
rs1043942390 Chr.4: 15604756-15604756 on Build GRCh38
rs778972837 Chr.4: 15604588-15604588 on Build GRCh38
rs1045715248 Chr.4: 15604449-15604450 on Build GRCh38
rs10303 Chr.4: 15604637-15604637 on Build GRCh38
rs918817605 Chr.4: 15604622-15604622 on Build GRCh38
rs191395223 Chr.4: 15604699-15604699 on Build GRCh38
rs149522678 Chr.4: 15604634-15604634 on Build GRCh38
rs28676493 Chr.4: 15604847-15604847 on Build GRCh38
rs116831681 Chr.4: 15604574-15604574 on Build GRCh38
rs753041581 Chr.4: 15604529-15604530 on Build GRCh38
rs370971976 Chr.4: 15604826-15604826 on Build GRCh38
rs781282876 Chr.4: 15604381-15604381 on Build GRCh38
rs199756293 Chr.4: 15604530-15604530 on Build GRCh38
rs1018267096 Chr.4: 15604587-15604587 on Build GRCh38
rs1048611725 Chr.4: 15604398-15604398 on Build GRCh38
rs902411326 Chr.4: 15604784-15604784 on Build GRCh38
rs143051099 Chr.4: 15604434-15604434 on Build GRCh38
rs78255455 Chr.4: 15604766-15604766 on Build GRCh38
rs563204468 Chr.4: 15604615-15604616 on Build GRCh38
rs984256770 Chr.4: 15604688-15604688 on Build GRCh38
rs10214 Chr.4: 15604808-15604808 on Build GRCh38
rs1018872748 Chr.4: 15604470-15604472 on Build GRCh38
rs538470652 Chr.4: 15604482-15604482 on Build GRCh38
rs11338780 Chr.4: 15604530-15604530 on Build GRCh38
rs958074833 Chr.4: 15604560-15604560 on Build GRCh38

View Full Product Details