Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 97765811-97766321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6
Entrez Gene ID
84100
Gene Name
ADP ribosylation factor like GTPase 6
Gene Aliases
BBS3, RP55
UniGene
Hs.373801
Gene Chromosome Location
Chr.3: 97762875-97812585 on Build GRCh38
Transcripts
NR_103511.1 XM_006713783.3 XM_017007311.1 XM_017007312.1 XM_017007313.1

SNP Detail

SNP ID Chromosome Location
rs576058272 Chr.3: 97766013-97766013 on Build GRCh38
rs935026780 Chr.3: 97766090-97766090 on Build GRCh38
rs912099075 Chr.3: 97766011-97766011 on Build GRCh38
rs372695040 Chr.3: 97766129-97766129 on Build GRCh38
rs975061545 Chr.3: 97765936-97765936 on Build GRCh38
rs1043141732 Chr.3: 97765877-97765877 on Build GRCh38
rs893073017 Chr.3: 97766179-97766181 on Build GRCh38
rs960048168 Chr.3: 97765943-97765943 on Build GRCh38
rs994058363 Chr.3: 97765978-97765978 on Build GRCh38
rs762368870 Chr.3: 97766289-97766289 on Build GRCh38
rs955208428 Chr.3: 97766242-97766242 on Build GRCh38
rs983994814 Chr.3: 97766009-97766009 on Build GRCh38
rs972334225 Chr.3: 97766051-97766051 on Build GRCh38
rs894574930 Chr.3: 97765839-97765839 on Build GRCh38
rs1001632567 Chr.3: 97765899-97765899 on Build GRCh38
rs142961263 Chr.3: 97765930-97765930 on Build GRCh38
rs952505968 Chr.3: 97766006-97766006 on Build GRCh38
rs962521199 Chr.3: 97766047-97766047 on Build GRCh38
rs542933306 Chr.3: 97765881-97765881 on Build GRCh38
rs539427414 Chr.3: 97766182-97766182 on Build GRCh38
rs1057233002 Chr.3: 97766164-97766164 on Build GRCh38
rs764097936 Chr.3: 97766034-97766034 on Build GRCh38
rs569831661 Chr.3: 97765983-97765983 on Build GRCh38
rs957058050 Chr.3: 97765932-97765932 on Build GRCh38
rs920837165 Chr.3: 97766086-97766086 on Build GRCh38
rs976546381 Chr.3: 97766065-97766065 on Build GRCh38
rs201662985 Chr.3: 97765907-97765907 on Build GRCh38
rs998816027 Chr.3: 97765922-97765922 on Build GRCh38
rs370610677 Chr.3: 97766283-97766283 on Build GRCh38
rs1002003102 Chr.3: 97766163-97766163 on Build GRCh38
rs556492608 Chr.3: 97766244-97766244 on Build GRCh38
rs147462580 Chr.3: 97766159-97766159 on Build GRCh38
rs1053279715 Chr.3: 97766100-97766100 on Build GRCh38
rs1506445 Chr.3: 97765869-97765869 on Build GRCh38
rs3806612 Chr.3: 97766169-97766169 on Build GRCh38
rs548784307 Chr.3: 97766107-97766107 on Build GRCh38
rs534070732 Chr.3: 97766144-97766144 on Build GRCh38
rs576679778 Chr.3: 97766275-97766275 on Build GRCh38
rs376935720 Chr.3: 97766183-97766183 on Build GRCh38
rs986923110 Chr.3: 97766008-97766008 on Build GRCh38
rs913523479 Chr.3: 97766106-97766106 on Build GRCh38
rs1054328198 Chr.3: 97766172-97766172 on Build GRCh38
rs1018499393 Chr.3: 97766269-97766269 on Build GRCh38
rs865803400 Chr.3: 97765889-97765889 on Build GRCh38
rs529160570 Chr.3: 97766019-97766019 on Build GRCh38
rs963922850 Chr.3: 97766285-97766285 on Build GRCh38
rs922232804 Chr.3: 97766085-97766085 on Build GRCh38
rs1005476884 Chr.3: 97766249-97766249 on Build GRCh38
rs950170036 Chr.3: 97766152-97766152 on Build GRCh38
rs1025564749 Chr.3: 97766219-97766219 on Build GRCh38
rs996679976 Chr.3: 97766179-97766179 on Build GRCh38
rs1055820037 Chr.3: 97766137-97766137 on Build GRCh38
rs1015743050 Chr.3: 97766253-97766253 on Build GRCh38

View Full Product Details