Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 71029609-71030197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCK
Entrez Gene ID
1633
Gene Name
deoxycytidine kinase
Gene Aliases
-
UniGene
Hs.709
Gene Chromosome Location
Chr.4: 70993548-71030914 on Build GRCh38
Transcripts
NM_000788.2

SNP Detail

SNP ID Chromosome Location
rs902597426 Chr.4: 71029916-71029916 on Build GRCh38
rs900866564 Chr.4: 71029995-71029995 on Build GRCh38
rs950487737 Chr.4: 71029653-71029653 on Build GRCh38
rs908900341 Chr.4: 71029756-71029756 on Build GRCh38
rs1030205146 Chr.4: 71030083-71030083 on Build GRCh38
rs916844978 Chr.4: 71029870-71029870 on Build GRCh38
rs1009940125 Chr.4: 71029977-71029977 on Build GRCh38
rs536504363 Chr.4: 71030155-71030155 on Build GRCh38
rs539071298 Chr.4: 71029982-71029982 on Build GRCh38
rs199750727 Chr.4: 71030096-71030096 on Build GRCh38
rs181308455 Chr.4: 71029671-71029671 on Build GRCh38
rs1016425253 Chr.4: 71030158-71030158 on Build GRCh38
rs953563440 Chr.4: 71029695-71029696 on Build GRCh38
rs72552079 Chr.4: 71029951-71029951 on Build GRCh38
rs751051297 Chr.4: 71030078-71030078 on Build GRCh38
rs1046586428 Chr.4: 71029910-71029910 on Build GRCh38
rs963558882 Chr.4: 71030163-71030168 on Build GRCh38
rs113605070 Chr.4: 71030063-71030063 on Build GRCh38
rs900419590 Chr.4: 71030005-71030005 on Build GRCh38
rs965072787 Chr.4: 71029639-71029639 on Build GRCh38
rs530782013 Chr.4: 71029717-71029717 on Build GRCh38
rs991449786 Chr.4: 71029854-71029854 on Build GRCh38
rs72552076 Chr.4: 71029709-71029709 on Build GRCh38
rs909475249 Chr.4: 71029755-71029755 on Build GRCh38
rs1005248926 Chr.4: 71030153-71030153 on Build GRCh38
rs972339545 Chr.4: 71029792-71029792 on Build GRCh38
rs937831293 Chr.4: 71029930-71029930 on Build GRCh38
rs942308318 Chr.4: 71029790-71029790 on Build GRCh38
rs72552078 Chr.4: 71029948-71029948 on Build GRCh38
rs976934596 Chr.4: 71029677-71029677 on Build GRCh38
rs115880528 Chr.4: 71029925-71029925 on Build GRCh38
rs1017905970 Chr.4: 71029996-71029996 on Build GRCh38
rs941724104 Chr.4: 71029821-71029821 on Build GRCh38
rs554766435 Chr.4: 71030165-71030165 on Build GRCh38
rs72552080 Chr.4: 71030118-71030118 on Build GRCh38
rs186925584 Chr.4: 71029747-71029747 on Build GRCh38
rs548895796 Chr.4: 71029690-71029690 on Build GRCh38
rs557069570 Chr.4: 71030029-71030029 on Build GRCh38
rs1020853959 Chr.4: 71029981-71029986 on Build GRCh38
rs72552077 Chr.4: 71029923-71029923 on Build GRCh38
rs568776014 Chr.4: 71030039-71030039 on Build GRCh38
rs375322096 Chr.4: 71030113-71030113 on Build GRCh38
rs949542856 Chr.4: 71029889-71029889 on Build GRCh38
rs1004609981 Chr.4: 71030117-71030117 on Build GRCh38
rs762403825 Chr.4: 71029746-71029746 on Build GRCh38

View Full Product Details