Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42411221-42411782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8A1
Entrez Gene ID
10396
Gene Name
ATPase phospholipid transporting 8A1
Gene Aliases
ATPASEII, ATPIA, ATPP2
UniGene
Hs.435052
Gene Chromosome Location
Chr.4: 42408373-42657134 on Build GRCh38
Transcripts
NM_001105529.1 NM_006095.2 XM_005248043.2 XM_011513615.1 XM_011513616.2 XM_011513618.1 XM_017007645.1 XM_017007646.1 XM_017007647.1

SNP Detail

SNP ID Chromosome Location
rs1009104320 Chr.4: 42411693-42411693 on Build GRCh38
rs983317115 Chr.4: 42411745-42411745 on Build GRCh38
rs965029711 Chr.4: 42411381-42411382 on Build GRCh38
rs531550355 Chr.4: 42411460-42411460 on Build GRCh38
rs73235558 Chr.4: 42411711-42411711 on Build GRCh38
rs577062389 Chr.4: 42411360-42411360 on Build GRCh38
rs893988447 Chr.4: 42411341-42411341 on Build GRCh38
rs966922756 Chr.4: 42411349-42411349 on Build GRCh38
rs1034165905 Chr.4: 42411473-42411473 on Build GRCh38
rs1021187849 Chr.4: 42411348-42411349 on Build GRCh38
rs185598471 Chr.4: 42411303-42411303 on Build GRCh38
rs900686580 Chr.4: 42411342-42411342 on Build GRCh38
rs754887760 Chr.4: 42411723-42411723 on Build GRCh38
rs1016218771 Chr.4: 42411443-42411443 on Build GRCh38
rs981317048 Chr.4: 42411527-42411527 on Build GRCh38
rs34850463 Chr.4: 42411740-42411741 on Build GRCh38
rs182431909 Chr.4: 42411692-42411692 on Build GRCh38
rs561976672 Chr.4: 42411689-42411689 on Build GRCh38
rs760959717 Chr.4: 42411440-42411440 on Build GRCh38
rs967160104 Chr.4: 42411516-42411516 on Build GRCh38
rs563902073 Chr.4: 42411326-42411326 on Build GRCh38
rs532269565 Chr.4: 42411746-42411746 on Build GRCh38
rs907820286 Chr.4: 42411479-42411479 on Build GRCh38
rs768245638 Chr.4: 42411274-42411274 on Build GRCh38
rs974861007 Chr.4: 42411396-42411396 on Build GRCh38
rs963229219 Chr.4: 42411446-42411446 on Build GRCh38
rs936974646 Chr.4: 42411595-42411595 on Build GRCh38
rs562945755 Chr.4: 42411709-42411709 on Build GRCh38
rs1052267844 Chr.4: 42411361-42411361 on Build GRCh38
rs190010043 Chr.4: 42411535-42411535 on Build GRCh38
rs1039173824 Chr.4: 42411335-42411335 on Build GRCh38
rs992704848 Chr.4: 42411485-42411485 on Build GRCh38
rs951930590 Chr.4: 42411420-42411420 on Build GRCh38
rs542981073 Chr.4: 42411676-42411676 on Build GRCh38
rs936914036 Chr.4: 42411670-42411670 on Build GRCh38
rs946917331 Chr.4: 42411683-42411683 on Build GRCh38
rs926988206 Chr.4: 42411571-42411571 on Build GRCh38
rs1004836026 Chr.4: 42411405-42411405 on Build GRCh38
rs925523706 Chr.4: 42411321-42411321 on Build GRCh38
rs548582554 Chr.4: 42411646-42411646 on Build GRCh38
rs945682893 Chr.4: 42411292-42411292 on Build GRCh38
rs900511440 Chr.4: 42411712-42411712 on Build GRCh38
rs960127394 Chr.4: 42411478-42411478 on Build GRCh38
rs16854343 Chr.4: 42411727-42411727 on Build GRCh38
rs978385169 Chr.4: 42411590-42411590 on Build GRCh38
rs1045412180 Chr.4: 42411302-42411302 on Build GRCh38
rs1054580842 Chr.4: 42411327-42411327 on Build GRCh38
rs952096663 Chr.4: 42411744-42411744 on Build GRCh38
rs886558240 Chr.4: 42411374-42411374 on Build GRCh38
rs912978876 Chr.4: 42411508-42411508 on Build GRCh38
rs558450448 Chr.4: 42411318-42411318 on Build GRCh38
rs748815395 Chr.4: 42411248-42411248 on Build GRCh38
rs925621957 Chr.4: 42411612-42411612 on Build GRCh38
rs543454440 Chr.4: 42411753-42411753 on Build GRCh38
rs559421014 Chr.4: 42411501-42411501 on Build GRCh38
rs779019821 Chr.4: 42411555-42411555 on Build GRCh38
rs892860924 Chr.4: 42411658-42411658 on Build GRCh38
rs1004927952 Chr.4: 42411755-42411755 on Build GRCh38
rs1054231566 Chr.4: 42411613-42411613 on Build GRCh38

View Full Product Details