Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76002114-76002705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL9
Entrez Gene ID
4283
Gene Name
C-X-C motif chemokine ligand 9
Gene Aliases
CMK, Humig, MIG, SCYB9, crg-10
UniGene
Hs.77367
Gene Chromosome Location
Chr.4: 76001342-76007523 on Build GRCh38
Transcripts
NM_002416.2 NR_125906.1

Gene Symbol
LOC101928809
Entrez Gene ID
101928809
Gene Name
uncharacterized LOC101928809
Gene Aliases
-
UniGene
Hs.637575
Gene Chromosome Location
Chr.4: 75980790-76005942 on Build GRCh38
Transcripts
NM_002416.2 NR_125906.1

SNP Detail

SNP ID Chromosome Location
rs753005874 Chr.4: 76002276-76002276 on Build GRCh38
rs975863110 Chr.4: 76002624-76002624 on Build GRCh38
rs942200640 Chr.4: 76002429-76002429 on Build GRCh38
rs759893960 Chr.4: 76002198-76002198 on Build GRCh38
rs1031549085 Chr.4: 76002265-76002265 on Build GRCh38
rs1027444633 Chr.4: 76002656-76002656 on Build GRCh38
rs1055268580 Chr.4: 76002444-76002444 on Build GRCh38
rs369441150 Chr.4: 76002650-76002650 on Build GRCh38
rs999444753 Chr.4: 76002502-76002502 on Build GRCh38
rs74454210 Chr.4: 76002330-76002330 on Build GRCh38
rs966413089 Chr.4: 76002299-76002299 on Build GRCh38
rs572210393 Chr.4: 76002197-76002197 on Build GRCh38
rs1046449116 Chr.4: 76002493-76002493 on Build GRCh38
rs895236614 Chr.4: 76002464-76002464 on Build GRCh38
rs148944642 Chr.4: 76002315-76002315 on Build GRCh38
rs1044021203 Chr.4: 76002575-76002575 on Build GRCh38
rs909423934 Chr.4: 76002386-76002386 on Build GRCh38
rs1013693410 Chr.4: 76002479-76002479 on Build GRCh38
rs540515956 Chr.4: 76002379-76002379 on Build GRCh38
rs910307249 Chr.4: 76002287-76002287 on Build GRCh38
rs994538608 Chr.4: 76002648-76002648 on Build GRCh38
rs565188736 Chr.4: 76002495-76002495 on Build GRCh38
rs541833050 Chr.4: 76002294-76002294 on Build GRCh38
rs1031560535 Chr.4: 76002523-76002523 on Build GRCh38
rs964889233 Chr.4: 76002611-76002611 on Build GRCh38
rs41484052 Chr.4: 76002193-76002193 on Build GRCh38
rs974759923 Chr.4: 76002300-76002300 on Build GRCh38
rs532549301 Chr.4: 76002645-76002645 on Build GRCh38
rs979548877 Chr.4: 76002323-76002323 on Build GRCh38
rs550697087 Chr.4: 76002646-76002646 on Build GRCh38
rs896747073 Chr.4: 76002488-76002488 on Build GRCh38
rs574382075 Chr.4: 76002217-76002217 on Build GRCh38
rs546265910 Chr.4: 76002331-76002331 on Build GRCh38
rs879786803 Chr.4: 76002538-76002538 on Build GRCh38
rs957283879 Chr.4: 76002542-76002542 on Build GRCh38
rs1027887545 Chr.4: 76002188-76002188 on Build GRCh38
rs1057081515 Chr.4: 76002372-76002372 on Build GRCh38
rs1017462467 Chr.4: 76002602-76002602 on Build GRCh38
rs767663473 Chr.4: 76002212-76002212 on Build GRCh38
rs760955174 Chr.4: 76002314-76002314 on Build GRCh38
rs989968069 Chr.4: 76002286-76002286 on Build GRCh38
rs998764839 Chr.4: 76002228-76002228 on Build GRCh38

View Full Product Details