Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14681257-14681734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTULIN
Entrez Gene ID
90268
Gene Name
OTU deubiquitinase with linear linkage specificity
Gene Aliases
FAM105B, GUM
UniGene
Hs.406335
Gene Chromosome Location
Chr.5: 14660794-14716552 on Build GRCh38
Transcripts
NM_138348.4 XM_011514151.2 XM_011514152.2 XM_011514154.2 XM_017010015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366936 c.345G>T p.Q115H Chr.5: 14681484-14681484 on Build GRCh38
6658044 c.362G>A p.R121Q Chr.5: 14681501-14681501 on Build GRCh38
269128 c.438G>A p.P146P Chr.5: 14681577-14681577 on Build GRCh38
391707 c.440_441insG p.W148fs*31 Chr.5: 14681579-14681580 on Build GRCh38
5402607 c.353C>T p.T118I Chr.5: 14681492-14681492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781200982 Chr.5: 14681367-14681367 on Build GRCh38
rs11953822 Chr.5: 14681602-14681602 on Build GRCh38
rs781683012 Chr.5: 14681650-14681650 on Build GRCh38
rs774554765 Chr.5: 14681596-14681596 on Build GRCh38
rs1039488403 Chr.5: 14681699-14681699 on Build GRCh38
rs780480926 Chr.5: 14681415-14681415 on Build GRCh38
rs773573756 Chr.5: 14681438-14681438 on Build GRCh38
rs754165084 Chr.5: 14681669-14681670 on Build GRCh38
rs937321859 Chr.5: 14681384-14681384 on Build GRCh38
rs147790160 Chr.5: 14681484-14681484 on Build GRCh38
rs375220163 Chr.5: 14681540-14681540 on Build GRCh38
rs756870978 Chr.5: 14681606-14681606 on Build GRCh38
rs757852944 Chr.5: 14681646-14681646 on Build GRCh38
rs367819985 Chr.5: 14681611-14681611 on Build GRCh38
rs1020407759 Chr.5: 14681496-14681496 on Build GRCh38
rs762108193 Chr.5: 14681430-14681430 on Build GRCh38
rs756713494 Chr.5: 14681656-14681656 on Build GRCh38
rs201942769 Chr.5: 14681601-14681601 on Build GRCh38
rs752485795 Chr.5: 14681471-14681471 on Build GRCh38
rs58610082 Chr.5: 14681446-14681446 on Build GRCh38
rs115519180 Chr.5: 14681544-14681544 on Build GRCh38
rs767400148 Chr.5: 14681461-14681461 on Build GRCh38
rs981905489 Chr.5: 14681358-14681358 on Build GRCh38
rs753698273 Chr.5: 14681605-14681605 on Build GRCh38
rs773077455 Chr.5: 14681666-14681666 on Build GRCh38
rs768701532 Chr.5: 14681359-14681359 on Build GRCh38
rs758569042 Chr.5: 14681350-14681350 on Build GRCh38
rs755888615 Chr.5: 14681478-14681478 on Build GRCh38
rs931948491 Chr.5: 14681697-14681697 on Build GRCh38
rs917710256 Chr.5: 14681339-14681339 on Build GRCh38
rs1050498102 Chr.5: 14681698-14681698 on Build GRCh38
rs751757853 Chr.5: 14681448-14681448 on Build GRCh38
rs371959714 Chr.5: 14681578-14681578 on Build GRCh38
rs1047329342 Chr.5: 14681668-14681668 on Build GRCh38
rs745422073 Chr.5: 14681532-14681532 on Build GRCh38
rs899587491 Chr.5: 14681704-14681704 on Build GRCh38
rs1038656189 Chr.5: 14681389-14681389 on Build GRCh38
rs544028294 Chr.5: 14681382-14681382 on Build GRCh38
rs547080675 Chr.5: 14681594-14681594 on Build GRCh38
rs771323670 Chr.5: 14681595-14681595 on Build GRCh38
rs576975020 Chr.5: 14681379-14681379 on Build GRCh38
rs749723052 Chr.5: 14681670-14681670 on Build GRCh38
rs767444462 Chr.5: 14681599-14681599 on Build GRCh38
rs549107191 Chr.5: 14681309-14681309 on Build GRCh38
rs759489559 Chr.5: 14681453-14681453 on Build GRCh38
rs10214386 Chr.5: 14681688-14681688 on Build GRCh38
rs940584420 Chr.5: 14681362-14681362 on Build GRCh38
rs770017442 Chr.5: 14681431-14681431 on Build GRCh38
rs903395664 Chr.5: 14681504-14681504 on Build GRCh38
rs769108997 Chr.5: 14681424-14681424 on Build GRCh38
rs557797037 Chr.5: 14681445-14681445 on Build GRCh38
rs958675266 Chr.5: 14681577-14681577 on Build GRCh38
rs747154736 Chr.5: 14681311-14681311 on Build GRCh38
rs975975881 Chr.5: 14681681-14681681 on Build GRCh38
rs113459239 Chr.5: 14681708-14681708 on Build GRCh38
rs907731094 Chr.5: 14681322-14681322 on Build GRCh38
rs778388838 Chr.5: 14681526-14681526 on Build GRCh38
rs747501936 Chr.5: 14681419-14681419 on Build GRCh38
rs756960841 Chr.5: 14681514-14681514 on Build GRCh38
rs749909461 Chr.5: 14681640-14681640 on Build GRCh38
rs944199863 Chr.5: 14681693-14681693 on Build GRCh38
rs748593845 Chr.5: 14681654-14681654 on Build GRCh38
rs759553465 Chr.5: 14681598-14681598 on Build GRCh38
rs950156142 Chr.5: 14681346-14681347 on Build GRCh38
rs777032858 Chr.5: 14681426-14681426 on Build GRCh38
rs377713255 Chr.5: 14681574-14681574 on Build GRCh38
rs773774176 Chr.5: 14681592-14681592 on Build GRCh38
rs974490802 Chr.5: 14681616-14681616 on Build GRCh38
rs35731374 Chr.5: 14681461-14681462 on Build GRCh38
rs748478710 Chr.5: 14681559-14681559 on Build GRCh38
rs753500942 Chr.5: 14681499-14681499 on Build GRCh38

View Full Product Details