Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183446183-183446560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2AIP
Entrez Gene ID
55602
Gene Name
CDKN2A interacting protein
Gene Aliases
CARF
UniGene
Hs.592508
Gene Chromosome Location
Chr.4: 183444591-183449064 on Build GRCh38
Transcripts
NM_001317343.1 NM_017632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
447654 c.839C>T p.S280L Chr.4: 183446523-183446523 on Build GRCh38
5949970 c.864C>G p.I288M Chr.4: 183446548-183446548 on Build GRCh38
4455484 c.796A>G p.S266G Chr.4: 183446480-183446480 on Build GRCh38
4439065 c.547A>T p.I183F Chr.4: 183446231-183446231 on Build GRCh38
6341299 c.539G>T p.G180V Chr.4: 183446223-183446223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755589619 Chr.4: 183446259-183446259 on Build GRCh38
rs929929895 Chr.4: 183446235-183446235 on Build GRCh38
rs761803248 Chr.4: 183446312-183446312 on Build GRCh38
rs888308083 Chr.4: 183446255-183446255 on Build GRCh38
rs753204795 Chr.4: 183446433-183446433 on Build GRCh38
rs745670845 Chr.4: 183446390-183446390 on Build GRCh38
rs764720312 Chr.4: 183446513-183446513 on Build GRCh38
rs781055322 Chr.4: 183446469-183446469 on Build GRCh38
rs761903332 Chr.4: 183446217-183446217 on Build GRCh38
rs777293151 Chr.4: 183446262-183446262 on Build GRCh38
rs747629501 Chr.4: 183446258-183446258 on Build GRCh38
rs753830347 Chr.4: 183446215-183446215 on Build GRCh38
rs748942087 Chr.4: 183446265-183446265 on Build GRCh38
rs556450977 Chr.4: 183446333-183446333 on Build GRCh38
rs150786153 Chr.4: 183446311-183446311 on Build GRCh38
rs916334668 Chr.4: 183446437-183446437 on Build GRCh38
rs773479173 Chr.4: 183446326-183446326 on Build GRCh38
rs139205806 Chr.4: 183446322-183446322 on Build GRCh38
rs758183982 Chr.4: 183446457-183446457 on Build GRCh38
rs746956972 Chr.4: 183446460-183446460 on Build GRCh38
rs766766778 Chr.4: 183446336-183446336 on Build GRCh38
rs201678930 Chr.4: 183446481-183446481 on Build GRCh38
rs762977550 Chr.4: 183446421-183446421 on Build GRCh38
rs953732365 Chr.4: 183446260-183446260 on Build GRCh38
rs755649490 Chr.4: 183446370-183446370 on Build GRCh38
rs759827030 Chr.4: 183446427-183446427 on Build GRCh38
rs774502460 Chr.4: 183446426-183446426 on Build GRCh38
rs746949845 Chr.4: 183446403-183446403 on Build GRCh38
rs10533201 Chr.4: 183446407-183446409 on Build GRCh38
rs79181273 Chr.4: 183446452-183446452 on Build GRCh38
rs73870525 Chr.4: 183446458-183446458 on Build GRCh38
rs927673768 Chr.4: 183446459-183446459 on Build GRCh38
rs142730545 Chr.4: 183446380-183446380 on Build GRCh38
rs1047001698 Chr.4: 183446474-183446474 on Build GRCh38
rs200822074 Chr.4: 183446482-183446482 on Build GRCh38
rs767923076 Chr.4: 183446431-183446431 on Build GRCh38
rs775355984 Chr.4: 183446401-183446401 on Build GRCh38
rs771934109 Chr.4: 183446298-183446298 on Build GRCh38
rs780386797 Chr.4: 183446233-183446233 on Build GRCh38
rs752112414 Chr.4: 183446244-183446244 on Build GRCh38
rs764824709 Chr.4: 183446525-183446525 on Build GRCh38
rs755325470 Chr.4: 183446355-183446355 on Build GRCh38
rs759793629 Chr.4: 183446483-183446483 on Build GRCh38
rs977726997 Chr.4: 183446332-183446332 on Build GRCh38
rs776451267 Chr.4: 183446405-183446406 on Build GRCh38
rs998186656 Chr.4: 183446310-183446310 on Build GRCh38
rs766607169 Chr.4: 183446425-183446425 on Build GRCh38
rs375534313 Chr.4: 183446375-183446375 on Build GRCh38
rs372176577 Chr.4: 183446315-183446315 on Build GRCh38
rs576415495 Chr.4: 183446372-183446372 on Build GRCh38
rs758829615 Chr.4: 183446231-183446231 on Build GRCh38
rs772933113 Chr.4: 183446208-183446208 on Build GRCh38
rs1018373042 Chr.4: 183446289-183446289 on Build GRCh38
rs148559030 Chr.4: 183446533-183446533 on Build GRCh38
rs774264646 Chr.4: 183446274-183446274 on Build GRCh38
rs763738595 Chr.4: 183446436-183446436 on Build GRCh38
rs775441345 Chr.4: 183446306-183446306 on Build GRCh38
rs773241785 Chr.4: 183446413-183446413 on Build GRCh38
rs201501817 Chr.4: 183446408-183446408 on Build GRCh38
rs770365259 Chr.4: 183446218-183446219 on Build GRCh38
rs374122485 Chr.4: 183446335-183446335 on Build GRCh38
rs756623161 Chr.4: 183446434-183446434 on Build GRCh38
rs771832415 Chr.4: 183446399-183446399 on Build GRCh38
rs753184856 Chr.4: 183446358-183446358 on Build GRCh38
rs141294633 Chr.4: 183446272-183446272 on Build GRCh38
rs148244824 Chr.4: 183446218-183446218 on Build GRCh38
rs376179805 Chr.4: 183446203-183446203 on Build GRCh38
rs143679105 Chr.4: 183446485-183446485 on Build GRCh38
rs202014271 Chr.4: 183446497-183446497 on Build GRCh38
rs767927562 Chr.4: 183446357-183446357 on Build GRCh38
rs147291711 Chr.4: 183446406-183446408 on Build GRCh38
rs866523012 Chr.4: 183446404-183446405 on Build GRCh38
rs199828861 Chr.4: 183446250-183446250 on Build GRCh38
rs750578423 Chr.4: 183446226-183446226 on Build GRCh38
rs926650223 Chr.4: 183446429-183446429 on Build GRCh38
rs200693061 Chr.4: 183446376-183446376 on Build GRCh38
rs778429266 Chr.4: 183446355-183446356 on Build GRCh38
rs751948368 Chr.4: 183446352-183446352 on Build GRCh38
rs367772349 Chr.4: 183446516-183446516 on Build GRCh38
rs901340479 Chr.4: 183446486-183446486 on Build GRCh38
rs564435376 Chr.4: 183446520-183446520 on Build GRCh38
rs750085143 Chr.4: 183446453-183446453 on Build GRCh38
rs145782483 Chr.4: 183446487-183446487 on Build GRCh38
rs759531881 Chr.4: 183446214-183446214 on Build GRCh38
rs768667094 Chr.4: 183446468-183446468 on Build GRCh38
rs749379115 Chr.4: 183446480-183446480 on Build GRCh38
rs745840281 Chr.4: 183446276-183446276 on Build GRCh38
rs760909541 Chr.4: 183446491-183446491 on Build GRCh38
rs899892479 Chr.4: 183446295-183446295 on Build GRCh38
rs764522552 Chr.4: 183446408-183446410 on Build GRCh38

View Full Product Details