Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 13717162-13717697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
UniGene
Hs.212360
Gene Chromosome Location
Chr.5: 13690328-14011829 on Build GRCh38
Transcripts
NM_001369.2 XM_005248262.3 XM_017009177.1 XM_017009178.1 XM_017009179.1 XM_017009180.1 XM_017009185.1 XM_017009186.1 XM_017009188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671511 c.12625C>T p.P4209S Chr.5: 13717395-13717395 on Build GRCh38
1061175 c.12631G>A p.E4211K Chr.5: 13717389-13717389 on Build GRCh38
5356440 c.12608C>T p.A4203V Chr.5: 13717412-13717412 on Build GRCh38
1061176 c.12631G>T p.E4211* Chr.5: 13717389-13717389 on Build GRCh38
331435 c.12617G>T p.W4206L Chr.5: 13717403-13717403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370958379 Chr.5: 13717393-13717393 on Build GRCh38
rs1029078940 Chr.5: 13717607-13717607 on Build GRCh38
rs992629268 Chr.5: 13717465-13717465 on Build GRCh38
rs548589680 Chr.5: 13717273-13717273 on Build GRCh38
rs141559600 Chr.5: 13717266-13717266 on Build GRCh38
rs564134623 Chr.5: 13717326-13717326 on Build GRCh38
rs748903850 Chr.5: 13717416-13717416 on Build GRCh38
rs537158218 Chr.5: 13717211-13717211 on Build GRCh38
rs141993196 Chr.5: 13717461-13717461 on Build GRCh38
rs779993876 Chr.5: 13717314-13717314 on Build GRCh38
rs374874272 Chr.5: 13717425-13717425 on Build GRCh38
rs372037071 Chr.5: 13717539-13717539 on Build GRCh38
rs113533336 Chr.5: 13717588-13717588 on Build GRCh38
rs375694023 Chr.5: 13717424-13717424 on Build GRCh38
rs769834014 Chr.5: 13717546-13717546 on Build GRCh38
rs906933901 Chr.5: 13717188-13717188 on Build GRCh38
rs79185772 Chr.5: 13717406-13717406 on Build GRCh38
rs755584441 Chr.5: 13717482-13717482 on Build GRCh38
rs758172356 Chr.5: 13717305-13717305 on Build GRCh38
rs767521567 Chr.5: 13717184-13717184 on Build GRCh38
rs200564274 Chr.5: 13717276-13717276 on Build GRCh38
rs894186234 Chr.5: 13717457-13717457 on Build GRCh38
rs1022306164 Chr.5: 13717639-13717639 on Build GRCh38
rs758897089 Chr.5: 13717417-13717417 on Build GRCh38
rs752622622 Chr.5: 13717481-13717481 on Build GRCh38
rs762673561 Chr.5: 13717389-13717389 on Build GRCh38
rs765409506 Chr.5: 13717380-13717380 on Build GRCh38
rs952344053 Chr.5: 13717264-13717264 on Build GRCh38
rs150824951 Chr.5: 13717669-13717669 on Build GRCh38
rs375727884 Chr.5: 13717292-13717292 on Build GRCh38
rs200056509 Chr.5: 13717531-13717531 on Build GRCh38
rs148373144 Chr.5: 13717254-13717254 on Build GRCh38
rs146330391 Chr.5: 13717474-13717474 on Build GRCh38
rs368456286 Chr.5: 13717345-13717345 on Build GRCh38
rs766099303 Chr.5: 13717390-13717390 on Build GRCh38
rs555395280 Chr.5: 13717213-13717213 on Build GRCh38
rs572053952 Chr.5: 13717280-13717280 on Build GRCh38
rs768917457 Chr.5: 13717374-13717374 on Build GRCh38
rs200969063 Chr.5: 13717376-13717376 on Build GRCh38
rs953387830 Chr.5: 13717611-13717611 on Build GRCh38
rs773331673 Chr.5: 13717547-13717547 on Build GRCh38
rs374063061 Chr.5: 13717334-13717334 on Build GRCh38
rs777303722 Chr.5: 13717412-13717412 on Build GRCh38
rs794727887 Chr.5: 13717365-13717365 on Build GRCh38
rs747881064 Chr.5: 13717328-13717328 on Build GRCh38
rs760850770 Chr.5: 13717295-13717295 on Build GRCh38
rs2277046 Chr.5: 13717362-13717362 on Build GRCh38
rs897366089 Chr.5: 13717561-13717561 on Build GRCh38
rs56949645 Chr.5: 13717577-13717577 on Build GRCh38
rs773314785 Chr.5: 13717458-13717458 on Build GRCh38
rs771171095 Chr.5: 13717553-13717553 on Build GRCh38
rs990507266 Chr.5: 13717306-13717306 on Build GRCh38
rs368413636 Chr.5: 13717463-13717463 on Build GRCh38
rs755597752 Chr.5: 13717405-13717405 on Build GRCh38
rs186878182 Chr.5: 13717545-13717545 on Build GRCh38
rs1027905118 Chr.5: 13717263-13717263 on Build GRCh38
rs547445590 Chr.5: 13717507-13717507 on Build GRCh38
rs573712461 Chr.5: 13717234-13717234 on Build GRCh38
rs191082214 Chr.5: 13717320-13717320 on Build GRCh38
rs903957388 Chr.5: 13717649-13717649 on Build GRCh38
rs1041529275 Chr.5: 13717370-13717370 on Build GRCh38
rs749548795 Chr.5: 13717552-13717552 on Build GRCh38
rs767375182 Chr.5: 13717480-13717480 on Build GRCh38
rs754177491 Chr.5: 13717302-13717302 on Build GRCh38
rs748817131 Chr.5: 13717462-13717462 on Build GRCh38
rs921893160 Chr.5: 13717300-13717300 on Build GRCh38
rs148123430 Chr.5: 13717497-13717497 on Build GRCh38
rs748199626 Chr.5: 13717446-13717446 on Build GRCh38
rs866782197 Chr.5: 13717477-13717477 on Build GRCh38
rs765130526 Chr.5: 13717304-13717304 on Build GRCh38
rs139501386 Chr.5: 13717450-13717450 on Build GRCh38
rs754490730 Chr.5: 13717322-13717322 on Build GRCh38
rs746241201 Chr.5: 13717335-13717335 on Build GRCh38
rs780718067 Chr.5: 13717398-13717398 on Build GRCh38
rs138788207 Chr.5: 13717454-13717454 on Build GRCh38
rs531483632 Chr.5: 13717330-13717330 on Build GRCh38
rs889974036 Chr.5: 13717612-13717612 on Build GRCh38
rs376439430 Chr.5: 13717498-13717498 on Build GRCh38
rs948460982 Chr.5: 13717219-13717219 on Build GRCh38
rs747409181 Chr.5: 13717518-13717518 on Build GRCh38
rs868560935 Chr.5: 13717537-13717537 on Build GRCh38
rs747568741 Chr.5: 13717439-13717439 on Build GRCh38
rs747309196 Chr.5: 13717504-13717504 on Build GRCh38
rs868612793 Chr.5: 13717395-13717395 on Build GRCh38
rs756891359 Chr.5: 13717468-13717468 on Build GRCh38
rs372829113 Chr.5: 13717487-13717487 on Build GRCh38
rs1006915400 Chr.5: 13717615-13717615 on Build GRCh38
rs370688558 Chr.5: 13717299-13717299 on Build GRCh38
rs886059965 Chr.5: 13717434-13717434 on Build GRCh38
rs771382650 Chr.5: 13717271-13717271 on Build GRCh38
rs769256866 Chr.5: 13717443-13717443 on Build GRCh38
rs772505955 Chr.5: 13717338-13717338 on Build GRCh38
rs751567040 Chr.5: 13717391-13717391 on Build GRCh38
rs372118787 Chr.5: 13717403-13717403 on Build GRCh38
rs895412535 Chr.5: 13717246-13717246 on Build GRCh38
rs756052803 Chr.5: 13717385-13717385 on Build GRCh38
rs897423035 Chr.5: 13717307-13717307 on Build GRCh38
rs776552880 Chr.5: 13717375-13717375 on Build GRCh38
rs1051200747 Chr.5: 13717578-13717578 on Build GRCh38
rs763160068 Chr.5: 13717459-13717459 on Build GRCh38
rs749015987 Chr.5: 13717253-13717253 on Build GRCh38

View Full Product Details