Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7867634-7868104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD3
Entrez Gene ID
79072
Gene Name
FAST kinase domains 3
Gene Aliases
-
UniGene
Hs.653162
Gene Chromosome Location
Chr.5: 7859159-7869037 on Build GRCh38
Transcripts
NM_024091.3 NR_036553.1 NR_073608.1 XM_006714498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
421174 c.388G>A p.A130T Chr.5: 7867696-7867696 on Build GRCh38
6852048 c.432T>A p.D144E Chr.5: 7867652-7867652 on Build GRCh38
4160016 c.75A>T p.L25F Chr.5: 7868009-7868009 on Build GRCh38
4160014 c.77A>C p.K26T Chr.5: 7868007-7868007 on Build GRCh38
3429676 c.159C>T p.F53F Chr.5: 7867925-7867925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036126568 Chr.5: 7868068-7868068 on Build GRCh38
rs761131854 Chr.5: 7868000-7868000 on Build GRCh38
rs751102537 Chr.5: 7868034-7868034 on Build GRCh38
rs143616198 Chr.5: 7867760-7867760 on Build GRCh38
rs375491136 Chr.5: 7867862-7867862 on Build GRCh38
rs144204302 Chr.5: 7867860-7867860 on Build GRCh38
rs775525968 Chr.5: 7867976-7867976 on Build GRCh38
rs765139932 Chr.5: 7868008-7868008 on Build GRCh38
rs138057208 Chr.5: 7867738-7867738 on Build GRCh38
rs770611081 Chr.5: 7867663-7867663 on Build GRCh38
rs141321021 Chr.5: 7867823-7867823 on Build GRCh38
rs149819178 Chr.5: 7867946-7867946 on Build GRCh38
rs748726932 Chr.5: 7867653-7867653 on Build GRCh38
rs767484855 Chr.5: 7867766-7867766 on Build GRCh38
rs747401284 Chr.5: 7867681-7867681 on Build GRCh38
rs763263583 Chr.5: 7867711-7867711 on Build GRCh38
rs868466752 Chr.5: 7867741-7867741 on Build GRCh38
rs762452287 Chr.5: 7867721-7867721 on Build GRCh38
rs778056443 Chr.5: 7868057-7868057 on Build GRCh38
rs150802478 Chr.5: 7867844-7867844 on Build GRCh38
rs756128288 Chr.5: 7867775-7867775 on Build GRCh38
rs764113909 Chr.5: 7867883-7867883 on Build GRCh38
rs756126035 Chr.5: 7867991-7867992 on Build GRCh38
rs751723624 Chr.5: 7867783-7867784 on Build GRCh38
rs754653963 Chr.5: 7868049-7868049 on Build GRCh38
rs1002007394 Chr.5: 7868016-7868016 on Build GRCh38
rs1025134782 Chr.5: 7867837-7867837 on Build GRCh38
rs749876184 Chr.5: 7867953-7867953 on Build GRCh38
rs371427214 Chr.5: 7867923-7867923 on Build GRCh38
rs2966952 Chr.5: 7867917-7867917 on Build GRCh38
rs761060758 Chr.5: 7867939-7867941 on Build GRCh38
rs763134231 Chr.5: 7867828-7867828 on Build GRCh38
rs757795521 Chr.5: 7868022-7868022 on Build GRCh38
rs753721111 Chr.5: 7867884-7867884 on Build GRCh38
rs79525539 Chr.5: 7867806-7867806 on Build GRCh38
rs760533511 Chr.5: 7867992-7867992 on Build GRCh38
rs549311765 Chr.5: 7867759-7867759 on Build GRCh38
rs56856157 Chr.5: 7868033-7868033 on Build GRCh38
rs756768452 Chr.5: 7867892-7867892 on Build GRCh38
rs770362537 Chr.5: 7867818-7867818 on Build GRCh38
rs1051586462 Chr.5: 7867711-7867711 on Build GRCh38
rs774620297 Chr.5: 7867963-7867963 on Build GRCh38
rs767614377 Chr.5: 7867859-7867859 on Build GRCh38
rs756707899 Chr.5: 7867797-7867797 on Build GRCh38
rs767701698 Chr.5: 7867985-7867986 on Build GRCh38
rs755015909 Chr.5: 7867806-7867806 on Build GRCh38
rs768954890 Chr.5: 7867695-7867695 on Build GRCh38
rs200439700 Chr.5: 7867807-7867807 on Build GRCh38
rs766613241 Chr.5: 7867741-7867743 on Build GRCh38
rs773378598 Chr.5: 7867732-7867732 on Build GRCh38
rs978202380 Chr.5: 7867870-7867870 on Build GRCh38
rs896244670 Chr.5: 7868076-7868076 on Build GRCh38
rs368807642 Chr.5: 7867677-7867677 on Build GRCh38
rs967028899 Chr.5: 7867784-7867784 on Build GRCh38
rs777542253 Chr.5: 7867777-7867777 on Build GRCh38
rs144077073 Chr.5: 7867793-7867793 on Build GRCh38
rs761798590 Chr.5: 7868004-7868004 on Build GRCh38
rs759658836 Chr.5: 7867965-7867965 on Build GRCh38
rs369437252 Chr.5: 7867995-7867995 on Build GRCh38
rs774567361 Chr.5: 7867849-7867849 on Build GRCh38
rs778464630 Chr.5: 7867901-7867901 on Build GRCh38
rs376052553 Chr.5: 7867936-7867936 on Build GRCh38
rs777011399 Chr.5: 7867701-7867701 on Build GRCh38
rs778554530 Chr.5: 7867801-7867801 on Build GRCh38
rs1012411193 Chr.5: 7867832-7867832 on Build GRCh38
rs773607911 Chr.5: 7867656-7867656 on Build GRCh38
rs146944469 Chr.5: 7867734-7867734 on Build GRCh38
rs931830327 Chr.5: 7867708-7867708 on Build GRCh38
rs772372098 Chr.5: 7867975-7867975 on Build GRCh38
rs962848866 Chr.5: 7867678-7867678 on Build GRCh38
rs3733782 Chr.5: 7868075-7868075 on Build GRCh38
rs766064274 Chr.5: 7868024-7868024 on Build GRCh38
rs771550627 Chr.5: 7867958-7867958 on Build GRCh38
rs540585802 Chr.5: 7867966-7867966 on Build GRCh38
rs765636666 Chr.5: 7867729-7867729 on Build GRCh38
rs759894376 Chr.5: 7867854-7867854 on Build GRCh38
rs1033440805 Chr.5: 7868045-7868045 on Build GRCh38
rs145797848 Chr.5: 7868014-7868014 on Build GRCh38
rs139472729 Chr.5: 7867855-7867855 on Build GRCh38
rs752706112 Chr.5: 7867988-7867991 on Build GRCh38
rs140095335 Chr.5: 7867921-7867921 on Build GRCh38
rs1039189888 Chr.5: 7867816-7867816 on Build GRCh38
rs1053719628 Chr.5: 7867657-7867657 on Build GRCh38
rs369497874 Chr.5: 7867773-7867773 on Build GRCh38

View Full Product Details