Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43446249-43446791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM267
Entrez Gene ID
64417
Gene Name
transmembrane protein 267
Gene Aliases
C5orf28
UniGene
Hs.732093
Gene Chromosome Location
Chr.5: 43444252-43484660 on Build GRCh38
Transcripts
NM_022483.4 XM_005248333.4 XM_005248335.3 XM_006714486.3 XM_011514074.2 XM_011514075.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
449701 c.458C>A p.T153N Chr.5: 43446412-43446412 on Build GRCh38
738718 c.327C>G p.L109L Chr.5: 43446543-43446543 on Build GRCh38
4501182 c.582C>T p.F194F Chr.5: 43446288-43446288 on Build GRCh38
738719 c.534C>T p.F178F Chr.5: 43446336-43446336 on Build GRCh38
4566783 c.471_472CC>TT p.R158* Chr.5: 43446398-43446399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367731073 Chr.5: 43446578-43446578 on Build GRCh38
rs769587606 Chr.5: 43446431-43446431 on Build GRCh38
rs551239004 Chr.5: 43446630-43446630 on Build GRCh38
rs766682510 Chr.5: 43446593-43446593 on Build GRCh38
rs943386576 Chr.5: 43446577-43446577 on Build GRCh38
rs746207444 Chr.5: 43446280-43446280 on Build GRCh38
rs35426098 Chr.5: 43446538-43446538 on Build GRCh38
rs375909375 Chr.5: 43446575-43446575 on Build GRCh38
rs767920811 Chr.5: 43446462-43446462 on Build GRCh38
rs750875918 Chr.5: 43446397-43446397 on Build GRCh38
rs762743321 Chr.5: 43446357-43446357 on Build GRCh38
rs371437370 Chr.5: 43446546-43446546 on Build GRCh38
rs1053087020 Chr.5: 43446324-43446324 on Build GRCh38
rs777381857 Chr.5: 43446416-43446416 on Build GRCh38
rs749822109 Chr.5: 43446406-43446406 on Build GRCh38
rs762980305 Chr.5: 43446437-43446437 on Build GRCh38
rs373129040 Chr.5: 43446310-43446310 on Build GRCh38
rs774698216 Chr.5: 43446456-43446456 on Build GRCh38
rs540906289 Chr.5: 43446430-43446430 on Build GRCh38
rs746478658 Chr.5: 43446422-43446422 on Build GRCh38
rs377061101 Chr.5: 43446385-43446385 on Build GRCh38
rs541749580 Chr.5: 43446321-43446321 on Build GRCh38
rs751146213 Chr.5: 43446292-43446292 on Build GRCh38
rs780501864 Chr.5: 43446404-43446404 on Build GRCh38
rs780594006 Chr.5: 43446503-43446503 on Build GRCh38
rs555109567 Chr.5: 43446297-43446297 on Build GRCh38
rs780758109 Chr.5: 43446427-43446427 on Build GRCh38
rs901951971 Chr.5: 43446624-43446624 on Build GRCh38
rs1042440487 Chr.5: 43446748-43446748 on Build GRCh38
rs761014978 Chr.5: 43446592-43446592 on Build GRCh38
rs183681009 Chr.5: 43446654-43446654 on Build GRCh38
rs747729445 Chr.5: 43446287-43446287 on Build GRCh38
rs764259047 Chr.5: 43446364-43446364 on Build GRCh38
rs775219655 Chr.5: 43446434-43446434 on Build GRCh38
rs564897515 Chr.5: 43446755-43446755 on Build GRCh38
rs769398385 Chr.5: 43446540-43446540 on Build GRCh38
rs756544607 Chr.5: 43446401-43446401 on Build GRCh38
rs199788584 Chr.5: 43446579-43446579 on Build GRCh38
rs751517883 Chr.5: 43446428-43446428 on Build GRCh38
rs1007527077 Chr.5: 43446300-43446300 on Build GRCh38
rs902683358 Chr.5: 43446753-43446753 on Build GRCh38
rs745945815 Chr.5: 43446465-43446466 on Build GRCh38
rs775319507 Chr.5: 43446513-43446515 on Build GRCh38
rs187882024 Chr.5: 43446533-43446533 on Build GRCh38
rs772122326 Chr.5: 43446466-43446466 on Build GRCh38
rs756352849 Chr.5: 43446496-43446496 on Build GRCh38
rs906049515 Chr.5: 43446448-43446448 on Build GRCh38
rs745544379 Chr.5: 43446539-43446539 on Build GRCh38
rs146367439 Chr.5: 43446504-43446504 on Build GRCh38
rs752633432 Chr.5: 43446333-43446333 on Build GRCh38
rs754282609 Chr.5: 43446595-43446595 on Build GRCh38
rs530596159 Chr.5: 43446766-43446766 on Build GRCh38
rs377043111 Chr.5: 43446476-43446476 on Build GRCh38
rs769002867 Chr.5: 43446413-43446413 on Build GRCh38
rs142187685 Chr.5: 43446451-43446451 on Build GRCh38
rs370051225 Chr.5: 43446398-43446398 on Build GRCh38
rs574967741 Chr.5: 43446315-43446315 on Build GRCh38
rs767721725 Chr.5: 43446583-43446583 on Build GRCh38
rs765221777 Chr.5: 43446328-43446328 on Build GRCh38
rs755932050 Chr.5: 43446453-43446453 on Build GRCh38
rs901920688 Chr.5: 43446373-43446373 on Build GRCh38
rs537974960 Chr.5: 43446284-43446284 on Build GRCh38
rs781542455 Chr.5: 43446386-43446386 on Build GRCh38
rs770431146 Chr.5: 43446522-43446522 on Build GRCh38
rs192398187 Chr.5: 43446541-43446541 on Build GRCh38
rs749029011 Chr.5: 43446626-43446626 on Build GRCh38
rs575644770 Chr.5: 43446283-43446283 on Build GRCh38
rs759308930 Chr.5: 43446323-43446323 on Build GRCh38
rs374248815 Chr.5: 43446532-43446532 on Build GRCh38
rs771342363 Chr.5: 43446348-43446350 on Build GRCh38
rs1026834129 Chr.5: 43446724-43446724 on Build GRCh38
rs746596063 Chr.5: 43446511-43446511 on Build GRCh38
rs756796297 Chr.5: 43446305-43446305 on Build GRCh38
rs749015076 Chr.5: 43446543-43446543 on Build GRCh38
rs773383632 Chr.5: 43446585-43446585 on Build GRCh38
rs751556472 Chr.5: 43446383-43446383 on Build GRCh38

View Full Product Details